Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4654155-4654672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFA9
Entrez Gene ID
4704
Gene Name
NADH:ubiquinone oxidoreductase subunit A9
Gene Aliases
CC6, CI-39k, CI39k, NDUFS2L, SDR22E1
UniGene
Hs.75227
Gene Chromosome Location
Chr.12: 4649098-4687554 on Build GRCh38
Transcripts
NM_005002.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1746997 c.112C>G p.H38D Chr.12: 4654354-4654354 on Build GRCh38
124702 c.220G>C p.G74R Chr.12: 4654462-4654462 on Build GRCh38
7168419 c.101G>T p.R34L Chr.12: 4654343-4654343 on Build GRCh38
3739678 c.51T>C p.R17R Chr.12: 4654293-4654293 on Build GRCh38
4467297 c.147C>T p.S49S Chr.12: 4654389-4654389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761352178 Chr.12: 4654285-4654285 on Build GRCh38
rs764535850 Chr.12: 4654338-4654338 on Build GRCh38
rs746773892 Chr.12: 4654357-4654357 on Build GRCh38
rs1017963381 Chr.12: 4654194-4654194 on Build GRCh38
rs779748295 Chr.12: 4654354-4654354 on Build GRCh38
rs912394102 Chr.12: 4654647-4654647 on Build GRCh38
rs768493880 Chr.12: 4654358-4654358 on Build GRCh38
rs527679033 Chr.12: 4654603-4654603 on Build GRCh38
rs781211782 Chr.12: 4654376-4654376 on Build GRCh38
rs540881913 Chr.12: 4654282-4654282 on Build GRCh38
rs769488091 Chr.12: 4654379-4654379 on Build GRCh38
rs746965071 Chr.12: 4654262-4654262 on Build GRCh38
rs758574474 Chr.12: 4654441-4654441 on Build GRCh38
rs960949361 Chr.12: 4654461-4654461 on Build GRCh38
rs551597883 Chr.12: 4654502-4654502 on Build GRCh38
rs779882427 Chr.12: 4654253-4654253 on Build GRCh38
rs200951099 Chr.12: 4654409-4654409 on Build GRCh38
rs768375490 Chr.12: 4654204-4654204 on Build GRCh38
rs748147127 Chr.12: 4654469-4654469 on Build GRCh38
rs766569587 Chr.12: 4654442-4654442 on Build GRCh38
rs575343474 Chr.12: 4654205-4654205 on Build GRCh38
rs756088751 Chr.12: 4654483-4654483 on Build GRCh38
rs765346077 Chr.12: 4654419-4654419 on Build GRCh38
rs753019597 Chr.12: 4654503-4654503 on Build GRCh38
rs1033044835 Chr.12: 4654252-4654252 on Build GRCh38
rs531865660 Chr.12: 4654427-4654427 on Build GRCh38
rs777180772 Chr.12: 4654488-4654488 on Build GRCh38
rs943920731 Chr.12: 4654241-4654241 on Build GRCh38
rs768601855 Chr.12: 4654266-4654266 on Build GRCh38
rs370239160 Chr.12: 4654382-4654382 on Build GRCh38
rs863224087 Chr.12: 4654424-4654424 on Build GRCh38
rs3832857 Chr.12: 4654618-4654619 on Build GRCh38
rs762646907 Chr.12: 4654277-4654277 on Build GRCh38
rs748781226 Chr.12: 4654492-4654492 on Build GRCh38
rs571586267 Chr.12: 4654570-4654570 on Build GRCh38
rs192359316 Chr.12: 4654632-4654632 on Build GRCh38
rs767821094 Chr.12: 4654329-4654329 on Build GRCh38
rs863224088 Chr.12: 4654445-4654445 on Build GRCh38
rs758620918 Chr.12: 4654348-4654348 on Build GRCh38
rs762655026 Chr.12: 4654318-4654318 on Build GRCh38
rs150945569 Chr.12: 4654393-4654393 on Build GRCh38
rs776307606 Chr.12: 4654274-4654274 on Build GRCh38
rs919692372 Chr.12: 4654616-4654616 on Build GRCh38
rs764587106 Chr.12: 4654388-4654388 on Build GRCh38
rs371538909 Chr.12: 4654342-4654342 on Build GRCh38
rs942049578 Chr.12: 4654531-4654531 on Build GRCh38
rs186636159 Chr.12: 4654202-4654202 on Build GRCh38
rs769063148 Chr.12: 4654514-4654514 on Build GRCh38
rs748095158 Chr.12: 4654377-4654377 on Build GRCh38
rs2302247 Chr.12: 4654183-4654183 on Build GRCh38
rs61909964 Chr.12: 4654238-4654238 on Build GRCh38
rs745501996 Chr.12: 4654509-4654509 on Build GRCh38
rs368824479 Chr.12: 4654447-4654447 on Build GRCh38
rs769578248 Chr.12: 4654292-4654292 on Build GRCh38
rs772826689 Chr.12: 4654381-4654381 on Build GRCh38
rs2302246 Chr.12: 4654234-4654234 on Build GRCh38
rs770501176 Chr.12: 4654500-4654500 on Build GRCh38
rs778957183 Chr.12: 4654343-4654343 on Build GRCh38
rs3217056 Chr.12: 4654615-4654616 on Build GRCh38
rs145275641 Chr.12: 4654384-4654384 on Build GRCh38
rs529503724 Chr.12: 4654330-4654330 on Build GRCh38
rs543849651 Chr.12: 4654206-4654206 on Build GRCh38
rs951103764 Chr.12: 4654619-4654619 on Build GRCh38
rs969066855 Chr.12: 4654592-4654592 on Build GRCh38
rs148447690 Chr.12: 4654300-4654300 on Build GRCh38
rs546449323 Chr.12: 4654612-4654612 on Build GRCh38
rs375524647 Chr.12: 4654385-4654385 on Build GRCh38
rs770577184 Chr.12: 4654383-4654383 on Build GRCh38

View Full Product Details