Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4668225-4668649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFA9
Entrez Gene ID
4704
Gene Name
NADH:ubiquinone oxidoreductase subunit A9
Gene Aliases
CC6, CI-39k, CI39k, NDUFS2L, SDR22E1
UniGene
Hs.75227
Gene Chromosome Location
Chr.12: 4649098-4687554 on Build GRCh38
Transcripts
NM_005002.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6723702 c.702G>A p.K234K Chr.12: 4668503-4668503 on Build GRCh38
6462625 c.670G>A p.G224S Chr.12: 4668471-4668471 on Build GRCh38
6723704 c.698G>A p.W233* Chr.12: 4668499-4668499 on Build GRCh38
5498048 c.656-6C>T p.? Chr.12: 4668451-4668451 on Build GRCh38
1216914 c.664C>T p.R222W Chr.12: 4668465-4668465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752577103 Chr.12: 4668516-4668516 on Build GRCh38
rs915064202 Chr.12: 4668553-4668553 on Build GRCh38
rs138785086 Chr.12: 4668451-4668451 on Build GRCh38
rs749007855 Chr.12: 4668525-4668525 on Build GRCh38
rs976703741 Chr.12: 4668251-4668251 on Build GRCh38
rs902227801 Chr.12: 4668345-4668345 on Build GRCh38
rs941576315 Chr.12: 4668575-4668575 on Build GRCh38
rs967906249 Chr.12: 4668574-4668574 on Build GRCh38
rs533044724 Chr.12: 4668402-4668402 on Build GRCh38
rs775798853 Chr.12: 4668457-4668457 on Build GRCh38
rs747603490 Chr.12: 4668429-4668429 on Build GRCh38
rs143907732 Chr.12: 4668463-4668463 on Build GRCh38
rs932499468 Chr.12: 4668255-4668255 on Build GRCh38
rs564391848 Chr.12: 4668337-4668337 on Build GRCh38
rs375511497 Chr.12: 4668466-4668466 on Build GRCh38
rs373257507 Chr.12: 4668546-4668546 on Build GRCh38
rs780415526 Chr.12: 4668558-4668558 on Build GRCh38
rs896932586 Chr.12: 4668273-4668273 on Build GRCh38
rs192223259 Chr.12: 4668446-4668446 on Build GRCh38
rs200198459 Chr.12: 4668465-4668465 on Build GRCh38
rs966066971 Chr.12: 4668467-4668467 on Build GRCh38
rs770527387 Chr.12: 4668555-4668555 on Build GRCh38
rs139012735 Chr.12: 4668523-4668523 on Build GRCh38
rs542668265 Chr.12: 4668279-4668279 on Build GRCh38
rs369529412 Chr.12: 4668483-4668483 on Build GRCh38
rs755710465 Chr.12: 4668522-4668522 on Build GRCh38
rs945368283 Chr.12: 4668436-4668436 on Build GRCh38
rs55872857 Chr.12: 4668290-4668290 on Build GRCh38
rs747311627 Chr.12: 4668561-4668561 on Build GRCh38
rs1056671519 Chr.12: 4668386-4668386 on Build GRCh38
rs750185999 Chr.12: 4668408-4668408 on Build GRCh38
rs769336263 Chr.12: 4668431-4668431 on Build GRCh38
rs781209611 Chr.12: 4668504-4668504 on Build GRCh38
rs761201299 Chr.12: 4668461-4668461 on Build GRCh38
rs68144469 Chr.12: 4668308-4668308 on Build GRCh38
rs34954412 Chr.12: 4668277-4668278 on Build GRCh38
rs746985978 Chr.12: 4668421-4668421 on Build GRCh38
rs372555061 Chr.12: 4668615-4668616 on Build GRCh38
rs957271840 Chr.12: 4668351-4668351 on Build GRCh38
rs571242168 Chr.12: 4668545-4668545 on Build GRCh38
rs1032930891 Chr.12: 4668322-4668322 on Build GRCh38
rs766311501 Chr.12: 4668485-4668485 on Build GRCh38
rs779980094 Chr.12: 4668420-4668420 on Build GRCh38
rs754616889 Chr.12: 4668365-4668365 on Build GRCh38
rs965993881 Chr.12: 4668248-4668248 on Build GRCh38
rs531316763 Chr.12: 4668490-4668490 on Build GRCh38
rs758276766 Chr.12: 4668409-4668409 on Build GRCh38
rs540543130 Chr.12: 4668432-4668432 on Build GRCh38
rs758519893 Chr.12: 4668478-4668478 on Build GRCh38
rs765224137 Chr.12: 4668474-4668474 on Build GRCh38
rs895383352 Chr.12: 4668396-4668396 on Build GRCh38
rs1005379172 Chr.12: 4668454-4668454 on Build GRCh38
rs146434380 Chr.12: 4668498-4668498 on Build GRCh38
rs750713184 Chr.12: 4668475-4668475 on Build GRCh38
rs757503969 Chr.12: 4668283-4668283 on Build GRCh38
rs187016424 Chr.12: 4668280-4668280 on Build GRCh38
rs184286793 Chr.12: 4668452-4668452 on Build GRCh38
rs978280249 Chr.12: 4668580-4668580 on Build GRCh38
rs985272933 Chr.12: 4668318-4668318 on Build GRCh38
rs922503458 Chr.12: 4668254-4668254 on Build GRCh38
rs761882524 Chr.12: 4668471-4668471 on Build GRCh38
rs778711240 Chr.12: 4668427-4668427 on Build GRCh38
rs749710181 Chr.12: 4668507-4668507 on Build GRCh38

View Full Product Details