Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6461948-6462443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAPBPL
Entrez Gene ID
55080
Gene Name
TAP binding protein like
Gene Aliases
TAPBP-R, TAPBPR
UniGene
Hs.504597
Gene Chromosome Location
Chr.12: 6451655-6472006 on Build GRCh38
Transcripts
NM_018009.4 XM_017019545.1 XM_017019546.1 NM_001297438.1 NM_014231.4 NM_016830.3 NM_199245.2 NR_123717.1

Gene Symbol
VAMP1
Entrez Gene ID
6843
Gene Name
vesicle associated membrane protein 1
Gene Aliases
SPAX1, SYB1, VAMP-1
UniGene
Hs.20021
Gene Chromosome Location
Chr.12: 6462237-6470899 on Build GRCh38
Transcripts
NM_018009.4 XM_017019545.1 XM_017019546.1 NM_001297438.1 NM_014231.4 NM_016830.3 NM_199245.2 NR_123717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1991559 c.1306G>A p.G436R Chr.12: 6462048-6462048 on Build GRCh38
7118264 c.1348G>T p.D450Y Chr.12: 6462090-6462090 on Build GRCh38
942569 c.1379G>A p.R460H Chr.12: 6462121-6462121 on Build GRCh38
6049241 c.1389T>C p.R463R Chr.12: 6462131-6462131 on Build GRCh38
5015069 c.1331A>G p.E444G Chr.12: 6462073-6462073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760929425 Chr.12: 6462184-6462184 on Build GRCh38
rs757058767 Chr.12: 6462199-6462199 on Build GRCh38
rs747950246 Chr.12: 6462010-6462010 on Build GRCh38
rs369660460 Chr.12: 6462107-6462107 on Build GRCh38
rs900415872 Chr.12: 6462400-6462400 on Build GRCh38
rs71584826 Chr.12: 6462128-6462128 on Build GRCh38
rs777594925 Chr.12: 6462108-6462108 on Build GRCh38
rs745380231 Chr.12: 6462050-6462050 on Build GRCh38
rs1010702319 Chr.12: 6462122-6462122 on Build GRCh38
rs200778376 Chr.12: 6462120-6462120 on Build GRCh38
rs1000146139 Chr.12: 6462405-6462405 on Build GRCh38
rs952148697 Chr.12: 6462410-6462410 on Build GRCh38
rs773776530 Chr.12: 6462047-6462047 on Build GRCh38
rs145531892 Chr.12: 6462243-6462243 on Build GRCh38
rs781469049 Chr.12: 6462006-6462006 on Build GRCh38
rs760341891 Chr.12: 6462070-6462070 on Build GRCh38
rs779060711 Chr.12: 6462130-6462130 on Build GRCh38
rs753690548 Chr.12: 6462191-6462191 on Build GRCh38
rs371361079 Chr.12: 6461990-6461990 on Build GRCh38
rs566314360 Chr.12: 6461998-6461998 on Build GRCh38
rs537189889 Chr.12: 6462031-6462031 on Build GRCh38
rs1128466 Chr.12: 6462299-6462299 on Build GRCh38
rs530955951 Chr.12: 6462360-6462360 on Build GRCh38
rs755916605 Chr.12: 6462024-6462024 on Build GRCh38
rs758938038 Chr.12: 6461999-6461999 on Build GRCh38
rs1024377698 Chr.12: 6462331-6462331 on Build GRCh38
rs563759407 Chr.12: 6462272-6462272 on Build GRCh38
rs71584828 Chr.12: 6462181-6462181 on Build GRCh38
rs980151204 Chr.12: 6461977-6461977 on Build GRCh38
rs911660001 Chr.12: 6462217-6462217 on Build GRCh38
rs138172426 Chr.12: 6462066-6462066 on Build GRCh38
rs765643644 Chr.12: 6461995-6461995 on Build GRCh38
rs34434676 Chr.12: 6462125-6462125 on Build GRCh38
rs139613151 Chr.12: 6462131-6462131 on Build GRCh38
rs980507554 Chr.12: 6462340-6462340 on Build GRCh38
rs1010092748 Chr.12: 6462196-6462196 on Build GRCh38
rs770316205 Chr.12: 6462072-6462072 on Build GRCh38
rs753601393 Chr.12: 6462112-6462112 on Build GRCh38
rs541708097 Chr.12: 6462157-6462157 on Build GRCh38
rs543548641 Chr.12: 6462182-6462182 on Build GRCh38
rs199779151 Chr.12: 6462048-6462048 on Build GRCh38
rs747870941 Chr.12: 6462092-6462092 on Build GRCh38
rs141838188 Chr.12: 6462121-6462121 on Build GRCh38
rs937649407 Chr.12: 6462011-6462011 on Build GRCh38
rs1039337190 Chr.12: 6462085-6462085 on Build GRCh38
rs759893027 Chr.12: 6462176-6462176 on Build GRCh38
rs752966254 Chr.12: 6462098-6462098 on Build GRCh38
rs773749610 Chr.12: 6462148-6462148 on Build GRCh38
rs927277424 Chr.12: 6462353-6462353 on Build GRCh38
rs920228805 Chr.12: 6462188-6462188 on Build GRCh38
rs1010255163 Chr.12: 6462419-6462419 on Build GRCh38
rs762227755 Chr.12: 6461994-6461994 on Build GRCh38
rs369831782 Chr.12: 6462002-6462002 on Build GRCh38
rs201607140 Chr.12: 6462158-6462158 on Build GRCh38
rs573060784 Chr.12: 6462143-6462143 on Build GRCh38
rs756207694 Chr.12: 6462198-6462198 on Build GRCh38
rs1037353376 Chr.12: 6462028-6462028 on Build GRCh38
rs112343707 Chr.12: 6462001-6462001 on Build GRCh38
rs575070676 Chr.12: 6462030-6462030 on Build GRCh38
rs768704434 Chr.12: 6462358-6462358 on Build GRCh38
rs767438563 Chr.12: 6462093-6462093 on Build GRCh38
rs139938131 Chr.12: 6462084-6462084 on Build GRCh38
rs764298551 Chr.12: 6462189-6462189 on Build GRCh38
rs891760516 Chr.12: 6462411-6462411 on Build GRCh38
rs552603034 Chr.12: 6461989-6461989 on Build GRCh38
rs1047913916 Chr.12: 6462206-6462206 on Build GRCh38
rs759162597 Chr.12: 6461986-6461986 on Build GRCh38
rs760981246 Chr.12: 6462281-6462281 on Build GRCh38
rs752832207 Chr.12: 6462186-6462186 on Build GRCh38
rs944527698 Chr.12: 6462219-6462219 on Build GRCh38
rs957905284 Chr.12: 6462190-6462190 on Build GRCh38
rs763601458 Chr.12: 6462077-6462077 on Build GRCh38
rs751653118 Chr.12: 6462090-6462090 on Build GRCh38
rs988974975 Chr.12: 6461980-6461980 on Build GRCh38
rs774935900 Chr.12: 6462162-6462162 on Build GRCh38
rs570055804 Chr.12: 6462113-6462113 on Build GRCh38
rs780541469 Chr.12: 6462067-6462067 on Build GRCh38
rs971501070 Chr.12: 6462338-6462338 on Build GRCh38
rs145996145 Chr.12: 6462003-6462003 on Build GRCh38
rs770834090 Chr.12: 6462043-6462043 on Build GRCh38
rs1054379721 Chr.12: 6462409-6462409 on Build GRCh38
rs376455133 Chr.12: 6462127-6462127 on Build GRCh38
rs568982467 Chr.12: 6462039-6462039 on Build GRCh38

View Full Product Details