Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15956855-15957357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERA
Entrez Gene ID
51071
Gene Name
deoxyribose-phosphate aldolase
Gene Aliases
CGI-26, DEOC
UniGene
Hs.39429
Gene Chromosome Location
Chr.12: 15911172-16037381 on Build GRCh38
Transcripts
NM_001300779.1 NM_015954.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
937643 c.117A>C p.K39N Chr.12: 15957021-15957021 on Build GRCh38
6223305 c.38G>C p.S13T Chr.12: 15956942-15956942 on Build GRCh38
6205307 c.68C>T p.P23L Chr.12: 15956972-15956972 on Build GRCh38
4677107 c.83G>A p.R28H Chr.12: 15956987-15956987 on Build GRCh38
5037849 c.129+8T>G p.? Chr.12: 15957041-15957041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201344317 Chr.12: 15957015-15957015 on Build GRCh38
rs780659292 Chr.12: 15956898-15956898 on Build GRCh38
rs750926445 Chr.12: 15956959-15956959 on Build GRCh38
rs748102136 Chr.12: 15957078-15957078 on Build GRCh38
rs773504125 Chr.12: 15957027-15957027 on Build GRCh38
rs763017089 Chr.12: 15956935-15956935 on Build GRCh38
rs528030813 Chr.12: 15957021-15957021 on Build GRCh38
rs1041363604 Chr.12: 15957076-15957076 on Build GRCh38
rs780761055 Chr.12: 15956991-15956991 on Build GRCh38
rs758691540 Chr.12: 15957041-15957041 on Build GRCh38
rs995307849 Chr.12: 15957151-15957151 on Build GRCh38
rs750065752 Chr.12: 15957049-15957049 on Build GRCh38
rs1010136944 Chr.12: 15957229-15957229 on Build GRCh38
rs371541567 Chr.12: 15956973-15956973 on Build GRCh38
rs766572270 Chr.12: 15956975-15956975 on Build GRCh38
rs779390653 Chr.12: 15956904-15956904 on Build GRCh38
rs891661075 Chr.12: 15957175-15957175 on Build GRCh38
rs761047506 Chr.12: 15957028-15957028 on Build GRCh38
rs746617704 Chr.12: 15956912-15956912 on Build GRCh38
rs751818859 Chr.12: 15957224-15957224 on Build GRCh38
rs754469182 Chr.12: 15957067-15957067 on Build GRCh38
rs1039454175 Chr.12: 15957095-15957095 on Build GRCh38
rs772006617 Chr.12: 15957024-15957024 on Build GRCh38
rs755816139 Chr.12: 15957056-15957056 on Build GRCh38
rs865813421 Chr.12: 15956958-15956958 on Build GRCh38
rs779403940 Chr.12: 15957058-15957058 on Build GRCh38
rs568016967 Chr.12: 15957298-15957298 on Build GRCh38
rs757980392 Chr.12: 15957035-15957036 on Build GRCh38
rs117958162 Chr.12: 15957324-15957324 on Build GRCh38
rs1008091590 Chr.12: 15957094-15957094 on Build GRCh38
rs922000014 Chr.12: 15957098-15957098 on Build GRCh38
rs549871116 Chr.12: 15957247-15957247 on Build GRCh38
rs764507137 Chr.12: 15957046-15957046 on Build GRCh38
rs560848992 Chr.12: 15957008-15957008 on Build GRCh38
rs754384751 Chr.12: 15956902-15956902 on Build GRCh38
rs753064457 Chr.12: 15956987-15956987 on Build GRCh38
rs376072127 Chr.12: 15956968-15956968 on Build GRCh38
rs942076775 Chr.12: 15957062-15957062 on Build GRCh38
rs375069071 Chr.12: 15957146-15957146 on Build GRCh38
rs368479950 Chr.12: 15956996-15956996 on Build GRCh38
rs201549411 Chr.12: 15956990-15956990 on Build GRCh38
rs768683510 Chr.12: 15957016-15957016 on Build GRCh38
rs77428417 Chr.12: 15957036-15957036 on Build GRCh38
rs375606723 Chr.12: 15957030-15957030 on Build GRCh38
rs374442153 Chr.12: 15956986-15956986 on Build GRCh38
rs775336699 Chr.12: 15956934-15956934 on Build GRCh38
rs768324553 Chr.12: 15956946-15956946 on Build GRCh38
rs776811322 Chr.12: 15957035-15957035 on Build GRCh38
rs770657016 Chr.12: 15956917-15956917 on Build GRCh38
rs748795653 Chr.12: 15957066-15957066 on Build GRCh38
rs745521985 Chr.12: 15957007-15957007 on Build GRCh38
rs11056740 Chr.12: 15957240-15957240 on Build GRCh38
rs774189773 Chr.12: 15957017-15957017 on Build GRCh38
rs556620793 Chr.12: 15956976-15956976 on Build GRCh38
rs145891923 Chr.12: 15957153-15957153 on Build GRCh38
rs992264498 Chr.12: 15956971-15956971 on Build GRCh38
rs771990304 Chr.12: 15957081-15957081 on Build GRCh38
rs1001172300 Chr.12: 15957270-15957270 on Build GRCh38
rs778853893 Chr.12: 15957077-15957077 on Build GRCh38
rs766580388 Chr.12: 15957141-15957141 on Build GRCh38
rs765725900 Chr.12: 15957037-15957037 on Build GRCh38
rs530126231 Chr.12: 15956926-15956926 on Build GRCh38
rs552448445 Chr.12: 15957325-15957325 on Build GRCh38
rs372692257 Chr.12: 15957248-15957248 on Build GRCh38
rs368086194 Chr.12: 15957040-15957040 on Build GRCh38
rs945677741 Chr.12: 15957055-15957055 on Build GRCh38
rs777724422 Chr.12: 15957307-15957307 on Build GRCh38
rs755555578 Chr.12: 15956982-15956982 on Build GRCh38
rs755232499 Chr.12: 15956903-15956903 on Build GRCh38
rs761913704 Chr.12: 15956961-15956961 on Build GRCh38
rs372595872 Chr.12: 15956942-15956942 on Build GRCh38
rs376410058 Chr.12: 15957102-15957102 on Build GRCh38
rs750696055 Chr.12: 15956972-15956972 on Build GRCh38
rs748027054 Chr.12: 15957282-15957282 on Build GRCh38
rs779720533 Chr.12: 15957056-15957061 on Build GRCh38
rs745330176 Chr.12: 15956927-15956927 on Build GRCh38
rs780823124 Chr.12: 15957267-15957267 on Build GRCh38
rs769304465 Chr.12: 15956931-15956931 on Build GRCh38
rs552899576 Chr.12: 15957105-15957105 on Build GRCh38

View Full Product Details