Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 19439931-19440462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEBP2
Entrez Gene ID
121536
Gene Name
AE binding protein 2
Gene Aliases
-
UniGene
Hs.126497
Gene Chromosome Location
Chr.12: 19404101-19522239 on Build GRCh38
Transcripts
NM_001114176.1 NM_153207.4 XM_006719031.3 XM_011520560.2 XM_017018806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4040686 c.636T>C p.D212D Chr.12: 19440335-19440335 on Build GRCh38
6426894 c.413G>T p.R138L Chr.12: 19440112-19440112 on Build GRCh38
6408788 c.266G>T p.S89I Chr.12: 19439965-19439965 on Build GRCh38
5714948 c.305A>G p.D102G Chr.12: 19440004-19440004 on Build GRCh38
6409131 c.272G>A p.S91N Chr.12: 19439971-19439971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752111783 Chr.12: 19440205-19440205 on Build GRCh38
rs770859882 Chr.12: 19440179-19440179 on Build GRCh38
rs774463361 Chr.12: 19440399-19440399 on Build GRCh38
rs934350168 Chr.12: 19440142-19440142 on Build GRCh38
rs924401332 Chr.12: 19440439-19440439 on Build GRCh38
rs919380121 Chr.12: 19440341-19440341 on Build GRCh38
rs754445302 Chr.12: 19440152-19440152 on Build GRCh38
rs918408344 Chr.12: 19440044-19440044 on Build GRCh38
rs777509556 Chr.12: 19440291-19440291 on Build GRCh38
rs768159445 Chr.12: 19439987-19439987 on Build GRCh38
rs374698851 Chr.12: 19440347-19440347 on Build GRCh38
rs752403929 Chr.12: 19440354-19440354 on Build GRCh38
rs764578035 Chr.12: 19440203-19440203 on Build GRCh38
rs553262655 Chr.12: 19440386-19440386 on Build GRCh38
rs956894611 Chr.12: 19440001-19440001 on Build GRCh38
rs936375319 Chr.12: 19440423-19440423 on Build GRCh38
rs777377666 Chr.12: 19440359-19440359 on Build GRCh38
rs1036469988 Chr.12: 19440028-19440028 on Build GRCh38
rs761201177 Chr.12: 19440002-19440003 on Build GRCh38
rs534661378 Chr.12: 19440402-19440402 on Build GRCh38
rs751515893 Chr.12: 19440373-19440373 on Build GRCh38
rs752248302 Chr.12: 19440414-19440414 on Build GRCh38
rs759181822 Chr.12: 19440349-19440349 on Build GRCh38
rs764498202 Chr.12: 19440109-19440109 on Build GRCh38
rs894364926 Chr.12: 19439991-19440005 on Build GRCh38
rs201164752 Chr.12: 19440405-19440405 on Build GRCh38
rs982678184 Chr.12: 19439966-19439966 on Build GRCh38
rs769969258 Chr.12: 19440300-19440300 on Build GRCh38
rs774223696 Chr.12: 19439988-19439988 on Build GRCh38
rs754672775 Chr.12: 19440237-19440237 on Build GRCh38
rs774679287 Chr.12: 19440317-19440317 on Build GRCh38
rs773053149 Chr.12: 19440171-19440171 on Build GRCh38
rs778471668 Chr.12: 19440239-19440239 on Build GRCh38
rs763494698 Chr.12: 19440419-19440419 on Build GRCh38
rs761668112 Chr.12: 19440409-19440409 on Build GRCh38
rs986396285 Chr.12: 19440236-19440236 on Build GRCh38
rs942067892 Chr.12: 19440022-19440022 on Build GRCh38
rs577707353 Chr.12: 19440422-19440422 on Build GRCh38
rs762346825 Chr.12: 19440326-19440326 on Build GRCh38
rs750843512 Chr.12: 19440344-19440344 on Build GRCh38
rs367737820 Chr.12: 19440392-19440392 on Build GRCh38
rs185496830 Chr.12: 19440158-19440158 on Build GRCh38
rs762874575 Chr.12: 19440308-19440308 on Build GRCh38
rs780082277 Chr.12: 19440397-19440397 on Build GRCh38
rs929830498 Chr.12: 19440046-19440046 on Build GRCh38
rs764726940 Chr.12: 19440353-19440353 on Build GRCh38
rs1000585042 Chr.12: 19439990-19439990 on Build GRCh38
rs757927196 Chr.12: 19440281-19440281 on Build GRCh38
rs1012172849 Chr.12: 19440132-19440132 on Build GRCh38
rs748961386 Chr.12: 19440149-19440149 on Build GRCh38
rs536867351 Chr.12: 19440305-19440305 on Build GRCh38
rs1056381683 Chr.12: 19440095-19440095 on Build GRCh38
rs764841025 Chr.12: 19440134-19440136 on Build GRCh38
rs767899562 Chr.12: 19440221-19440221 on Build GRCh38
rs760970280 Chr.12: 19440406-19440406 on Build GRCh38
rs771958543 Chr.12: 19440170-19440170 on Build GRCh38
rs541690522 Chr.12: 19440428-19440429 on Build GRCh38
rs751929284 Chr.12: 19440125-19440125 on Build GRCh38
rs771648202 Chr.12: 19440417-19440417 on Build GRCh38
rs1043209682 Chr.12: 19440107-19440107 on Build GRCh38
rs779615599 Chr.12: 19440143-19440143 on Build GRCh38
rs976348631 Chr.12: 19440002-19440002 on Build GRCh38
rs892172992 Chr.12: 19440431-19440431 on Build GRCh38
rs760935165 Chr.12: 19440006-19440006 on Build GRCh38
rs763504577 Chr.12: 19440200-19440200 on Build GRCh38
rs781123543 Chr.12: 19440391-19440391 on Build GRCh38
rs909222405 Chr.12: 19440014-19440014 on Build GRCh38
rs757316121 Chr.12: 19440378-19440378 on Build GRCh38
rs775484809 Chr.12: 19440196-19440196 on Build GRCh38
rs951359643 Chr.12: 19440306-19440306 on Build GRCh38
rs551364854 Chr.12: 19440041-19440041 on Build GRCh38
rs753457683 Chr.12: 19440226-19440226 on Build GRCh38
rs759627999 Chr.12: 19440047-19440047 on Build GRCh38
rs757561419 Chr.12: 19440216-19440216 on Build GRCh38
rs915731722 Chr.12: 19440008-19440008 on Build GRCh38
rs924264574 Chr.12: 19440111-19440111 on Build GRCh38
rs758872734 Chr.12: 19440092-19440092 on Build GRCh38
rs768586142 Chr.12: 19440398-19440398 on Build GRCh38
rs776948418 Chr.12: 19440181-19440181 on Build GRCh38
rs370357606 Chr.12: 19440298-19440298 on Build GRCh38
rs757793677 Chr.12: 19440356-19440356 on Build GRCh38
rs765433182 Chr.12: 19440049-19440049 on Build GRCh38
rs747676646 Chr.12: 19440251-19440251 on Build GRCh38
rs181009844 Chr.12: 19440065-19440065 on Build GRCh38
rs144161986 Chr.12: 19440443-19440443 on Build GRCh38
rs755760623 Chr.12: 19440136-19440136 on Build GRCh38
rs531149839 Chr.12: 19440110-19440110 on Build GRCh38
rs986499826 Chr.12: 19440010-19440010 on Build GRCh38
rs746069011 Chr.12: 19440299-19440299 on Build GRCh38
rs746711559 Chr.12: 19440178-19440178 on Build GRCh38
rs767844361 Chr.12: 19440343-19440343 on Build GRCh38
rs761687041 Chr.12: 19439993-19439993 on Build GRCh38
rs928576508 Chr.12: 19439978-19439978 on Build GRCh38
rs565770036 Chr.12: 19440394-19440394 on Build GRCh38
rs896353189 Chr.12: 19440129-19440129 on Build GRCh38
rs748095646 Chr.12: 19440159-19440159 on Build GRCh38
rs766772255 Chr.12: 19440032-19440032 on Build GRCh38
rs770812990 Chr.12: 19440292-19440292 on Build GRCh38
rs567796065 Chr.12: 19440184-19440184 on Build GRCh38
rs765380250 Chr.12: 19440187-19440187 on Build GRCh38
rs953725478 Chr.12: 19440224-19440224 on Build GRCh38
rs776123281 Chr.12: 19439990-19439991 on Build GRCh38
rs768795871 Chr.12: 19440313-19440313 on Build GRCh38
rs997285101 Chr.12: 19440436-19440436 on Build GRCh38
rs367880727 Chr.12: 19440005-19440005 on Build GRCh38

View Full Product Details