Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48106020-48106515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFKM
Entrez Gene ID
5213
Gene Name
phosphofructokinase, muscle
Gene Aliases
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
UniGene
Hs.75160
Gene Chromosome Location
Chr.12: 48105278-48146404 on Build GRCh38
Transcripts
NM_001166686.1 XM_005268974.1 XM_005268975.1 XM_005268976.3 XM_005268977.1 XM_005268978.2 XM_005268979.1 XM_011538488.2 XM_017019467.1 XM_017019468.1 XM_017019469.1 NM_001267594.1 XM_006719362.2 XM_017019232.1 XM_017019235.1 XM_017019237.1

Gene Symbol
SENP1
Entrez Gene ID
29843
Gene Name
SUMO1/sentrin specific peptidase 1
Gene Aliases
SuPr-2
UniGene
Hs.371957
Gene Chromosome Location
Chr.12: 48042893-48106308 on Build GRCh38
Transcripts
NM_001166686.1 XM_005268974.1 XM_005268975.1 XM_005268976.3 XM_005268977.1 XM_005268978.2 XM_005268979.1 XM_011538488.2 XM_017019467.1 XM_017019468.1 XM_017019469.1 NM_001267594.1 XM_006719362.2 XM_017019232.1 XM_017019235.1 XM_017019237.1

SNP Detail

SNP ID Chromosome Location
rs560507779 Chr.12: 48106178-48106178 on Build GRCh38
rs1039673183 Chr.12: 48106228-48106228 on Build GRCh38
rs936141337 Chr.12: 48106054-48106054 on Build GRCh38
rs990296905 Chr.12: 48106248-48106248 on Build GRCh38
rs900224966 Chr.12: 48106172-48106172 on Build GRCh38
rs182767535 Chr.12: 48106385-48106385 on Build GRCh38
rs1003071315 Chr.12: 48106343-48106343 on Build GRCh38
rs942490866 Chr.12: 48106211-48106211 on Build GRCh38
rs759611657 Chr.12: 48106390-48106391 on Build GRCh38
rs377385692 Chr.12: 48106068-48106068 on Build GRCh38
rs990633415 Chr.12: 48106093-48106093 on Build GRCh38
rs968242137 Chr.12: 48106162-48106162 on Build GRCh38
rs1056880499 Chr.12: 48106345-48106345 on Build GRCh38
rs775212149 Chr.12: 48106153-48106153 on Build GRCh38
rs4760619 Chr.12: 48106148-48106148 on Build GRCh38
rs928620812 Chr.12: 48106299-48106299 on Build GRCh38
rs1051464938 Chr.12: 48106409-48106409 on Build GRCh38
rs567575939 Chr.12: 48106358-48106358 on Build GRCh38
rs765992625 Chr.12: 48106158-48106158 on Build GRCh38
rs755024725 Chr.12: 48106370-48106370 on Build GRCh38
rs530783211 Chr.12: 48106277-48106277 on Build GRCh38
rs561284771 Chr.12: 48106142-48106142 on Build GRCh38
rs919856349 Chr.12: 48106275-48106275 on Build GRCh38
rs898185606 Chr.12: 48106040-48106040 on Build GRCh38
rs886878478 Chr.12: 48106058-48106058 on Build GRCh38
rs1029987503 Chr.12: 48106194-48106194 on Build GRCh38
rs1017773655 Chr.12: 48106168-48106168 on Build GRCh38
rs896671138 Chr.12: 48106361-48106361 on Build GRCh38
rs1031451615 Chr.12: 48106478-48106478 on Build GRCh38
rs140895856 Chr.12: 48106115-48106115 on Build GRCh38
rs188020136 Chr.12: 48106199-48106199 on Build GRCh38
rs562822545 Chr.12: 48106245-48106245 on Build GRCh38
rs1048114132 Chr.12: 48106316-48106316 on Build GRCh38
rs575822837 Chr.12: 48106078-48106078 on Build GRCh38
rs957781894 Chr.12: 48106218-48106218 on Build GRCh38
rs970087601 Chr.12: 48106268-48106268 on Build GRCh38
rs1038836176 Chr.12: 48106371-48106371 on Build GRCh38
rs552675197 Chr.12: 48106216-48106216 on Build GRCh38
rs562635071 Chr.12: 48106372-48106372 on Build GRCh38
rs867639684 Chr.12: 48106208-48106208 on Build GRCh38
rs1001037808 Chr.12: 48106396-48106396 on Build GRCh38
rs762577518 Chr.12: 48106166-48106166 on Build GRCh38
rs886744973 Chr.12: 48106322-48106322 on Build GRCh38
rs968273074 Chr.12: 48106387-48106387 on Build GRCh38
rs941875849 Chr.12: 48106369-48106369 on Build GRCh38
rs1045686477 Chr.12: 48106119-48106119 on Build GRCh38
rs763966220 Chr.12: 48106171-48106171 on Build GRCh38
rs540234596 Chr.12: 48106155-48106155 on Build GRCh38
rs999159617 Chr.12: 48106489-48106489 on Build GRCh38
rs955629113 Chr.12: 48106195-48106195 on Build GRCh38
rs924752777 Chr.12: 48106048-48106048 on Build GRCh38
rs931195837 Chr.12: 48106284-48106284 on Build GRCh38
rs1031874518 Chr.12: 48106202-48106202 on Build GRCh38
rs767127969 Chr.12: 48106097-48106097 on Build GRCh38
rs1023735497 Chr.12: 48106380-48106380 on Build GRCh38
rs916000215 Chr.12: 48106255-48106255 on Build GRCh38
rs540598020 Chr.12: 48106355-48106355 on Build GRCh38
rs745406901 Chr.12: 48106141-48106141 on Build GRCh38
rs192246141 Chr.12: 48106301-48106301 on Build GRCh38
rs902175867 Chr.12: 48106411-48106411 on Build GRCh38
rs151237831 Chr.12: 48106091-48106092 on Build GRCh38

View Full Product Details