Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3865681-3866184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFB
Entrez Gene ID
1677
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
CAD, CPAN, DFF-40, DFF2, DFF40
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3857143-3885429 on Build GRCh38
Transcripts
NM_001282669.1 NM_001320132.1 NM_001320136.1 NM_004402.3 NR_104222.1 NR_135150.1 NR_135151.1 NR_135152.1 XM_011540865.2 XM_017000498.1 XM_017000499.1 XM_017000500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285027 c.430+5G>A p.? Chr.1: 3866005-3866005 on Build GRCh38
908981 c.393G>T p.A131A Chr.1: 3865963-3865963 on Build GRCh38
908980 c.307G>A p.A103T Chr.1: 3865877-3865877 on Build GRCh38
7364175 c.391G>A p.A131T Chr.1: 3865961-3865961 on Build GRCh38
426205 c.406G>A p.A136T Chr.1: 3865976-3865976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764000947 Chr.1: 3866019-3866019 on Build GRCh38
rs763914002 Chr.1: 3865941-3865943 on Build GRCh38
rs1037280222 Chr.1: 3865900-3865900 on Build GRCh38
rs768449630 Chr.1: 3865783-3865783 on Build GRCh38
rs139836683 Chr.1: 3865987-3865987 on Build GRCh38
rs757222871 Chr.1: 3865974-3865974 on Build GRCh38
rs904405024 Chr.1: 3865984-3865984 on Build GRCh38
rs375484294 Chr.1: 3865829-3865829 on Build GRCh38
rs775646835 Chr.1: 3865911-3865911 on Build GRCh38
rs373685537 Chr.1: 3866107-3866107 on Build GRCh38
rs762775624 Chr.1: 3865803-3865805 on Build GRCh38
rs1041052283 Chr.1: 3865962-3865962 on Build GRCh38
rs773522402 Chr.1: 3865989-3865989 on Build GRCh38
rs940296744 Chr.1: 3865898-3865898 on Build GRCh38
rs771995734 Chr.1: 3865796-3865796 on Build GRCh38
rs779628227 Chr.1: 3865800-3865800 on Build GRCh38
rs754422248 Chr.1: 3865965-3865965 on Build GRCh38
rs142035604 Chr.1: 3866004-3866004 on Build GRCh38
rs980236012 Chr.1: 3865932-3865932 on Build GRCh38
rs61035383 Chr.1: 3866070-3866070 on Build GRCh38
rs750921231 Chr.1: 3865865-3865865 on Build GRCh38
rs761749838 Chr.1: 3865739-3865739 on Build GRCh38
rs778700566 Chr.1: 3865787-3865787 on Build GRCh38
rs780305831 Chr.1: 3865975-3865975 on Build GRCh38
rs371235013 Chr.1: 3865945-3865945 on Build GRCh38
rs191694797 Chr.1: 3865781-3865781 on Build GRCh38
rs539442846 Chr.1: 3865709-3865709 on Build GRCh38
rs763211797 Chr.1: 3865830-3865830 on Build GRCh38
rs756153261 Chr.1: 3865813-3865813 on Build GRCh38
rs371766008 Chr.1: 3865990-3865990 on Build GRCh38
rs774418366 Chr.1: 3865943-3865943 on Build GRCh38
rs368962997 Chr.1: 3865831-3865831 on Build GRCh38
rs746930748 Chr.1: 3865905-3865905 on Build GRCh38
rs770008628 Chr.1: 3865906-3865906 on Build GRCh38
rs550477801 Chr.1: 3865986-3865986 on Build GRCh38
rs752737114 Chr.1: 3865754-3865755 on Build GRCh38
rs777653882 Chr.1: 3865971-3865971 on Build GRCh38
rs753490602 Chr.1: 3865876-3865876 on Build GRCh38
rs768591886 Chr.1: 3865985-3865985 on Build GRCh38
rs751358351 Chr.1: 3866021-3866021 on Build GRCh38
rs573639008 Chr.1: 3865819-3865819 on Build GRCh38
rs184201765 Chr.1: 3866096-3866096 on Build GRCh38
rs770918230 Chr.1: 3865743-3865743 on Build GRCh38
rs372200331 Chr.1: 3865850-3865850 on Build GRCh38
rs903253232 Chr.1: 3866099-3866099 on Build GRCh38
rs777925265 Chr.1: 3865892-3865892 on Build GRCh38
rs777090524 Chr.1: 3866003-3866003 on Build GRCh38
rs749554515 Chr.1: 3865925-3865925 on Build GRCh38
rs927037870 Chr.1: 3865710-3865710 on Build GRCh38
rs545284178 Chr.1: 3865963-3865963 on Build GRCh38
rs757083936 Chr.1: 3866023-3866023 on Build GRCh38
rs368953379 Chr.1: 3865879-3865879 on Build GRCh38
rs766919853 Chr.1: 3866001-3866001 on Build GRCh38
rs911376719 Chr.1: 3866102-3866102 on Build GRCh38
rs748922534 Chr.1: 3865782-3865782 on Build GRCh38
rs773718457 Chr.1: 3865947-3865947 on Build GRCh38
rs757718602 Chr.1: 3865772-3865772 on Build GRCh38
rs901164589 Chr.1: 3866151-3866151 on Build GRCh38
rs757928685 Chr.1: 3866017-3866017 on Build GRCh38
rs778593878 Chr.1: 3865878-3865878 on Build GRCh38
rs998910707 Chr.1: 3866106-3866106 on Build GRCh38
rs768877624 Chr.1: 3865926-3865926 on Build GRCh38
rs201202604 Chr.1: 3865755-3865755 on Build GRCh38
rs10909809 Chr.1: 3866044-3866044 on Build GRCh38
rs772681407 Chr.1: 3866143-3866152 on Build GRCh38
rs916184791 Chr.1: 3866069-3866069 on Build GRCh38
rs752936656 Chr.1: 3865754-3865754 on Build GRCh38
rs1049853496 Chr.1: 3865958-3865958 on Build GRCh38
rs201987832 Chr.1: 3865907-3865907 on Build GRCh38
rs774210099 Chr.1: 3865719-3865719 on Build GRCh38
rs768123263 Chr.1: 3865861-3865861 on Build GRCh38
rs780602443 Chr.1: 3865774-3865774 on Build GRCh38
rs758096361 Chr.1: 3865884-3865884 on Build GRCh38
rs367884441 Chr.1: 3865960-3865960 on Build GRCh38
rs530698986 Chr.1: 3865973-3865973 on Build GRCh38
rs140438463 Chr.1: 3865966-3865966 on Build GRCh38
rs749438529 Chr.1: 3865983-3865983 on Build GRCh38
rs907412057 Chr.1: 3865798-3865798 on Build GRCh38
rs758272319 Chr.1: 3865969-3865969 on Build GRCh38
rs769990187 Chr.1: 3865823-3865823 on Build GRCh38
rs375297483 Chr.1: 3865868-3865868 on Build GRCh38
rs571628688 Chr.1: 3866104-3866104 on Build GRCh38
rs764649530 Chr.1: 3866005-3866005 on Build GRCh38
rs765225919 Chr.1: 3865713-3865713 on Build GRCh38
rs752398061 Chr.1: 3866010-3866010 on Build GRCh38
rs938964317 Chr.1: 3865815-3865815 on Build GRCh38
rs748064829 Chr.1: 3865791-3865791 on Build GRCh38
rs542750177 Chr.1: 3865849-3865849 on Build GRCh38
rs754617230 Chr.1: 3865877-3865877 on Build GRCh38
rs759859504 Chr.1: 3865747-3865747 on Build GRCh38
rs755094492 Chr.1: 3866052-3866052 on Build GRCh38
rs767755693 Chr.1: 3865946-3865946 on Build GRCh38
rs112043792 Chr.1: 3866080-3866080 on Build GRCh38
rs781323826 Chr.1: 3866027-3866027 on Build GRCh38
rs145965678 Chr.1: 3865961-3865961 on Build GRCh38
rs768431349 Chr.1: 3865714-3865714 on Build GRCh38
rs749889720 Chr.1: 3865776-3865776 on Build GRCh38
rs752023286 Chr.1: 3865758-3865758 on Build GRCh38
rs772999602 Chr.1: 3865798-3865798 on Build GRCh38
rs891834717 Chr.1: 3866146-3866146 on Build GRCh38
rs765605743 Chr.1: 3865749-3865749 on Build GRCh38
rs764508148 Chr.1: 3865757-3865757 on Build GRCh38
rs920129801 Chr.1: 3865944-3865944 on Build GRCh38

View Full Product Details