Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55469840-55470348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR6C70
Entrez Gene ID
390327
Gene Name
olfactory receptor family 6 subfamily C member 70
Gene Aliases
-
UniGene
Hs.632718
Gene Chromosome Location
Chr.12: 55469200-55470138 on Build GRCh38
Transcripts
NM_001005499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3463099 c.272C>T p.S91F Chr.12: 55469867-55469867 on Build GRCh38
7449418 c.146T>C p.L49P Chr.12: 55469993-55469993 on Build GRCh38
5671602 c.109A>G p.M37V Chr.12: 55470030-55470030 on Build GRCh38
7234177 c.188A>G p.N63S Chr.12: 55469951-55469951 on Build GRCh38
5669008 c.230C>A p.P77H Chr.12: 55469909-55469909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751103583 Chr.12: 55469954-55469955 on Build GRCh38
rs371909341 Chr.12: 55469994-55469994 on Build GRCh38
rs750858025 Chr.12: 55470101-55470101 on Build GRCh38
rs539606491 Chr.12: 55470081-55470081 on Build GRCh38
rs781124816 Chr.12: 55470178-55470178 on Build GRCh38
rs775257011 Chr.12: 55470090-55470090 on Build GRCh38
rs759268344 Chr.12: 55469980-55469980 on Build GRCh38
rs990028194 Chr.12: 55470216-55470216 on Build GRCh38
rs868413154 Chr.12: 55469970-55469970 on Build GRCh38
rs533924506 Chr.12: 55470126-55470126 on Build GRCh38
rs780399246 Chr.12: 55470014-55470014 on Build GRCh38
rs375430845 Chr.12: 55470092-55470092 on Build GRCh38
rs754846624 Chr.12: 55470152-55470153 on Build GRCh38
rs756519489 Chr.12: 55469918-55469918 on Build GRCh38
rs918477482 Chr.12: 55470315-55470315 on Build GRCh38
rs775208768 Chr.12: 55470232-55470232 on Build GRCh38
rs566528196 Chr.12: 55470123-55470123 on Build GRCh38
rs147176459 Chr.12: 55469893-55469893 on Build GRCh38
rs776384068 Chr.12: 55469977-55469977 on Build GRCh38
rs753998115 Chr.12: 55470024-55470024 on Build GRCh38
rs778984312 Chr.12: 55470118-55470118 on Build GRCh38
rs764871551 Chr.12: 55469909-55469909 on Build GRCh38
rs139560912 Chr.12: 55470286-55470286 on Build GRCh38
rs144439557 Chr.12: 55469940-55469940 on Build GRCh38
rs201511358 Chr.12: 55470116-55470116 on Build GRCh38
rs373857105 Chr.12: 55469916-55469916 on Build GRCh38
rs936991202 Chr.12: 55470169-55470169 on Build GRCh38
rs748971133 Chr.12: 55470180-55470180 on Build GRCh38
rs777523851 Chr.12: 55470048-55470048 on Build GRCh38
rs760616040 Chr.12: 55469892-55469892 on Build GRCh38
rs746887633 Chr.12: 55470135-55470135 on Build GRCh38
rs192355567 Chr.12: 55469899-55469899 on Build GRCh38
rs141663640 Chr.12: 55470293-55470293 on Build GRCh38
rs997774783 Chr.12: 55470242-55470242 on Build GRCh38
rs757691426 Chr.12: 55469911-55469911 on Build GRCh38
rs111804291 Chr.12: 55469922-55469922 on Build GRCh38
rs772873385 Chr.12: 55469889-55469889 on Build GRCh38
rs762320452 Chr.12: 55470091-55470091 on Build GRCh38
rs779263968 Chr.12: 55470026-55470026 on Build GRCh38
rs985508963 Chr.12: 55469945-55469945 on Build GRCh38
rs573589963 Chr.12: 55470244-55470244 on Build GRCh38
rs868102243 Chr.12: 55470083-55470083 on Build GRCh38
rs749540632 Chr.12: 55469919-55469919 on Build GRCh38
rs371806756 Chr.12: 55469914-55469914 on Build GRCh38
rs776582475 Chr.12: 55470062-55470062 on Build GRCh38
rs759468909 Chr.12: 55470071-55470071 on Build GRCh38
rs764997542 Chr.12: 55469991-55469991 on Build GRCh38
rs558218918 Chr.12: 55470313-55470313 on Build GRCh38
rs746186135 Chr.12: 55469915-55469915 on Build GRCh38
rs767288966 Chr.12: 55469880-55469880 on Build GRCh38
rs111912512 Chr.12: 55470237-55470237 on Build GRCh38
rs200202218 Chr.12: 55470050-55470050 on Build GRCh38
rs895196789 Chr.12: 55470009-55470009 on Build GRCh38
rs148484851 Chr.12: 55470004-55470004 on Build GRCh38
rs761383184 Chr.12: 55470187-55470187 on Build GRCh38
rs755182949 Chr.12: 55470025-55470025 on Build GRCh38
rs953594599 Chr.12: 55470276-55470276 on Build GRCh38
rs912266609 Chr.12: 55470287-55470287 on Build GRCh38
rs367581109 Chr.12: 55470041-55470041 on Build GRCh38
rs985151334 Chr.12: 55470240-55470240 on Build GRCh38
rs777915727 Chr.12: 55470131-55470131 on Build GRCh38
rs201032439 Chr.12: 55469998-55469998 on Build GRCh38
rs746975519 Chr.12: 55470049-55470049 on Build GRCh38
rs1029047053 Chr.12: 55470268-55470268 on Build GRCh38
rs774300923 Chr.12: 55469872-55469872 on Build GRCh38
rs560172351 Chr.12: 55469866-55469866 on Build GRCh38
rs753596075 Chr.12: 55469894-55469894 on Build GRCh38
rs770603206 Chr.12: 55470222-55470222 on Build GRCh38
rs376579312 Chr.12: 55470012-55470012 on Build GRCh38
rs761658078 Chr.12: 55469877-55469877 on Build GRCh38
rs767991148 Chr.12: 55470181-55470181 on Build GRCh38
rs368053282 Chr.12: 55469954-55469954 on Build GRCh38
rs747307958 Chr.12: 55470137-55470137 on Build GRCh38
rs745635956 Chr.12: 55470152-55470152 on Build GRCh38
rs773834991 Chr.12: 55470182-55470182 on Build GRCh38
rs757255436 Chr.12: 55470140-55470140 on Build GRCh38
rs766932410 Chr.12: 55470109-55470109 on Build GRCh38
rs147422635 Chr.12: 55469962-55469962 on Build GRCh38
rs1031193321 Chr.12: 55470166-55470166 on Build GRCh38
rs1029513133 Chr.12: 55470007-55470007 on Build GRCh38
rs773245045 Chr.12: 55469951-55469951 on Build GRCh38
rs372370284 Chr.12: 55470040-55470040 on Build GRCh38
rs754200243 Chr.12: 55470114-55470114 on Build GRCh38
rs200802708 Chr.12: 55470001-55470001 on Build GRCh38
rs774961437 Chr.12: 55470160-55470160 on Build GRCh38
rs567592422 Chr.12: 55469910-55469910 on Build GRCh38
rs897030754 Chr.12: 55469985-55469985 on Build GRCh38
rs556553413 Chr.12: 55470290-55470290 on Build GRCh38
rs956724419 Chr.12: 55470312-55470312 on Build GRCh38
rs765755155 Chr.12: 55469898-55469898 on Build GRCh38
rs184693499 Chr.12: 55470309-55470309 on Build GRCh38
rs201863379 Chr.12: 55469924-55469924 on Build GRCh38
rs748321392 Chr.12: 55469925-55469925 on Build GRCh38
rs927048168 Chr.12: 55470156-55470156 on Build GRCh38
rs769652454 Chr.12: 55470159-55470159 on Build GRCh38
rs781317451 Chr.12: 55470149-55470149 on Build GRCh38

View Full Product Details