Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 66348086-66348590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP1
Entrez Gene ID
23426
Gene Name
glutamate receptor interacting protein 1
Gene Aliases
GRIP
UniGene
Hs.505946
Gene Chromosome Location
Chr.12: 66347431-67069265 on Build GRCh38
Transcripts
NM_001178074.1 NM_021150.3 XM_005268754.4 XM_005268757.4 XM_011538093.2 XM_017019098.1 XM_017019099.1 XM_017019100.1 XM_017019101.1 XM_017019102.1

SNP Detail

SNP ID Chromosome Location
rs750996493 Chr.12: 66348566-66348567 on Build GRCh38
rs140338954 Chr.12: 66348237-66348237 on Build GRCh38
rs1014922583 Chr.12: 66348265-66348265 on Build GRCh38
rs1049682789 Chr.12: 66348539-66348539 on Build GRCh38
rs1056183700 Chr.12: 66348257-66348261 on Build GRCh38
rs889589974 Chr.12: 66348267-66348267 on Build GRCh38
rs897920394 Chr.12: 66348283-66348283 on Build GRCh38
rs937965691 Chr.12: 66348231-66348231 on Build GRCh38
rs950020301 Chr.12: 66348468-66348468 on Build GRCh38
rs967825487 Chr.12: 66348389-66348389 on Build GRCh38
rs886049794 Chr.12: 66348293-66348294 on Build GRCh38
rs769815711 Chr.12: 66348294-66348295 on Build GRCh38
rs1056357623 Chr.12: 66348451-66348452 on Build GRCh38
rs1168308 Chr.12: 66348403-66348403 on Build GRCh38
rs1014226858 Chr.12: 66348262-66348266 on Build GRCh38
rs907995613 Chr.12: 66348409-66348409 on Build GRCh38
rs570075195 Chr.12: 66348386-66348386 on Build GRCh38
rs991448031 Chr.12: 66348388-66348388 on Build GRCh38
rs572566894 Chr.12: 66348520-66348520 on Build GRCh38
rs962084355 Chr.12: 66348428-66348428 on Build GRCh38
rs551666983 Chr.12: 66348280-66348281 on Build GRCh38
rs895785273 Chr.12: 66348260-66348261 on Build GRCh38
rs994957841 Chr.12: 66348293-66348294 on Build GRCh38
rs201977256 Chr.12: 66348321-66348322 on Build GRCh38
rs558588692 Chr.12: 66348215-66348216 on Build GRCh38
rs148302164 Chr.12: 66348261-66348262 on Build GRCh38
rs905104132 Chr.12: 66348224-66348224 on Build GRCh38
rs867272684 Chr.12: 66348198-66348198 on Build GRCh38
rs537491974 Chr.12: 66348162-66348162 on Build GRCh38
rs984899547 Chr.12: 66348408-66348408 on Build GRCh38
rs531925092 Chr.12: 66348303-66348303 on Build GRCh38
rs1010675056 Chr.12: 66348279-66348279 on Build GRCh38
rs897910505 Chr.12: 66348510-66348510 on Build GRCh38
rs1022425910 Chr.12: 66348295-66348295 on Build GRCh38
rs1050067594 Chr.12: 66348524-66348524 on Build GRCh38
rs1049610748 Chr.12: 66348235-66348235 on Build GRCh38
rs556413166 Chr.12: 66348511-66348511 on Build GRCh38
rs1043589565 Chr.12: 66348169-66348169 on Build GRCh38
rs191784180 Chr.12: 66348352-66348352 on Build GRCh38
rs796588029 Chr.12: 66348340-66348340 on Build GRCh38
rs367709432 Chr.12: 66348339-66348340 on Build GRCh38
rs923267234 Chr.12: 66348467-66348467 on Build GRCh38
rs182639017 Chr.12: 66348453-66348453 on Build GRCh38
rs999228165 Chr.12: 66348324-66348324 on Build GRCh38
rs946512379 Chr.12: 66348540-66348540 on Build GRCh38
rs376924032 Chr.12: 66348271-66348274 on Build GRCh38
rs911075021 Chr.12: 66348527-66348527 on Build GRCh38
rs886049793 Chr.12: 66348262-66348266 on Build GRCh38
rs138816960 Chr.12: 66348361-66348362 on Build GRCh38
rs554240061 Chr.12: 66348402-66348402 on Build GRCh38
rs116640382 Chr.12: 66348316-66348316 on Build GRCh38
rs1036353785 Chr.12: 66348472-66348472 on Build GRCh38
rs200677568 Chr.12: 66348368-66348369 on Build GRCh38

View Full Product Details