Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 66392226-66392722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP1
Entrez Gene ID
23426
Gene Name
glutamate receptor interacting protein 1
Gene Aliases
GRIP
UniGene
Hs.505946
Gene Chromosome Location
Chr.12: 66347431-67069265 on Build GRCh38
Transcripts
NM_001178074.1 NM_021150.3 XM_005268754.4 XM_005268757.4 XM_011538093.2 XM_017019098.1 XM_017019099.1 XM_017019100.1 XM_017019101.1 XM_017019102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4756769 c.2095A>C p.I699L Chr.12: 66392695-66392695 on Build GRCh38
7100553 c.2316G>A p.L772L Chr.12: 66392456-66392456 on Build GRCh38
6675657 c.2412T>C p.S804S Chr.12: 66392360-66392360 on Build GRCh38
3399002 c.2382G>A p.M794I Chr.12: 66392390-66392390 on Build GRCh38
6886201 c.2231G>A p.G744E Chr.12: 66392715-66392715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041091322 Chr.12: 66392458-66392458 on Build GRCh38
rs895784191 Chr.12: 66392600-66392600 on Build GRCh38
rs549783937 Chr.12: 66392283-66392283 on Build GRCh38
rs886049797 Chr.12: 66392411-66392411 on Build GRCh38
rs7970076 Chr.12: 66392293-66392293 on Build GRCh38
rs750338089 Chr.12: 66392286-66392286 on Build GRCh38
rs769389718 Chr.12: 66392375-66392375 on Build GRCh38
rs746288453 Chr.12: 66392340-66392340 on Build GRCh38
rs747437893 Chr.12: 66392654-66392654 on Build GRCh38
rs772516123 Chr.12: 66392446-66392446 on Build GRCh38
rs1014683071 Chr.12: 66392684-66392684 on Build GRCh38
rs1032463862 Chr.12: 66392279-66392279 on Build GRCh38
rs754096539 Chr.12: 66392515-66392515 on Build GRCh38
rs768254691 Chr.12: 66392260-66392260 on Build GRCh38
rs751518904 Chr.12: 66392385-66392385 on Build GRCh38
rs752872677 Chr.12: 66392396-66392396 on Build GRCh38
rs1043977323 Chr.12: 66392555-66392555 on Build GRCh38
rs757227837 Chr.12: 66392519-66392519 on Build GRCh38
rs747772513 Chr.12: 66392460-66392460 on Build GRCh38
rs745472874 Chr.12: 66392522-66392523 on Build GRCh38
rs766292531 Chr.12: 66392382-66392382 on Build GRCh38
rs749768786 Chr.12: 66392313-66392313 on Build GRCh38
rs397514485 Chr.12: 66392676-66392676 on Build GRCh38
rs189784716 Chr.12: 66392557-66392557 on Build GRCh38
rs776280921 Chr.12: 66392263-66392263 on Build GRCh38
rs752929795 Chr.12: 66392309-66392309 on Build GRCh38
rs757978398 Chr.12: 66392369-66392369 on Build GRCh38
rs770487416 Chr.12: 66392470-66392470 on Build GRCh38
rs771388269 Chr.12: 66392329-66392329 on Build GRCh38
rs556130145 Chr.12: 66392525-66392525 on Build GRCh38
rs762831429 Chr.12: 66392290-66392290 on Build GRCh38
rs944380005 Chr.12: 66392474-66392474 on Build GRCh38
rs779325243 Chr.12: 66392339-66392339 on Build GRCh38
rs371030807 Chr.12: 66392342-66392342 on Build GRCh38
rs971199891 Chr.12: 66392604-66392604 on Build GRCh38
rs767322707 Chr.12: 66392395-66392395 on Build GRCh38
rs761344168 Chr.12: 66392278-66392278 on Build GRCh38
rs1009619278 Chr.12: 66392569-66392569 on Build GRCh38
rs937950591 Chr.12: 66392568-66392568 on Build GRCh38
rs777229959 Chr.12: 66392681-66392681 on Build GRCh38
rs762197654 Chr.12: 66392624-66392624 on Build GRCh38
rs369165287 Chr.12: 66392482-66392482 on Build GRCh38
rs756364602 Chr.12: 66392422-66392422 on Build GRCh38
rs181365280 Chr.12: 66392445-66392445 on Build GRCh38
rs7970771 Chr.12: 66392561-66392561 on Build GRCh38
rs757781712 Chr.12: 66392442-66392442 on Build GRCh38
rs760872241 Chr.12: 66392502-66392502 on Build GRCh38
rs371343500 Chr.12: 66392437-66392437 on Build GRCh38
rs780597898 Chr.12: 66392453-66392453 on Build GRCh38
rs781324542 Chr.12: 66392308-66392308 on Build GRCh38
rs200863167 Chr.12: 66392489-66392489 on Build GRCh38
rs1021012419 Chr.12: 66392578-66392578 on Build GRCh38
rs569662809 Chr.12: 66392529-66392529 on Build GRCh38
rs753920435 Chr.12: 66392438-66392438 on Build GRCh38
rs777858404 Chr.12: 66392312-66392312 on Build GRCh38
rs755396456 Chr.12: 66392669-66392669 on Build GRCh38
rs780546938 Chr.12: 66392640-66392640 on Build GRCh38
rs13277 Chr.12: 66392311-66392311 on Build GRCh38
rs200363939 Chr.12: 66392379-66392379 on Build GRCh38
rs769621251 Chr.12: 66392462-66392462 on Build GRCh38
rs775312134 Chr.12: 66392490-66392490 on Build GRCh38
rs748677923 Chr.12: 66392466-66392466 on Build GRCh38
rs759500891 Chr.12: 66392483-66392483 on Build GRCh38
rs778663653 Chr.12: 66392430-66392432 on Build GRCh38
rs776369902 Chr.12: 66392344-66392344 on Build GRCh38
rs900115532 Chr.12: 66392481-66392481 on Build GRCh38
rs144494437 Chr.12: 66392391-66392391 on Build GRCh38
rs373844044 Chr.12: 66392443-66392443 on Build GRCh38
rs186127141 Chr.12: 66392510-66392510 on Build GRCh38
rs563213520 Chr.12: 66392292-66392292 on Build GRCh38
rs759017866 Chr.12: 66392547-66392547 on Build GRCh38
rs750858998 Chr.12: 66392527-66392527 on Build GRCh38

View Full Product Details