Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9602355-9602849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM201
Entrez Gene ID
199953
Gene Name
transmembrane protein 201
Gene Aliases
NET5
UniGene
Hs.632365
Gene Chromosome Location
Chr.1: 9588843-9614877 on Build GRCh38
Transcripts
NM_001010866.3 NM_001130924.2 XM_006710417.3 XM_011540910.2 XM_017000549.1 XM_017000550.1 XM_017000551.1 XM_017000552.1

SNP Detail

SNP ID Chromosome Location
rs368463902 Chr.1: 9602490-9602490 on Build GRCh38
rs974069705 Chr.1: 9602472-9602472 on Build GRCh38
rs1030303591 Chr.1: 9602384-9602384 on Build GRCh38
rs552829434 Chr.1: 9602593-9602593 on Build GRCh38
rs1042187693 Chr.1: 9602620-9602621 on Build GRCh38
rs1030449281 Chr.1: 9602736-9602736 on Build GRCh38
rs985996018 Chr.1: 9602439-9602439 on Build GRCh38
rs955152168 Chr.1: 9602443-9602443 on Build GRCh38
rs1029920084 Chr.1: 9602624-9602624 on Build GRCh38
rs927250207 Chr.1: 9602543-9602543 on Build GRCh38
rs139782863 Chr.1: 9602396-9602396 on Build GRCh38
rs1052185862 Chr.1: 9602825-9602825 on Build GRCh38
rs563209065 Chr.1: 9602753-9602753 on Build GRCh38
rs936398280 Chr.1: 9602636-9602636 on Build GRCh38
rs995779769 Chr.1: 9602710-9602710 on Build GRCh38
rs917875920 Chr.1: 9602763-9602763 on Build GRCh38
rs375133696 Chr.1: 9602383-9602383 on Build GRCh38
rs974144251 Chr.1: 9602711-9602711 on Build GRCh38
rs774581410 Chr.1: 9602815-9602815 on Build GRCh38
rs890705804 Chr.1: 9602742-9602742 on Build GRCh38
rs541484957 Chr.1: 9602693-9602693 on Build GRCh38
rs771511748 Chr.1: 9602769-9602769 on Build GRCh38
rs372707799 Chr.1: 9602754-9602754 on Build GRCh38
rs773892592 Chr.1: 9602586-9602586 on Build GRCh38
rs530723786 Chr.1: 9602812-9602812 on Build GRCh38
rs992107447 Chr.1: 9602503-9602503 on Build GRCh38
rs992822667 Chr.1: 9602802-9602802 on Build GRCh38
rs917788185 Chr.1: 9602534-9602534 on Build GRCh38
rs574354883 Chr.1: 9602689-9602689 on Build GRCh38
rs1025662528 Chr.1: 9602727-9602727 on Build GRCh38
rs576964186 Chr.1: 9602556-9602556 on Build GRCh38
rs559202583 Chr.1: 9602447-9602447 on Build GRCh38
rs929952599 Chr.1: 9602491-9602491 on Build GRCh38
rs200848529 Chr.1: 9602499-9602499 on Build GRCh38
rs1005549293 Chr.1: 9602743-9602743 on Build GRCh38
rs114457968 Chr.1: 9602574-9602574 on Build GRCh38
rs962763733 Chr.1: 9602469-9602469 on Build GRCh38
rs947921398 Chr.1: 9602544-9602544 on Build GRCh38
rs1018381645 Chr.1: 9602639-9602639 on Build GRCh38
rs892154550 Chr.1: 9602582-9602582 on Build GRCh38
rs962799353 Chr.1: 9602653-9602653 on Build GRCh38
rs947963578 Chr.1: 9602800-9602800 on Build GRCh38
rs1034418178 Chr.1: 9602783-9602783 on Build GRCh38
rs112835708 Chr.1: 9602638-9602638 on Build GRCh38
rs903627906 Chr.1: 9602565-9602565 on Build GRCh38
rs973502948 Chr.1: 9602489-9602489 on Build GRCh38
rs902345377 Chr.1: 9602626-9602626 on Build GRCh38
rs545547783 Chr.1: 9602820-9602820 on Build GRCh38
rs759870446 Chr.1: 9602470-9602470 on Build GRCh38
rs949364849 Chr.1: 9602580-9602580 on Build GRCh38
rs980657755 Chr.1: 9602810-9602810 on Build GRCh38
rs1015515644 Chr.1: 9602462-9602462 on Build GRCh38
rs996353277 Chr.1: 9602434-9602434 on Build GRCh38
rs936390999 Chr.1: 9602571-9602571 on Build GRCh38
rs900289498 Chr.1: 9602683-9602683 on Build GRCh38
rs955567941 Chr.1: 9602427-9602427 on Build GRCh38
rs749804566 Chr.1: 9602635-9602635 on Build GRCh38
rs868385142 Chr.1: 9602561-9602561 on Build GRCh38
rs911342846 Chr.1: 9602453-9602453 on Build GRCh38

View Full Product Details