Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116911029-116911524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW8
Entrez Gene ID
26259
Gene Name
F-box and WD repeat domain containing 8
Gene Aliases
FBW6, FBW8, FBX29, FBXO29, FBXW6
UniGene
Hs.624537
Gene Chromosome Location
Chr.12: 116910949-117032498 on Build GRCh38
Transcripts
NM_012174.1 NM_153348.2 XM_005253867.2 XM_005253868.3 XM_017019176.1 XM_017019177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500230 c.189G>C p.A63A Chr.12: 116911226-116911226 on Build GRCh38
6531678 c.155A>C p.D52A Chr.12: 116911192-116911192 on Build GRCh38
6395509 c.80G>A p.R27Q Chr.12: 116911117-116911117 on Build GRCh38
5429542 c.223G>C p.E75Q Chr.12: 116911260-116911260 on Build GRCh38
6424305 c.121G>A p.V41M Chr.12: 116911158-116911158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944257565 Chr.12: 116911149-116911149 on Build GRCh38
rs887301358 Chr.12: 116911144-116911144 on Build GRCh38
rs892914508 Chr.12: 116911188-116911188 on Build GRCh38
rs928437643 Chr.12: 116911379-116911379 on Build GRCh38
rs766691499 Chr.12: 116911146-116911146 on Build GRCh38
rs150631436 Chr.12: 116911340-116911340 on Build GRCh38
rs745843233 Chr.12: 116911061-116911061 on Build GRCh38
rs753708747 Chr.12: 116911496-116911496 on Build GRCh38
rs919989584 Chr.12: 116911271-116911271 on Build GRCh38
rs1047284849 Chr.12: 116911142-116911142 on Build GRCh38
rs576697686 Chr.12: 116911367-116911367 on Build GRCh38
rs774166467 Chr.12: 116911054-116911054 on Build GRCh38
rs557769265 Chr.12: 116911296-116911296 on Build GRCh38
rs906843527 Chr.12: 116911206-116911206 on Build GRCh38
rs1028689687 Chr.12: 116911169-116911169 on Build GRCh38
rs900125830 Chr.12: 116911322-116911322 on Build GRCh38
rs1011253485 Chr.12: 116911343-116911343 on Build GRCh38
rs928599826 Chr.12: 116911058-116911058 on Build GRCh38
rs112339051 Chr.12: 116911163-116911163 on Build GRCh38
rs535655795 Chr.12: 116911150-116911150 on Build GRCh38
rs761515590 Chr.12: 116911121-116911121 on Build GRCh38
rs770664743 Chr.12: 116911051-116911051 on Build GRCh38
rs749308734 Chr.12: 116911141-116911159 on Build GRCh38
rs981585219 Chr.12: 116911375-116911375 on Build GRCh38
rs948605230 Chr.12: 116911372-116911373 on Build GRCh38
rs1041306792 Chr.12: 116911160-116911160 on Build GRCh38
rs967251611 Chr.12: 116911214-116911214 on Build GRCh38
rs991522372 Chr.12: 116911259-116911259 on Build GRCh38
rs948489963 Chr.12: 116911182-116911182 on Build GRCh38
rs1011347996 Chr.12: 116911195-116911195 on Build GRCh38
rs900553882 Chr.12: 116911272-116911272 on Build GRCh38
rs17857139 Chr.12: 116911173-116911173 on Build GRCh38
rs1020042332 Chr.12: 116911350-116911350 on Build GRCh38
rs981733670 Chr.12: 116911229-116911229 on Build GRCh38
rs534476620 Chr.12: 116911423-116911423 on Build GRCh38
rs928952541 Chr.12: 116911441-116911442 on Build GRCh38
rs1047430612 Chr.12: 116911459-116911459 on Build GRCh38
rs902977775 Chr.12: 116911359-116911359 on Build GRCh38
rs1045380958 Chr.12: 116911184-116911184 on Build GRCh38
rs982899113 Chr.12: 116911288-116911288 on Build GRCh38
rs11068230 Chr.12: 116911209-116911209 on Build GRCh38
rs554181409 Chr.12: 116911437-116911437 on Build GRCh38
rs931635203 Chr.12: 116911324-116911324 on Build GRCh38
rs1023910487 Chr.12: 116911372-116911372 on Build GRCh38
rs779638551 Chr.12: 116911414-116911414 on Build GRCh38
rs944220735 Chr.12: 116911300-116911300 on Build GRCh38
rs1035615463 Chr.12: 116911237-116911237 on Build GRCh38
rs763638591 Chr.12: 116911091-116911091 on Build GRCh38
rs1050077671 Chr.12: 116911336-116911336 on Build GRCh38
rs1041689905 Chr.12: 116911311-116911311 on Build GRCh38
rs1038239071 Chr.12: 116911159-116911159 on Build GRCh38
rs924568152 Chr.12: 116911118-116911118 on Build GRCh38
rs981401436 Chr.12: 116911055-116911055 on Build GRCh38
rs374397512 Chr.12: 116911068-116911068 on Build GRCh38
rs958694273 Chr.12: 116911245-116911245 on Build GRCh38
rs1002749932 Chr.12: 116911422-116911422 on Build GRCh38
rs537978133 Chr.12: 116911282-116911282 on Build GRCh38
rs931462175 Chr.12: 116911120-116911120 on Build GRCh38
rs768078394 Chr.12: 116911170-116911170 on Build GRCh38
rs774930316 Chr.12: 116911066-116911066 on Build GRCh38
rs978104903 Chr.12: 116911333-116911333 on Build GRCh38
rs776443595 Chr.12: 116911113-116911113 on Build GRCh38
rs755492051 Chr.12: 116911166-116911166 on Build GRCh38
rs12815835 Chr.12: 116911208-116911208 on Build GRCh38
rs915749649 Chr.12: 116911366-116911366 on Build GRCh38
rs772032889 Chr.12: 116911062-116911062 on Build GRCh38

View Full Product Details