Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12755611-12756115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf158
Entrez Gene ID
93190
Gene Name
chromosome 1 open reading frame 158
Gene Aliases
-
UniGene
Hs.98095
Gene Chromosome Location
Chr.1: 12746186-12761169 on Build GRCh38
Transcripts
NM_152290.3 XM_011542434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3472533 c.210G>A p.G70G Chr.1: 12755798-12755798 on Build GRCh38
3472532 c.172C>T p.P58S Chr.1: 12755760-12755760 on Build GRCh38
4400329 c.197C>T p.S66F Chr.1: 12755785-12755785 on Build GRCh38
3862447 c.215G>A p.R72K Chr.1: 12755803-12755803 on Build GRCh38
894889 c.121G>T p.D41Y Chr.1: 12755709-12755709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142466978 Chr.1: 12755804-12755804 on Build GRCh38
rs750156644 Chr.1: 12755813-12755813 on Build GRCh38
rs772764831 Chr.1: 12755717-12755717 on Build GRCh38
rs776920542 Chr.1: 12755783-12755783 on Build GRCh38
rs920644521 Chr.1: 12756031-12756031 on Build GRCh38
rs948759549 Chr.1: 12755692-12755692 on Build GRCh38
rs761323513 Chr.1: 12755901-12755901 on Build GRCh38
rs3795747 Chr.1: 12755877-12755877 on Build GRCh38
rs571261017 Chr.1: 12756032-12756032 on Build GRCh38
rs771746865 Chr.1: 12755942-12755943 on Build GRCh38
rs973470866 Chr.1: 12755972-12755972 on Build GRCh38
rs186768572 Chr.1: 12756070-12756070 on Build GRCh38
rs558739935 Chr.1: 12756072-12756072 on Build GRCh38
rs71647059 Chr.1: 12755700-12755700 on Build GRCh38
rs1037733091 Chr.1: 12755635-12755635 on Build GRCh38
rs776194363 Chr.1: 12755661-12755661 on Build GRCh38
rs774838694 Chr.1: 12755944-12755944 on Build GRCh38
rs200193668 Chr.1: 12755668-12755668 on Build GRCh38
rs112964097 Chr.1: 12755980-12755980 on Build GRCh38
rs1029027851 Chr.1: 12755724-12755724 on Build GRCh38
rs201802847 Chr.1: 12755833-12755833 on Build GRCh38
rs866543154 Chr.1: 12755693-12755693 on Build GRCh38
rs1018671898 Chr.1: 12755864-12755864 on Build GRCh38
rs183593740 Chr.1: 12755988-12755988 on Build GRCh38
rs568981127 Chr.1: 12755897-12755897 on Build GRCh38
rs376083079 Chr.1: 12755786-12755786 on Build GRCh38
rs145375004 Chr.1: 12755708-12755708 on Build GRCh38
rs763902092 Chr.1: 12755713-12755714 on Build GRCh38
rs957701717 Chr.1: 12755835-12755835 on Build GRCh38
rs536689502 Chr.1: 12755671-12755671 on Build GRCh38
rs370999199 Chr.1: 12755828-12755828 on Build GRCh38
rs143607816 Chr.1: 12755725-12755725 on Build GRCh38
rs907271482 Chr.1: 12755950-12755950 on Build GRCh38
rs767779910 Chr.1: 12755767-12755767 on Build GRCh38
rs192321925 Chr.1: 12756073-12756073 on Build GRCh38
rs934730486 Chr.1: 12756058-12756058 on Build GRCh38
rs145410215 Chr.1: 12755765-12755765 on Build GRCh38
rs529212782 Chr.1: 12755852-12755852 on Build GRCh38
rs868812867 Chr.1: 12756064-12756064 on Build GRCh38
rs754806387 Chr.1: 12755692-12755692 on Build GRCh38
rs1055838838 Chr.1: 12756047-12756047 on Build GRCh38
rs147495703 Chr.1: 12755727-12755727 on Build GRCh38
rs1028536418 Chr.1: 12756074-12756074 on Build GRCh38
rs775128662 Chr.1: 12755788-12755788 on Build GRCh38
rs765116078 Chr.1: 12755696-12755703 on Build GRCh38
rs1045719397 Chr.1: 12755948-12755948 on Build GRCh38
rs758382403 Chr.1: 12755698-12755698 on Build GRCh38
rs955017650 Chr.1: 12755940-12755940 on Build GRCh38
rs72860701 Chr.1: 12755722-12755722 on Build GRCh38
rs915815223 Chr.1: 12755946-12755946 on Build GRCh38
rs761422183 Chr.1: 12755805-12755805 on Build GRCh38
rs762790685 Chr.1: 12755791-12755791 on Build GRCh38
rs551471790 Chr.1: 12756005-12756005 on Build GRCh38
rs142642917 Chr.1: 12755718-12755718 on Build GRCh38
rs764878308 Chr.1: 12755689-12755689 on Build GRCh38
rs756790033 Chr.1: 12755712-12755712 on Build GRCh38
rs747454026 Chr.1: 12755862-12755862 on Build GRCh38
rs948687619 Chr.1: 12755947-12755947 on Build GRCh38
rs780903660 Chr.1: 12755716-12755716 on Build GRCh38
rs759377017 Chr.1: 12755680-12755680 on Build GRCh38
rs930505989 Chr.1: 12755902-12755902 on Build GRCh38
rs753957921 Chr.1: 12755851-12755853 on Build GRCh38
rs766923856 Chr.1: 12755809-12755809 on Build GRCh38
rs115530775 Chr.1: 12755696-12755696 on Build GRCh38
rs770235297 Chr.1: 12755944-12755945 on Build GRCh38
rs369328058 Chr.1: 12755856-12755856 on Build GRCh38
rs766607538 Chr.1: 12755667-12755667 on Build GRCh38
rs374923758 Chr.1: 12755741-12755741 on Build GRCh38
rs965798440 Chr.1: 12755868-12755868 on Build GRCh38
rs779382238 Chr.1: 12755851-12755851 on Build GRCh38
rs760935545 Chr.1: 12755663-12755663 on Build GRCh38
rs937345008 Chr.1: 12756042-12756042 on Build GRCh38
rs764160115 Chr.1: 12755792-12755792 on Build GRCh38

View Full Product Details