Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 111217570-111218093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
UniGene
Hs.508738
Gene Chromosome Location
Chr.13: 111114619-111305734 on Build GRCh38
Transcripts
NM_001113511.2 NM_001113512.2 NM_001113513.1 NM_001320851.1 NM_001320852.1 NM_001320853.1 NM_003899.3 NM_145735.3 XM_006719956.3 XM_011521133.2 XM_017020814.1 XM_017020815.1 XM_017020816.1 XM_017020817.1 XM_017020818.1 XM_017020819.1 XM_017020820.1 XM_017020821.1 XM_017020822.1 XM_017020823.1 XM_017020824.1 XM_017020825.1 XM_017020826.1 XM_017020827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4419090 c.480T>G p.D160E Chr.13: 111217690-111217690 on Build GRCh38
4419089 c.543T>G p.D181E Chr.13: 111217690-111217690 on Build GRCh38
1365496 c.566T>C p.V189A Chr.13: 111217776-111217776 on Build GRCh38
2263422 c.641C>A p.T214N Chr.13: 111217788-111217788 on Build GRCh38
6604472 c.61A>G p.N21D Chr.13: 111217742-111217742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202035865 Chr.13: 111217834-111217834 on Build GRCh38
rs771116682 Chr.13: 111217904-111217904 on Build GRCh38
rs181527441 Chr.13: 111217961-111217961 on Build GRCh38
rs370423370 Chr.13: 111217895-111217895 on Build GRCh38
rs924694001 Chr.13: 111217964-111217964 on Build GRCh38
rs756748260 Chr.13: 111217774-111217774 on Build GRCh38
rs769310646 Chr.13: 111217923-111217923 on Build GRCh38
rs927245640 Chr.13: 111217780-111217780 on Build GRCh38
rs764919975 Chr.13: 111217791-111217791 on Build GRCh38
rs1027315638 Chr.13: 111217888-111217888 on Build GRCh38
rs748383971 Chr.13: 111217688-111217688 on Build GRCh38
rs145220586 Chr.13: 111217825-111217825 on Build GRCh38
rs762367710 Chr.13: 111217927-111217927 on Build GRCh38
rs375511480 Chr.13: 111217657-111217657 on Build GRCh38
rs548277731 Chr.13: 111217829-111217829 on Build GRCh38
rs562655661 Chr.13: 111217624-111217624 on Build GRCh38
rs924688701 Chr.13: 111217747-111217747 on Build GRCh38
rs559951800 Chr.13: 111217949-111217949 on Build GRCh38
rs550628372 Chr.13: 111217604-111217605 on Build GRCh38
rs9522157 Chr.13: 111217807-111217807 on Build GRCh38
rs777296458 Chr.13: 111217640-111217640 on Build GRCh38
rs773604491 Chr.13: 111217844-111217844 on Build GRCh38
rs369515749 Chr.13: 111217673-111217673 on Build GRCh38
rs758014704 Chr.13: 111217882-111217882 on Build GRCh38
rs768643344 Chr.13: 111217680-111217680 on Build GRCh38
rs761125942 Chr.13: 111217920-111217920 on Build GRCh38
rs372175572 Chr.13: 111217648-111217648 on Build GRCh38
rs760038830 Chr.13: 111217740-111217740 on Build GRCh38
rs200218776 Chr.13: 111217833-111217833 on Build GRCh38
rs550412959 Chr.13: 111218050-111218050 on Build GRCh38
rs766094366 Chr.13: 111217884-111217884 on Build GRCh38
rs9522158 Chr.13: 111217990-111217990 on Build GRCh38
rs956117303 Chr.13: 111217969-111217969 on Build GRCh38
rs749480802 Chr.13: 111217858-111217858 on Build GRCh38
rs147769518 Chr.13: 111217890-111217890 on Build GRCh38
rs759964144 Chr.13: 111217863-111217863 on Build GRCh38
rs771833196 Chr.13: 111217659-111217659 on Build GRCh38
rs756293293 Chr.13: 111217893-111217893 on Build GRCh38
rs574301804 Chr.13: 111217629-111217629 on Build GRCh38
rs376373165 Chr.13: 111217602-111217602 on Build GRCh38
rs780984892 Chr.13: 111217832-111217832 on Build GRCh38
rs1018636994 Chr.13: 111217608-111217608 on Build GRCh38
rs772645844 Chr.13: 111217925-111217925 on Build GRCh38
rs763456797 Chr.13: 111217693-111217693 on Build GRCh38
rs560236071 Chr.13: 111217654-111217654 on Build GRCh38
rs189635357 Chr.13: 111217908-111217908 on Build GRCh38
rs2296354 Chr.13: 111217690-111217690 on Build GRCh38
rs144138131 Chr.13: 111217707-111217707 on Build GRCh38
rs534618453 Chr.13: 111217764-111217764 on Build GRCh38
rs541604698 Chr.13: 111217942-111217942 on Build GRCh38
rs751435250 Chr.13: 111217886-111217886 on Build GRCh38
rs773551411 Chr.13: 111217692-111217692 on Build GRCh38
rs111652345 Chr.13: 111218030-111218030 on Build GRCh38
rs978770021 Chr.13: 111218043-111218043 on Build GRCh38
rs531616530 Chr.13: 111218047-111218047 on Build GRCh38
rs373978851 Chr.13: 111217876-111217876 on Build GRCh38
rs141281183 Chr.13: 111217917-111217917 on Build GRCh38
rs41275140 Chr.13: 111217879-111217879 on Build GRCh38
rs564454886 Chr.13: 111218042-111218042 on Build GRCh38
rs761352896 Chr.13: 111217874-111217874 on Build GRCh38
rs779901304 Chr.13: 111217666-111217666 on Build GRCh38
rs774717680 Chr.13: 111217862-111217862 on Build GRCh38
rs149212045 Chr.13: 111217801-111217801 on Build GRCh38
rs202171987 Chr.13: 111217891-111217891 on Build GRCh38
rs771220906 Chr.13: 111217859-111217859 on Build GRCh38
rs774750752 Chr.13: 111217726-111217726 on Build GRCh38
rs916077246 Chr.13: 111217708-111217708 on Build GRCh38
rs184758955 Chr.13: 111217687-111217687 on Build GRCh38
rs550206678 Chr.13: 111217750-111217750 on Build GRCh38
rs775954288 Chr.13: 111217865-111217865 on Build GRCh38
rs962002779 Chr.13: 111217643-111217643 on Build GRCh38
rs369053545 Chr.13: 111217633-111217633 on Build GRCh38
rs868083558 Chr.13: 111217605-111217605 on Build GRCh38
rs750102418 Chr.13: 111217880-111217880 on Build GRCh38
rs149136288 Chr.13: 111217828-111217828 on Build GRCh38
rs910442344 Chr.13: 111218067-111218067 on Build GRCh38
rs574267143 Chr.13: 111217919-111217919 on Build GRCh38
rs143217884 Chr.13: 111217837-111217837 on Build GRCh38
rs890519027 Chr.13: 111217948-111217948 on Build GRCh38
rs758225753 Chr.13: 111217820-111217820 on Build GRCh38
rs755618918 Chr.13: 111217632-111217632 on Build GRCh38
rs376000352 Chr.13: 111217902-111217902 on Build GRCh38
rs369467671 Chr.13: 111217864-111217864 on Build GRCh38
rs866839509 Chr.13: 111217685-111217685 on Build GRCh38
rs200899980 Chr.13: 111217720-111217720 on Build GRCh38
rs971527897 Chr.13: 111217712-111217712 on Build GRCh38
rs1054348272 Chr.13: 111217778-111217778 on Build GRCh38
rs753454100 Chr.13: 111217646-111217646 on Build GRCh38
rs914572616 Chr.13: 111218036-111218036 on Build GRCh38
rs1020351870 Chr.13: 111218014-111218014 on Build GRCh38
rs543786979 Chr.13: 111217722-111217722 on Build GRCh38
rs374096402 Chr.13: 111217910-111217910 on Build GRCh38

View Full Product Details