Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112067520-112068018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00403
Entrez Gene ID
100505996
Gene Name
long intergenic non-protein coding RNA 403
Gene Aliases
-
UniGene
Hs.654853
Gene Chromosome Location
Chr.13: 111972310-112108015 on Build GRCh38
Transcripts
NR_120392.1 NM_005986.2

Gene Symbol
SOX1
Entrez Gene ID
6656
Gene Name
SRY-box 1
Gene Aliases
-
UniGene
Hs.202526
Gene Chromosome Location
Chr.13: 112067599-112071706 on Build GRCh38
Transcripts
NR_120392.1 NM_005986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4729127 c.324C>T p.R108R Chr.13: 112067982-112067982 on Build GRCh38
2263543 c.318G>A p.R106R Chr.13: 112067976-112067976 on Build GRCh38
2263546 c.342G>A p.E114E Chr.13: 112068000-112068000 on Build GRCh38
6138515 c.51G>C p.Q17H Chr.13: 112067709-112067709 on Build GRCh38
695976 c.157C>T p.R53W Chr.13: 112067815-112067815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761294176 Chr.13: 112067630-112067630 on Build GRCh38
rs772315499 Chr.13: 112067736-112067736 on Build GRCh38
rs921314504 Chr.13: 112067796-112067796 on Build GRCh38
rs780949817 Chr.13: 112067713-112067713 on Build GRCh38
rs375647033 Chr.13: 112067658-112067658 on Build GRCh38
rs754659872 Chr.13: 112067656-112067656 on Build GRCh38
rs754244329 Chr.13: 112067690-112067690 on Build GRCh38
rs759899415 Chr.13: 112067618-112067618 on Build GRCh38
rs148751385 Chr.13: 112067811-112067811 on Build GRCh38
rs747265562 Chr.13: 112067916-112067916 on Build GRCh38
rs755806902 Chr.13: 112067718-112067718 on Build GRCh38
rs896164603 Chr.13: 112067760-112067760 on Build GRCh38
rs781178996 Chr.13: 112067657-112067657 on Build GRCh38
rs1054758083 Chr.13: 112067757-112067757 on Build GRCh38
rs887265059 Chr.13: 112067547-112067547 on Build GRCh38
rs767936500 Chr.13: 112067619-112067619 on Build GRCh38
rs771858425 Chr.13: 112067749-112067760 on Build GRCh38
rs768841222 Chr.13: 112067768-112067768 on Build GRCh38
rs749398083 Chr.13: 112067666-112067666 on Build GRCh38
rs748353566 Chr.13: 112067779-112067784 on Build GRCh38
rs768897965 Chr.13: 112067918-112067918 on Build GRCh38
rs113831383 Chr.13: 112067612-112067612 on Build GRCh38
rs966624526 Chr.13: 112067762-112067762 on Build GRCh38
rs759065233 Chr.13: 112067795-112067795 on Build GRCh38
rs766040889 Chr.13: 112067704-112067704 on Build GRCh38
rs780365443 Chr.13: 112067767-112067775 on Build GRCh38
rs760038168 Chr.13: 112067761-112067772 on Build GRCh38
rs878861984 Chr.13: 112067593-112067593 on Build GRCh38
rs966949879 Chr.13: 112067628-112067628 on Build GRCh38
rs746475496 Chr.13: 112067743-112067748 on Build GRCh38
rs773796467 Chr.13: 112067937-112067937 on Build GRCh38
rs878984680 Chr.13: 112067553-112067553 on Build GRCh38
rs775645164 Chr.13: 112067740-112067740 on Build GRCh38
rs375777001 Chr.13: 112067771-112067772 on Build GRCh38
rs754715087 Chr.13: 112067709-112067709 on Build GRCh38
rs151310706 Chr.13: 112067940-112067940 on Build GRCh38
rs763875400 Chr.13: 112067760-112067761 on Build GRCh38
rs749982953 Chr.13: 112067637-112067637 on Build GRCh38
rs896216943 Chr.13: 112067961-112067961 on Build GRCh38
rs764480197 Chr.13: 112067632-112067632 on Build GRCh38
rs144349683 Chr.13: 112067973-112067973 on Build GRCh38
rs768983817 Chr.13: 112067668-112067670 on Build GRCh38
rs748151218 Chr.13: 112067608-112067608 on Build GRCh38
rs777036757 Chr.13: 112067781-112067781 on Build GRCh38
rs772260690 Chr.13: 112067680-112067680 on Build GRCh38
rs535164513 Chr.13: 112067874-112067874 on Build GRCh38
rs747418130 Chr.13: 112067756-112067756 on Build GRCh38
rs760295282 Chr.13: 112067941-112067941 on Build GRCh38
rs570383720 Chr.13: 112067573-112067573 on Build GRCh38
rs747077010 Chr.13: 112067981-112067981 on Build GRCh38
rs770848740 Chr.13: 112067668-112067668 on Build GRCh38
rs753048556 Chr.13: 112067624-112067624 on Build GRCh38
rs758965697 Chr.13: 112067767-112067772 on Build GRCh38
rs769592363 Chr.13: 112067609-112067609 on Build GRCh38
rs1051311080 Chr.13: 112067543-112067543 on Build GRCh38
rs986792793 Chr.13: 112067861-112067861 on Build GRCh38
rs762372894 Chr.13: 112067703-112067703 on Build GRCh38
rs1038529918 Chr.13: 112067559-112067559 on Build GRCh38
rs759581145 Chr.13: 112067749-112067754 on Build GRCh38
rs750745411 Chr.13: 112067767-112067769 on Build GRCh38
rs758332040 Chr.13: 112067974-112067974 on Build GRCh38
rs368703682 Chr.13: 112067820-112067820 on Build GRCh38
rs781269504 Chr.13: 112067742-112067753 on Build GRCh38
rs765176352 Chr.13: 112067970-112067970 on Build GRCh38
rs762899161 Chr.13: 112067611-112067611 on Build GRCh38
rs533444455 Chr.13: 112067567-112067567 on Build GRCh38
rs774027018 Chr.13: 112067790-112067790 on Build GRCh38
rs1004349200 Chr.13: 112067552-112067552 on Build GRCh38
rs146638058 Chr.13: 112067979-112067979 on Build GRCh38
rs867966822 Chr.13: 112067696-112067696 on Build GRCh38
rs781530699 Chr.13: 112067778-112067779 on Build GRCh38
rs940272596 Chr.13: 112067883-112067883 on Build GRCh38
rs749159524 Chr.13: 112067742-112067765 on Build GRCh38
rs776896967 Chr.13: 112067921-112067921 on Build GRCh38
rs552131875 Chr.13: 112067568-112067568 on Build GRCh38
rs755005464 Chr.13: 112067775-112067776 on Build GRCh38
rs530891950 Chr.13: 112067578-112067578 on Build GRCh38
rs200478519 Chr.13: 112067685-112067685 on Build GRCh38
rs757801852 Chr.13: 112067643-112067643 on Build GRCh38
rs751660680 Chr.13: 112067773-112067784 on Build GRCh38
rs908752648 Chr.13: 112067743-112067745 on Build GRCh38
rs1001138683 Chr.13: 112067770-112067770 on Build GRCh38
rs779014597 Chr.13: 112067734-112067734 on Build GRCh38
rs749252099 Chr.13: 112067730-112067730 on Build GRCh38
rs779869825 Chr.13: 112067645-112067645 on Build GRCh38
rs139356700 Chr.13: 112067945-112067945 on Build GRCh38
rs1051790188 Chr.13: 112067742-112067742 on Build GRCh38
rs748481964 Chr.13: 112067928-112067928 on Build GRCh38
rs141390174 Chr.13: 112067988-112067988 on Build GRCh38
rs752557624 Chr.13: 112067715-112067715 on Build GRCh38
rs796868434 Chr.13: 112067556-112067556 on Build GRCh38
rs969826580 Chr.13: 112067579-112067579 on Build GRCh38
rs759813449 Chr.13: 112067671-112067671 on Build GRCh38
rs767290447 Chr.13: 112067802-112067802 on Build GRCh38
rs780331342 Chr.13: 112067898-112067898 on Build GRCh38
rs898226369 Chr.13: 112067569-112067569 on Build GRCh38
rs753476241 Chr.13: 112067761-112067778 on Build GRCh38
rs756999555 Chr.13: 112067952-112067952 on Build GRCh38
rs775081361 Chr.13: 112067755-112067778 on Build GRCh38
rs752223740 Chr.13: 112067805-112067805 on Build GRCh38
rs753646635 Chr.13: 112067829-112067829 on Build GRCh38
rs762213984 Chr.13: 112067766-112067767 on Build GRCh38
rs778783653 Chr.13: 112067873-112067873 on Build GRCh38
rs1035284117 Chr.13: 112067631-112067631 on Build GRCh38
rs751088858 Chr.13: 112067705-112067705 on Build GRCh38
rs745918667 Chr.13: 112067735-112067735 on Build GRCh38
rs774410719 Chr.13: 112067669-112067669 on Build GRCh38
rs758418512 Chr.13: 112067895-112067895 on Build GRCh38
rs1024252507 Chr.13: 112067571-112067571 on Build GRCh38
rs777521736 Chr.13: 112067659-112067659 on Build GRCh38
rs879824266 Chr.13: 112067595-112067595 on Build GRCh38
rs373278017 Chr.13: 112067946-112067946 on Build GRCh38
rs1001889852 Chr.13: 112067755-112067760 on Build GRCh38
rs777771477 Chr.13: 112067726-112067726 on Build GRCh38
rs142328530 Chr.13: 112067856-112067856 on Build GRCh38
rs751319698 Chr.13: 112067653-112067653 on Build GRCh38
rs775874402 Chr.13: 112067683-112067683 on Build GRCh38
rs770444907 Chr.13: 112067932-112067932 on Build GRCh38
rs373543612 Chr.13: 112067778-112067778 on Build GRCh38
rs770831480 Chr.13: 112067743-112067772 on Build GRCh38
rs770843831 Chr.13: 112067733-112067733 on Build GRCh38

View Full Product Details