Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 37591526-37592031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA1
Entrez Gene ID
3169
Gene Name
forkhead box A1
Gene Aliases
HNF3A, TCF3A
UniGene
Hs.163484
Gene Chromosome Location
Chr.14: 37589552-37595120 on Build GRCh38
Transcripts
NM_004496.3 XM_017021246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5423879 c.1173C>A p.D391E Chr.14: 37591611-37591611 on Build GRCh38
6934277 c.1188C>A p.F396L Chr.14: 37591596-37591596 on Build GRCh38
6945894 c.797delT p.F266fs*55 Chr.14: 37591987-37591987 on Build GRCh38
6916846 c.958_979del22 p.L320fs*23 Chr.14: 37591805-37591826 on Build GRCh38
5773482 c.759G>A p.M253I Chr.14: 37592025-37592025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879940115 Chr.14: 37591987-37591987 on Build GRCh38
rs201646206 Chr.14: 37591750-37591750 on Build GRCh38
rs777608823 Chr.14: 37591625-37591625 on Build GRCh38
rs773567107 Chr.14: 37591935-37591936 on Build GRCh38
rs999190675 Chr.14: 37591896-37591896 on Build GRCh38
rs781104468 Chr.14: 37591954-37591954 on Build GRCh38
rs1056757373 Chr.14: 37591902-37591902 on Build GRCh38
rs779832519 Chr.14: 37591926-37591926 on Build GRCh38
rs777832140 Chr.14: 37591775-37591775 on Build GRCh38
rs772117929 Chr.14: 37592007-37592007 on Build GRCh38
rs1016447126 Chr.14: 37591910-37591910 on Build GRCh38
rs1035265550 Chr.14: 37591743-37591743 on Build GRCh38
rs780031833 Chr.14: 37591668-37591668 on Build GRCh38
rs556143176 Chr.14: 37591789-37591789 on Build GRCh38
rs962828688 Chr.14: 37591967-37591967 on Build GRCh38
rs748980881 Chr.14: 37591629-37591629 on Build GRCh38
rs201764769 Chr.14: 37591776-37591776 on Build GRCh38
rs764636331 Chr.14: 37591872-37591872 on Build GRCh38
rs759868036 Chr.14: 37591821-37591821 on Build GRCh38
rs779670401 Chr.14: 37591755-37591755 on Build GRCh38
rs926719052 Chr.14: 37591808-37591808 on Build GRCh38
rs911805778 Chr.14: 37591815-37591815 on Build GRCh38
rs763172562 Chr.14: 37591685-37591685 on Build GRCh38
rs768062390 Chr.14: 37591609-37591609 on Build GRCh38
rs547183484 Chr.14: 37591614-37591614 on Build GRCh38
rs1019963817 Chr.14: 37591594-37591594 on Build GRCh38
rs867404999 Chr.14: 37591714-37591714 on Build GRCh38
rs537882736 Chr.14: 37591841-37591841 on Build GRCh38
rs568751362 Chr.14: 37591804-37591804 on Build GRCh38
rs200879514 Chr.14: 37591581-37591581 on Build GRCh38
rs903095058 Chr.14: 37591862-37591862 on Build GRCh38
rs1884802 Chr.14: 37591848-37591848 on Build GRCh38
rs746967980 Chr.14: 37591757-37591757 on Build GRCh38
rs963090162 Chr.14: 37591783-37591783 on Build GRCh38
rs771973050 Chr.14: 37591659-37591659 on Build GRCh38
rs138390380 Chr.14: 37591642-37591642 on Build GRCh38
rs762342618 Chr.14: 37591606-37591607 on Build GRCh38
rs944534633 Chr.14: 37591560-37591560 on Build GRCh38
rs775955726 Chr.14: 37591849-37591849 on Build GRCh38
rs749411524 Chr.14: 37591956-37591956 on Build GRCh38
rs756922846 Chr.14: 37591635-37591635 on Build GRCh38
rs567083055 Chr.14: 37591751-37591751 on Build GRCh38
rs751891516 Chr.14: 37591584-37591584 on Build GRCh38
rs1011397768 Chr.14: 37591854-37591854 on Build GRCh38
rs774644296 Chr.14: 37591689-37591689 on Build GRCh38
rs769889535 Chr.14: 37591671-37591671 on Build GRCh38
rs902772971 Chr.14: 37591676-37591676 on Build GRCh38
rs753224813 Chr.14: 37591715-37591715 on Build GRCh38
rs757337935 Chr.14: 37591978-37591978 on Build GRCh38
rs974119009 Chr.14: 37591795-37591795 on Build GRCh38
rs763720398 Chr.14: 37591737-37591737 on Build GRCh38
rs758194732 Chr.14: 37591754-37591754 on Build GRCh38
rs997215836 Chr.14: 37591876-37591876 on Build GRCh38
rs946422204 Chr.14: 37591860-37591860 on Build GRCh38
rs529354784 Chr.14: 37591653-37591653 on Build GRCh38
rs755443008 Chr.14: 37591588-37591588 on Build GRCh38
rs896728690 Chr.14: 37591919-37591919 on Build GRCh38
rs759930701 Chr.14: 37591691-37591691 on Build GRCh38
rs879499034 Chr.14: 37591986-37591986 on Build GRCh38
rs1024200279 Chr.14: 37591966-37591966 on Build GRCh38
rs764833346 Chr.14: 37591905-37591905 on Build GRCh38
rs778808721 Chr.14: 37591979-37591979 on Build GRCh38
rs765506027 Chr.14: 37591548-37591548 on Build GRCh38
rs767958377 Chr.14: 37591701-37591701 on Build GRCh38
rs35391958 Chr.14: 37591759-37591759 on Build GRCh38
rs1010598209 Chr.14: 37591639-37591639 on Build GRCh38
rs754968432 Chr.14: 37591951-37591951 on Build GRCh38
rs944578235 Chr.14: 37591830-37591830 on Build GRCh38
rs867473250 Chr.14: 37591903-37591903 on Build GRCh38
rs751299919 Chr.14: 37591928-37591928 on Build GRCh38
rs753013275 Chr.14: 37591859-37591859 on Build GRCh38
rs145896666 Chr.14: 37591575-37591575 on Build GRCh38
rs778587980 Chr.14: 37591648-37591648 on Build GRCh38
rs1030123417 Chr.14: 37591683-37591683 on Build GRCh38
rs773225075 Chr.14: 37591681-37591681 on Build GRCh38
rs1012429678 Chr.14: 37591962-37591962 on Build GRCh38
rs750117744 Chr.14: 37591915-37591915 on Build GRCh38
rs761225339 Chr.14: 37591730-37591730 on Build GRCh38
rs577631417 Chr.14: 37591894-37591894 on Build GRCh38
rs746132159 Chr.14: 37591983-37591983 on Build GRCh38
rs35342092 Chr.14: 37591563-37591563 on Build GRCh38
rs770099992 Chr.14: 37591933-37591933 on Build GRCh38
rs999746832 Chr.14: 37591734-37591734 on Build GRCh38
rs756977898 Chr.14: 37591748-37591748 on Build GRCh38
rs533489074 Chr.14: 37591612-37591612 on Build GRCh38
rs983365702 Chr.14: 37591810-37591810 on Build GRCh38
rs868027915 Chr.14: 37591756-37591756 on Build GRCh38
rs771116697 Chr.14: 37591688-37591688 on Build GRCh38
rs867182904 Chr.14: 37591670-37591670 on Build GRCh38
rs753342877 Chr.14: 37591742-37591742 on Build GRCh38
rs753453381 Chr.14: 37591883-37591883 on Build GRCh38
rs950642604 Chr.14: 37591809-37591809 on Build GRCh38
rs757951191 Chr.14: 37591916-37591916 on Build GRCh38

View Full Product Details