Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75958302-75958819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB3
Entrez Gene ID
7043
Gene Name
transforming growth factor beta 3
Gene Aliases
ARVD, ARVD1, LDS5, RNHF, TGF-beta3
UniGene
Hs.592317
Gene Chromosome Location
Chr.14: 75958097-75982022 on Build GRCh38
Transcripts
NM_003239.3

SNP Detail

SNP ID Chromosome Location
rs912129834 Chr.14: 75958439-75958439 on Build GRCh38
rs900494519 Chr.14: 75958599-75958599 on Build GRCh38
rs921896477 Chr.14: 75958769-75958769 on Build GRCh38
rs3917218 Chr.14: 75958367-75958367 on Build GRCh38
rs375812472 Chr.14: 75958608-75958608 on Build GRCh38
rs4252349 Chr.14: 75958505-75958505 on Build GRCh38
rs549051269 Chr.14: 75958705-75958706 on Build GRCh38
rs945861518 Chr.14: 75958607-75958607 on Build GRCh38
rs10129478 Chr.14: 75958491-75958491 on Build GRCh38
rs776109060 Chr.14: 75958685-75958688 on Build GRCh38
rs936061729 Chr.14: 75958526-75958526 on Build GRCh38
rs1026852691 Chr.14: 75958334-75958334 on Build GRCh38
rs1026883642 Chr.14: 75958636-75958636 on Build GRCh38
rs748634626 Chr.14: 75958591-75958591 on Build GRCh38
rs780261726 Chr.14: 75958419-75958419 on Build GRCh38
rs1042587 Chr.14: 75958654-75958654 on Build GRCh38
rs995223269 Chr.14: 75958655-75958655 on Build GRCh38
rs926152720 Chr.14: 75958497-75958497 on Build GRCh38
rs992986410 Chr.14: 75958738-75958738 on Build GRCh38
rs755049637 Chr.14: 75958520-75958520 on Build GRCh38
rs949012333 Chr.14: 75958482-75958482 on Build GRCh38
rs1001605472 Chr.14: 75958348-75958348 on Build GRCh38
rs554651460 Chr.14: 75958590-75958590 on Build GRCh38
rs1055444737 Chr.14: 75958644-75958644 on Build GRCh38
rs199646113 Chr.14: 75958705-75958705 on Build GRCh38
rs1054470418 Chr.14: 75958546-75958546 on Build GRCh38
rs61762491 Chr.14: 75958686-75958689 on Build GRCh38
rs770849223 Chr.14: 75958620-75958621 on Build GRCh38
rs3917217 Chr.14: 75958691-75958691 on Build GRCh38
rs1015808317 Chr.14: 75958675-75958675 on Build GRCh38
rs915524065 Chr.14: 75958452-75958452 on Build GRCh38
rs201078179 Chr.14: 75958705-75958705 on Build GRCh38
rs183740022 Chr.14: 75958674-75958674 on Build GRCh38
rs886050793 Chr.14: 75958682-75958682 on Build GRCh38
rs560085664 Chr.14: 75958357-75958357 on Build GRCh38
rs387906514 Chr.14: 75958692-75958692 on Build GRCh38
rs992514022 Chr.14: 75958420-75958420 on Build GRCh38
rs1027786497 Chr.14: 75958375-75958375 on Build GRCh38
rs944756712 Chr.14: 75958577-75958577 on Build GRCh38
rs966664355 Chr.14: 75958736-75958736 on Build GRCh38
rs950958552 Chr.14: 75958405-75958405 on Build GRCh38
rs116586110 Chr.14: 75958363-75958363 on Build GRCh38
rs907095469 Chr.14: 75958612-75958612 on Build GRCh38
rs35290094 Chr.14: 75958370-75958370 on Build GRCh38
rs745971119 Chr.14: 75958347-75958347 on Build GRCh38
rs568012370 Chr.14: 75958668-75958668 on Build GRCh38
rs4252348 Chr.14: 75958521-75958521 on Build GRCh38
rs552155925 Chr.14: 75958600-75958600 on Build GRCh38
rs780607751 Chr.14: 75958685-75958685 on Build GRCh38
rs868168827 Chr.14: 75958331-75958331 on Build GRCh38

View Full Product Details