Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100376142-100376672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WARS
Entrez Gene ID
7453
Gene Name
tryptophanyl-tRNA synthetase
Gene Aliases
GAMMA-2, IFI53, IFP53
UniGene
Hs.497599
Gene Chromosome Location
Chr.14: 100333788-100376343 on Build GRCh38
Transcripts
NM_173701.1 NM_213645.1 XM_011537133.2 XM_017021627.1 XM_017021629.1 NM_001161476.1 NM_024515.4 XM_005268056.4 XM_005268057.3 XM_005268058.4 XM_011537150.2

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
UniGene
Hs.497600
Gene Chromosome Location
Chr.14: 100376410-100530303 on Build GRCh38
Transcripts
NM_173701.1 NM_213645.1 XM_011537133.2 XM_017021627.1 XM_017021629.1 NM_001161476.1 NM_024515.4 XM_005268056.4 XM_005268057.3 XM_005268058.4 XM_011537150.2

SNP Detail

SNP ID Chromosome Location
rs1039263708 Chr.14: 100376267-100376267 on Build GRCh38
rs947011088 Chr.14: 100376417-100376417 on Build GRCh38
rs1023485168 Chr.14: 100376331-100376331 on Build GRCh38
rs552253787 Chr.14: 100376375-100376400 on Build GRCh38
rs952358257 Chr.14: 100376536-100376536 on Build GRCh38
rs368395425 Chr.14: 100376214-100376214 on Build GRCh38
rs940803777 Chr.14: 100376383-100376383 on Build GRCh38
rs966176691 Chr.14: 100376388-100376388 on Build GRCh38
rs986531856 Chr.14: 100376368-100376368 on Build GRCh38
rs561694454 Chr.14: 100376550-100376550 on Build GRCh38
rs922161268 Chr.14: 100376561-100376561 on Build GRCh38
rs922163298 Chr.14: 100376409-100376409 on Build GRCh38
rs1056405171 Chr.14: 100376640-100376640 on Build GRCh38
rs2273802 Chr.14: 100376562-100376562 on Build GRCh38
rs962672585 Chr.14: 100376337-100376337 on Build GRCh38
rs1056436607 Chr.14: 100376431-100376431 on Build GRCh38
rs2273804 Chr.14: 100376300-100376300 on Build GRCh38
rs963814795 Chr.14: 100376544-100376544 on Build GRCh38
rs773799113 Chr.14: 100376278-100376278 on Build GRCh38
rs966481561 Chr.14: 100376319-100376319 on Build GRCh38
rs550038194 Chr.14: 100376632-100376632 on Build GRCh38
rs60819702 Chr.14: 100376571-100376571 on Build GRCh38
rs564144777 Chr.14: 100376579-100376579 on Build GRCh38
rs922215419 Chr.14: 100376346-100376346 on Build GRCh38
rs572671553 Chr.14: 100376404-100376404 on Build GRCh38
rs995860087 Chr.14: 100376524-100376524 on Build GRCh38
rs894941349 Chr.14: 100376299-100376299 on Build GRCh38
rs570567497 Chr.14: 100376279-100376279 on Build GRCh38
rs561530686 Chr.14: 100376192-100376192 on Build GRCh38
rs577898050 Chr.14: 100376511-100376511 on Build GRCh38
rs924193504 Chr.14: 100376594-100376594 on Build GRCh38
rs912098170 Chr.14: 100376241-100376241 on Build GRCh38
rs973783797 Chr.14: 100376166-100376166 on Build GRCh38
rs975369889 Chr.14: 100376340-100376340 on Build GRCh38
rs974887668 Chr.14: 100376559-100376559 on Build GRCh38
rs962315822 Chr.14: 100376164-100376164 on Build GRCh38
rs533439683 Chr.14: 100376581-100376581 on Build GRCh38
rs940687913 Chr.14: 100376206-100376206 on Build GRCh38
rs984905509 Chr.14: 100376556-100376556 on Build GRCh38
rs977737947 Chr.14: 100376321-100376321 on Build GRCh38
rs770458794 Chr.14: 100376250-100376250 on Build GRCh38
rs1055415862 Chr.14: 100376438-100376438 on Build GRCh38
rs562483045 Chr.14: 100376476-100376476 on Build GRCh38
rs555072357 Chr.14: 100376186-100376186 on Build GRCh38
rs1033616618 Chr.14: 100376284-100376284 on Build GRCh38
rs935339534 Chr.14: 100376430-100376430 on Build GRCh38
rs1043760015 Chr.14: 100376294-100376294 on Build GRCh38
rs750345565 Chr.14: 100376622-100376623 on Build GRCh38
rs550117508 Chr.14: 100376311-100376311 on Build GRCh38
rs1029650651 Chr.14: 100376534-100376534 on Build GRCh38
rs965719721 Chr.14: 100376339-100376339 on Build GRCh38
rs776083912 Chr.14: 100376334-100376334 on Build GRCh38
rs1025332904 Chr.14: 100376347-100376347 on Build GRCh38
rs985011048 Chr.14: 100376365-100376365 on Build GRCh38
rs2273803 Chr.14: 100376441-100376441 on Build GRCh38
rs527525096 Chr.14: 100376207-100376207 on Build GRCh38
rs761031986 Chr.14: 100376474-100376474 on Build GRCh38
rs966064780 Chr.14: 100376517-100376517 on Build GRCh38
rs536128437 Chr.14: 100376379-100376379 on Build GRCh38
rs907952992 Chr.14: 100376378-100376378 on Build GRCh38
rs1044904918 Chr.14: 100376443-100376443 on Build GRCh38
rs1044113070 Chr.14: 100376419-100376419 on Build GRCh38
rs535176780 Chr.14: 100376421-100376421 on Build GRCh38
rs896595530 Chr.14: 100376439-100376439 on Build GRCh38
rs981591272 Chr.14: 100376528-100376528 on Build GRCh38
rs984007607 Chr.14: 100376216-100376216 on Build GRCh38
rs569975880 Chr.14: 100376354-100376354 on Build GRCh38
rs1018644411 Chr.14: 100376329-100376329 on Build GRCh38
rs545602144 Chr.14: 100376447-100376447 on Build GRCh38
rs1043606615 Chr.14: 100376475-100376475 on Build GRCh38
rs928723742 Chr.14: 100376593-100376593 on Build GRCh38
rs886564878 Chr.14: 100376197-100376197 on Build GRCh38
rs189291792 Chr.14: 100376393-100376393 on Build GRCh38
rs547484743 Chr.14: 100376260-100376260 on Build GRCh38
rs531605171 Chr.14: 100376171-100376171 on Build GRCh38
rs944862558 Chr.14: 100376253-100376253 on Build GRCh38
rs1022048817 Chr.14: 100376301-100376301 on Build GRCh38
rs928612445 Chr.14: 100376397-100376397 on Build GRCh38
rs113911074 Chr.14: 100376428-100376428 on Build GRCh38
rs906443324 Chr.14: 100376275-100376275 on Build GRCh38
rs541712238 Chr.14: 100376535-100376535 on Build GRCh38
rs202223402 Chr.14: 100376248-100376249 on Build GRCh38
rs999253097 Chr.14: 100376485-100376485 on Build GRCh38

View Full Product Details