Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65150355-65150848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPX
Entrez Gene ID
10845
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases
-
UniGene
Hs.113823
Gene Chromosome Location
Chr.15: 65148219-65185420 on Build GRCh38
Transcripts
NM_006660.4 NR_133680.1 XM_011521164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919575 c.1901A>G p.*634* Chr.15: 65150824-65150824 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs961728075 Chr.15: 65150777-65150777 on Build GRCh38
rs958598460 Chr.15: 65150518-65150518 on Build GRCh38
rs952483868 Chr.15: 65150636-65150636 on Build GRCh38
rs759503919 Chr.15: 65150713-65150713 on Build GRCh38
rs984832291 Chr.15: 65150760-65150760 on Build GRCh38
rs1003064151 Chr.15: 65150733-65150733 on Build GRCh38
rs996906024 Chr.15: 65150616-65150616 on Build GRCh38
rs781494673 Chr.15: 65150573-65150574 on Build GRCh38
rs757174098 Chr.15: 65150805-65150805 on Build GRCh38
rs942666761 Chr.15: 65150618-65150618 on Build GRCh38
rs1033654836 Chr.15: 65150410-65150411 on Build GRCh38
rs368753744 Chr.15: 65150827-65150827 on Build GRCh38
rs749201029 Chr.15: 65150801-65150801 on Build GRCh38
rs749171896 Chr.15: 65150673-65150673 on Build GRCh38
rs1046711911 Chr.15: 65150407-65150407 on Build GRCh38
rs567272403 Chr.15: 65150688-65150688 on Build GRCh38
rs112243325 Chr.15: 65150406-65150406 on Build GRCh38
rs984494046 Chr.15: 65150520-65150520 on Build GRCh38
rs893401197 Chr.15: 65150418-65150418 on Build GRCh38
rs893882240 Chr.15: 65150561-65150561 on Build GRCh38
rs777474164 Chr.15: 65150784-65150784 on Build GRCh38
rs569207573 Chr.15: 65150645-65150645 on Build GRCh38
rs375348760 Chr.15: 65150817-65150817 on Build GRCh38
rs778320231 Chr.15: 65150810-65150810 on Build GRCh38
rs765916502 Chr.15: 65150560-65150560 on Build GRCh38
rs114931353 Chr.15: 65150646-65150646 on Build GRCh38
rs938054268 Chr.15: 65150729-65150729 on Build GRCh38
rs530104331 Chr.15: 65150413-65150413 on Build GRCh38
rs915220663 Chr.15: 65150808-65150808 on Build GRCh38
rs532577668 Chr.15: 65150574-65150574 on Build GRCh38
rs867285511 Chr.15: 65150527-65150527 on Build GRCh38
rs376746728 Chr.15: 65150511-65150523 on Build GRCh38
rs560268279 Chr.15: 65150573-65150573 on Build GRCh38
rs771902132 Chr.15: 65150813-65150813 on Build GRCh38
rs965109734 Chr.15: 65150615-65150615 on Build GRCh38
rs138843437 Chr.15: 65150739-65150739 on Build GRCh38
rs368299030 Chr.15: 65150516-65150516 on Build GRCh38
rs867850478 Chr.15: 65150607-65150607 on Build GRCh38
rs961156382 Chr.15: 65150674-65150674 on Build GRCh38
rs73475043 Chr.15: 65150725-65150725 on Build GRCh38
rs1009158449 Chr.15: 65150599-65150599 on Build GRCh38
rs773799392 Chr.15: 65150598-65150598 on Build GRCh38
rs762310496 Chr.15: 65150464-65150464 on Build GRCh38
rs752543714 Chr.15: 65150774-65150774 on Build GRCh38
rs550293779 Chr.15: 65150672-65150672 on Build GRCh38
rs1019573210 Chr.15: 65150608-65150608 on Build GRCh38
rs201623357 Chr.15: 65150811-65150811 on Build GRCh38
rs981179140 Chr.15: 65150514-65150514 on Build GRCh38
rs1020581330 Chr.15: 65150717-65150717 on Build GRCh38
rs1028099635 Chr.15: 65150625-65150625 on Build GRCh38
rs938008635 Chr.15: 65150475-65150475 on Build GRCh38
rs1042459848 Chr.15: 65150816-65150816 on Build GRCh38
rs971592783 Chr.15: 65150507-65150508 on Build GRCh38
rs552396503 Chr.15: 65150631-65150631 on Build GRCh38
rs958497177 Chr.15: 65150760-65150760 on Build GRCh38
rs566610862 Chr.15: 65150791-65150791 on Build GRCh38
rs561369024 Chr.15: 65150380-65150380 on Build GRCh38
rs755896226 Chr.15: 65150779-65150779 on Build GRCh38
rs762937599 Chr.15: 65150782-65150784 on Build GRCh38
rs1034130070 Chr.15: 65150515-65150515 on Build GRCh38

View Full Product Details