Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8674327-8674827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABAT
Entrez Gene ID
18
Gene Name
4-aminobutyrate aminotransferase
Gene Aliases
GABA-AT, GABAT, NPD009
UniGene
Hs.336768
Gene Chromosome Location
Chr.16: 8674587-8784575 on Build GRCh38
Transcripts
NM_020686.5

SNP Detail

SNP ID Chromosome Location
rs547209295 Chr.16: 8674782-8674782 on Build GRCh38
rs941138625 Chr.16: 8674518-8674518 on Build GRCh38
rs559387602 Chr.16: 8674802-8674802 on Build GRCh38
rs982894944 Chr.16: 8674468-8674468 on Build GRCh38
rs907424505 Chr.16: 8674442-8674442 on Build GRCh38
rs867307243 Chr.16: 8674396-8674396 on Build GRCh38
rs574895183 Chr.16: 8674432-8674432 on Build GRCh38
rs937622924 Chr.16: 8674626-8674626 on Build GRCh38
rs564419998 Chr.16: 8674607-8674607 on Build GRCh38
rs985384087 Chr.16: 8674531-8674531 on Build GRCh38
rs912382849 Chr.16: 8674653-8674653 on Build GRCh38
rs961282282 Chr.16: 8674624-8674624 on Build GRCh38
rs983043168 Chr.16: 8674631-8674631 on Build GRCh38
rs891075601 Chr.16: 8674540-8674540 on Build GRCh38
rs989914531 Chr.16: 8674761-8674761 on Build GRCh38
rs1052890574 Chr.16: 8674546-8674546 on Build GRCh38
rs936118847 Chr.16: 8674393-8674393 on Build GRCh38
rs972645494 Chr.16: 8674349-8674349 on Build GRCh38
rs183460230 Chr.16: 8674595-8674595 on Build GRCh38
rs989567192 Chr.16: 8674403-8674403 on Build GRCh38
rs905327166 Chr.16: 8674756-8674756 on Build GRCh38
rs1030996889 Chr.16: 8674389-8674389 on Build GRCh38
rs962384025 Chr.16: 8674645-8674645 on Build GRCh38
rs1056048537 Chr.16: 8674644-8674644 on Build GRCh38
rs961037316 Chr.16: 8674491-8674492 on Build GRCh38
rs969503838 Chr.16: 8674448-8674448 on Build GRCh38
rs546011808 Chr.16: 8674597-8674597 on Build GRCh38
rs993914810 Chr.16: 8674680-8674680 on Build GRCh38
rs1015819506 Chr.16: 8674805-8674805 on Build GRCh38
rs1038109671 Chr.16: 8674528-8674528 on Build GRCh38
rs892447213 Chr.16: 8674554-8674554 on Build GRCh38
rs981002102 Chr.16: 8674458-8674458 on Build GRCh38
rs1029599659 Chr.16: 8674519-8674519 on Build GRCh38
rs932386524 Chr.16: 8674650-8674650 on Build GRCh38
rs886052387 Chr.16: 8674603-8674603 on Build GRCh38
rs1002289043 Chr.16: 8674599-8674599 on Build GRCh38
rs541932326 Chr.16: 8674436-8674436 on Build GRCh38
rs990197857 Chr.16: 8674567-8674567 on Build GRCh38
rs1010963814 Chr.16: 8674558-8674558 on Build GRCh38
rs1001774880 Chr.16: 8674453-8674453 on Build GRCh38
rs1022306303 Chr.16: 8674581-8674581 on Build GRCh38
rs1007258286 Chr.16: 8674726-8674726 on Build GRCh38
rs551953775 Chr.16: 8674768-8674768 on Build GRCh38
rs1043794082 Chr.16: 8674753-8674753 on Build GRCh38
rs531589337 Chr.16: 8674646-8674646 on Build GRCh38
rs908195129 Chr.16: 8674502-8674502 on Build GRCh38
rs908231030 Chr.16: 8674643-8674643 on Build GRCh38
rs543457242 Chr.16: 8674649-8674649 on Build GRCh38
rs779210592 Chr.16: 8674762-8674762 on Build GRCh38
rs928639371 Chr.16: 8674625-8674625 on Build GRCh38
rs998773352 Chr.16: 8674544-8674544 on Build GRCh38
rs932445804 Chr.16: 8674538-8674538 on Build GRCh38
rs1016652233 Chr.16: 8674490-8674490 on Build GRCh38
rs928187333 Chr.16: 8674464-8674464 on Build GRCh38
rs890468188 Chr.16: 8674373-8674373 on Build GRCh38
rs532269694 Chr.16: 8674701-8674701 on Build GRCh38
rs35440533 Chr.16: 8674371-8674371 on Build GRCh38
rs899686904 Chr.16: 8674534-8674534 on Build GRCh38
rs1051010107 Chr.16: 8674539-8674539 on Build GRCh38
rs1050946041 Chr.16: 8674652-8674652 on Build GRCh38
rs757633507 Chr.16: 8674742-8674742 on Build GRCh38
rs1035193070 Chr.16: 8674617-8674617 on Build GRCh38
rs1045991223 Chr.16: 8674439-8674439 on Build GRCh38
rs1018108908 Chr.16: 8674743-8674743 on Build GRCh38
rs561638701 Chr.16: 8674663-8674663 on Build GRCh38
rs537848639 Chr.16: 8674379-8674379 on Build GRCh38
rs1730963 Chr.16: 8674465-8674465 on Build GRCh38
rs1010827928 Chr.16: 8674752-8674752 on Build GRCh38
rs556101217 Chr.16: 8674381-8674381 on Build GRCh38

View Full Product Details