Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29995231-29995773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
969743 c.186+1G>A p.? Chr.16: 29995342-29995342 on Build GRCh38
328732 c.13A>G p.K5E Chr.16: 29995593-29995593 on Build GRCh38
969744 c.136_138delGAG p.E46delE Chr.16: 29995391-29995393 on Build GRCh38
1609192 c.66-5T>G p.? Chr.16: 29995468-29995468 on Build GRCh38
6044637 c.169_171delGAG p.E57delE Chr.16: 29995358-29995360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780302466 Chr.16: 29995531-29995531 on Build GRCh38
rs964100504 Chr.16: 29995714-29995714 on Build GRCh38
rs779589307 Chr.16: 29995525-29995525 on Build GRCh38
rs571949113 Chr.16: 29995631-29995631 on Build GRCh38
rs751218565 Chr.16: 29995564-29995564 on Build GRCh38
rs760488550 Chr.16: 29995440-29995440 on Build GRCh38
rs773062940 Chr.16: 29995561-29995561 on Build GRCh38
rs892132949 Chr.16: 29995465-29995465 on Build GRCh38
rs937724744 Chr.16: 29995740-29995740 on Build GRCh38
rs898672436 Chr.16: 29995687-29995687 on Build GRCh38
rs372142050 Chr.16: 29995617-29995617 on Build GRCh38
rs765862451 Chr.16: 29995562-29995562 on Build GRCh38
rs760413972 Chr.16: 29995323-29995323 on Build GRCh38
rs559367516 Chr.16: 29995642-29995642 on Build GRCh38
rs537101913 Chr.16: 29995721-29995721 on Build GRCh38
rs996129512 Chr.16: 29995691-29995691 on Build GRCh38
rs914460953 Chr.16: 29995680-29995680 on Build GRCh38
rs926312102 Chr.16: 29995739-29995739 on Build GRCh38
rs912669986 Chr.16: 29995272-29995272 on Build GRCh38
rs540649865 Chr.16: 29995281-29995283 on Build GRCh38
rs887378987 Chr.16: 29995705-29995705 on Build GRCh38
rs1055918273 Chr.16: 29995532-29995532 on Build GRCh38
rs761669772 Chr.16: 29995459-29995459 on Build GRCh38
rs770286943 Chr.16: 29995266-29995266 on Build GRCh38
rs1047305436 Chr.16: 29995694-29995694 on Build GRCh38
rs200825231 Chr.16: 29995619-29995619 on Build GRCh38
rs755898184 Chr.16: 29995644-29995644 on Build GRCh38
rs62057684 Chr.16: 29995498-29995498 on Build GRCh38
rs769675336 Chr.16: 29995554-29995554 on Build GRCh38
rs1003059588 Chr.16: 29995361-29995361 on Build GRCh38
rs749343900 Chr.16: 29995301-29995301 on Build GRCh38
rs773678589 Chr.16: 29995609-29995609 on Build GRCh38
rs757792570 Chr.16: 29995365-29995365 on Build GRCh38
rs759046093 Chr.16: 29995571-29995571 on Build GRCh38
rs942505711 Chr.16: 29995281-29995281 on Build GRCh38
rs781420621 Chr.16: 29995596-29995596 on Build GRCh38
rs755086752 Chr.16: 29995638-29995638 on Build GRCh38
rs986909087 Chr.16: 29995261-29995261 on Build GRCh38
rs776986383 Chr.16: 29995727-29995727 on Build GRCh38
rs374538636 Chr.16: 29995256-29995256 on Build GRCh38
rs770726112 Chr.16: 29995651-29995651 on Build GRCh38
rs755291709 Chr.16: 29995263-29995263 on Build GRCh38
rs370809064 Chr.16: 29995671-29995671 on Build GRCh38
rs1052506585 Chr.16: 29995336-29995336 on Build GRCh38
rs367786840 Chr.16: 29995530-29995530 on Build GRCh38
rs949121006 Chr.16: 29995467-29995467 on Build GRCh38
rs894650624 Chr.16: 29995692-29995692 on Build GRCh38
rs988419666 Chr.16: 29995731-29995731 on Build GRCh38
rs756484175 Chr.16: 29995487-29995487 on Build GRCh38
rs375289843 Chr.16: 29995309-29995309 on Build GRCh38
rs777362891 Chr.16: 29995647-29995647 on Build GRCh38
rs913801074 Chr.16: 29995733-29995733 on Build GRCh38
rs767641089 Chr.16: 29995318-29995318 on Build GRCh38
rs771132004 Chr.16: 29995394-29995394 on Build GRCh38
rs990132345 Chr.16: 29995560-29995560 on Build GRCh38
rs751550181 Chr.16: 29995636-29995636 on Build GRCh38
rs1046510975 Chr.16: 29995659-29995659 on Build GRCh38
rs768150257 Chr.16: 29995591-29995593 on Build GRCh38
rs773882611 Chr.16: 29995268-29995268 on Build GRCh38
rs758495523 Chr.16: 29995590-29995590 on Build GRCh38
rs779536055 Chr.16: 29995589-29995589 on Build GRCh38
rs774378518 Chr.16: 29995655-29995655 on Build GRCh38
rs762864243 Chr.16: 29995602-29995602 on Build GRCh38
rs563156257 Chr.16: 29995335-29995335 on Build GRCh38
rs757727801 Chr.16: 29995588-29995588 on Build GRCh38
rs745925567 Chr.16: 29995526-29995526 on Build GRCh38
rs913841475 Chr.16: 29995652-29995652 on Build GRCh38
rs151257215 Chr.16: 29995424-29995424 on Build GRCh38
rs544787245 Chr.16: 29995260-29995260 on Build GRCh38
rs1003113623 Chr.16: 29995654-29995654 on Build GRCh38
rs780067094 Chr.16: 29995591-29995591 on Build GRCh38
rs776538269 Chr.16: 29995454-29995454 on Build GRCh38
rs746279625 Chr.16: 29995367-29995368 on Build GRCh38
rs926362842 Chr.16: 29995665-29995665 on Build GRCh38
rs764053367 Chr.16: 29995331-29995331 on Build GRCh38
rs995734052 Chr.16: 29995581-29995581 on Build GRCh38
rs761614626 Chr.16: 29995553-29995553 on Build GRCh38
rs754207828 Chr.16: 29995582-29995582 on Build GRCh38
rs773186219 Chr.16: 29995601-29995601 on Build GRCh38
rs749865907 Chr.16: 29995380-29995380 on Build GRCh38
rs981772725 Chr.16: 29995736-29995736 on Build GRCh38
rs775300654 Chr.16: 29995322-29995322 on Build GRCh38
rs748501108 Chr.16: 29995365-29995365 on Build GRCh38
rs754031936 Chr.16: 29995629-29995629 on Build GRCh38
rs138012549 Chr.16: 29995593-29995593 on Build GRCh38
rs1032144071 Chr.16: 29995724-29995724 on Build GRCh38
rs776299420 Chr.16: 29995616-29995634 on Build GRCh38
rs763928002 Chr.16: 29995445-29995445 on Build GRCh38
rs778347328 Chr.16: 29995495-29995495 on Build GRCh38
rs1001640977 Chr.16: 29995506-29995506 on Build GRCh38
rs1019675345 Chr.16: 29995603-29995603 on Build GRCh38
rs996935872 Chr.16: 29995720-29995720 on Build GRCh38
rs749938559 Chr.16: 29995359-29995359 on Build GRCh38
rs771608969 Chr.16: 29995391-29995393 on Build GRCh38
rs200916858 Chr.16: 29995368-29995368 on Build GRCh38
rs936584951 Chr.16: 29995334-29995334 on Build GRCh38
rs759906736 Chr.16: 29995316-29995316 on Build GRCh38
rs754261304 Chr.16: 29995518-29995518 on Build GRCh38
rs150353935 Chr.16: 29995475-29995475 on Build GRCh38
rs778398805 Chr.16: 29995371-29995371 on Build GRCh38
rs761337804 Chr.16: 29995290-29995292 on Build GRCh38
rs1011992528 Chr.16: 29995461-29995461 on Build GRCh38
rs776350320 Chr.16: 29995540-29995540 on Build GRCh38
rs551166516 Chr.16: 29995710-29995710 on Build GRCh38
rs758010643 Chr.16: 29995362-29995362 on Build GRCh38
rs765317327 Chr.16: 29995647-29995649 on Build GRCh38
rs745340206 Chr.16: 29995656-29995656 on Build GRCh38
rs770904204 Chr.16: 29995604-29995604 on Build GRCh38
rs748426149 Chr.16: 29995643-29995643 on Build GRCh38
rs761816007 Chr.16: 29995637-29995637 on Build GRCh38
rs771180897 Chr.16: 29995306-29995306 on Build GRCh38
rs749123627 Chr.16: 29995649-29995649 on Build GRCh38
rs775605788 Chr.16: 29995438-29995438 on Build GRCh38
rs549214690 Chr.16: 29995573-29995573 on Build GRCh38
rs1055867370 Chr.16: 29995686-29995686 on Build GRCh38
rs925175337 Chr.16: 29995325-29995325 on Build GRCh38
rs745951353 Chr.16: 29995396-29995396 on Build GRCh38
rs79852981 Chr.16: 29995635-29995635 on Build GRCh38
rs767147321 Chr.16: 29995301-29995302 on Build GRCh38
rs757784789 Chr.16: 29995520-29995520 on Build GRCh38
rs747902326 Chr.16: 29995597-29995597 on Build GRCh38
rs751216828 Chr.16: 29995486-29995486 on Build GRCh38
rs765172291 Chr.16: 29995460-29995460 on Build GRCh38
rs957849852 Chr.16: 29995725-29995725 on Build GRCh38
rs779217282 Chr.16: 29995365-29995366 on Build GRCh38
rs781551200 Chr.16: 29995265-29995265 on Build GRCh38
rs768813952 Chr.16: 29995594-29995594 on Build GRCh38
rs775277813 Chr.16: 29995469-29995469 on Build GRCh38
rs756792861 Chr.16: 29995255-29995255 on Build GRCh38
rs187781939 Chr.16: 29995712-29995712 on Build GRCh38
rs764793156 Chr.16: 29995357-29995357 on Build GRCh38
rs762249768 Chr.16: 29995620-29995620 on Build GRCh38
rs1135661 Chr.16: 29995356-29995356 on Build GRCh38
rs898724670 Chr.16: 29995552-29995552 on Build GRCh38
rs746642279 Chr.16: 29995579-29995581 on Build GRCh38
rs747255129 Chr.16: 29995536-29995536 on Build GRCh38
rs971082185 Chr.16: 29995734-29995734 on Build GRCh38
rs766929180 Chr.16: 29995634-29995634 on Build GRCh38
rs528467410 Chr.16: 29995614-29995614 on Build GRCh38
rs768868878 Chr.16: 29995538-29995538 on Build GRCh38
rs766019347 Chr.16: 29995477-29995477 on Build GRCh38

View Full Product Details