Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57020820-57021314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
UniGene
Hs.528836
Gene Chromosome Location
Chr.16: 56989547-57083524 on Build GRCh38
Transcripts
NM_032206.4 XM_005256193.2 XM_005256194.3 XM_005256195.2 XM_005256196.2 XM_005256197.2 XM_005256199.3 XM_005256201.3 XM_006721297.2 XM_006721298.2 XM_006721300.2 XM_011523373.2 XM_011523375.2 XM_017023766.1 XM_017023767.1 XM_017023768.1 XM_017023769.1 XM_017023770.1 XM_017023771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6079330 c.116C>T p.T39M Chr.16: 57020828-57020828 on Build GRCh38
5224923 c.185A>G p.H62R Chr.16: 57020897-57020897 on Build GRCh38
6485466 c.220C>T p.H74Y Chr.16: 57020932-57020932 on Build GRCh38
6485465 c.220C>T p.H74Y Chr.16: 57020932-57020932 on Build GRCh38
2836146 c.136G>A p.E46K Chr.16: 57020848-57020848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767100380 Chr.16: 57020866-57020866 on Build GRCh38
rs111249602 Chr.16: 57021054-57021054 on Build GRCh38
rs184991438 Chr.16: 57021118-57021118 on Build GRCh38
rs759262585 Chr.16: 57021124-57021124 on Build GRCh38
rs114632706 Chr.16: 57021142-57021142 on Build GRCh38
rs771869422 Chr.16: 57020982-57020982 on Build GRCh38
rs368996988 Chr.16: 57020848-57020848 on Build GRCh38
rs753541934 Chr.16: 57020905-57020905 on Build GRCh38
rs774081805 Chr.16: 57021100-57021100 on Build GRCh38
rs147394286 Chr.16: 57020990-57020990 on Build GRCh38
rs1009119016 Chr.16: 57021161-57021161 on Build GRCh38
rs757409351 Chr.16: 57021029-57021029 on Build GRCh38
rs777526365 Chr.16: 57021053-57021053 on Build GRCh38
rs950851042 Chr.16: 57020983-57020983 on Build GRCh38
rs772786249 Chr.16: 57020867-57020867 on Build GRCh38
rs761583329 Chr.16: 57020850-57020850 on Build GRCh38
rs540291182 Chr.16: 57021119-57021119 on Build GRCh38
rs765035595 Chr.16: 57020909-57020909 on Build GRCh38
rs368715111 Chr.16: 57020997-57020997 on Build GRCh38
rs112858120 Chr.16: 57021130-57021130 on Build GRCh38
rs763147979 Chr.16: 57021017-57021017 on Build GRCh38
rs375166779 Chr.16: 57021040-57021040 on Build GRCh38
rs777656667 Chr.16: 57020911-57020911 on Build GRCh38
rs868761728 Chr.16: 57021057-57021057 on Build GRCh38
rs746312170 Chr.16: 57020847-57020847 on Build GRCh38
rs764942760 Chr.16: 57021006-57021006 on Build GRCh38
rs545659470 Chr.16: 57021067-57021067 on Build GRCh38
rs752502662 Chr.16: 57021014-57021014 on Build GRCh38
rs748115787 Chr.16: 57020851-57020851 on Build GRCh38
rs749732904 Chr.16: 57021041-57021041 on Build GRCh38
rs577454687 Chr.16: 57021207-57021207 on Build GRCh38
rs9939318 Chr.16: 57021272-57021272 on Build GRCh38
rs955022333 Chr.16: 57020842-57020842 on Build GRCh38
rs769824909 Chr.16: 57020883-57020883 on Build GRCh38
rs752265314 Chr.16: 57021188-57021188 on Build GRCh38
rs534263309 Chr.16: 57020952-57020952 on Build GRCh38
rs555349987 Chr.16: 57020959-57020959 on Build GRCh38
rs865865861 Chr.16: 57021185-57021185 on Build GRCh38
rs750431597 Chr.16: 57021013-57021014 on Build GRCh38
rs999678985 Chr.16: 57021121-57021121 on Build GRCh38
rs771588780 Chr.16: 57021046-57021046 on Build GRCh38
rs182021192 Chr.16: 57021028-57021028 on Build GRCh38
rs564279381 Chr.16: 57021080-57021080 on Build GRCh38
rs368103963 Chr.16: 57021283-57021283 on Build GRCh38
rs766403859 Chr.16: 57020901-57020901 on Build GRCh38
rs770236357 Chr.16: 57020968-57020968 on Build GRCh38
rs767013416 Chr.16: 57021168-57021168 on Build GRCh38
rs184877810 Chr.16: 57020906-57020906 on Build GRCh38
rs747170829 Chr.16: 57020913-57020913 on Build GRCh38
rs537423505 Chr.16: 57021066-57021066 on Build GRCh38
rs375380698 Chr.16: 57021039-57021039 on Build GRCh38
rs190154357 Chr.16: 57020910-57020910 on Build GRCh38
rs760268034 Chr.16: 57020988-57020988 on Build GRCh38
rs573651232 Chr.16: 57020979-57020979 on Build GRCh38
rs757701534 Chr.16: 57020943-57020943 on Build GRCh38
rs770750307 Chr.16: 57020989-57020989 on Build GRCh38
rs759500634 Chr.16: 57021003-57021003 on Build GRCh38

View Full Product Details