Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75567945-75568454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABARAPL2
Entrez Gene ID
11345
Gene Name
GABA type A receptor associated protein like 2
Gene Aliases
ATG8, ATG8C, GATE-16, GATE16, GEF-2, GEF2
UniGene
Hs.461379
Gene Chromosome Location
Chr.16: 75566351-75577881 on Build GRCh38
Transcripts
NM_007285.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2691445 c.116C>T p.S39F Chr.16: 75568062-75568062 on Build GRCh38
973749 c.141G>A p.R47R Chr.16: 75568087-75568087 on Build GRCh38
6283455 c.94A>C p.I32L Chr.16: 75568040-75568040 on Build GRCh38
973750 c.250G>T p.V84F Chr.16: 75568196-75568196 on Build GRCh38
3421163 c.225G>A p.A75A Chr.16: 75568171-75568171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747764185 Chr.16: 75568195-75568195 on Build GRCh38
rs530257337 Chr.16: 75568267-75568267 on Build GRCh38
rs769203122 Chr.16: 75568229-75568230 on Build GRCh38
rs775400512 Chr.16: 75568070-75568070 on Build GRCh38
rs147967714 Chr.16: 75568039-75568039 on Build GRCh38
rs902216804 Chr.16: 75568289-75568289 on Build GRCh38
rs549374958 Chr.16: 75568022-75568022 on Build GRCh38
rs990040996 Chr.16: 75568023-75568023 on Build GRCh38
rs113667686 Chr.16: 75567997-75567997 on Build GRCh38
rs928106556 Chr.16: 75568170-75568170 on Build GRCh38
rs956995613 Chr.16: 75568338-75568338 on Build GRCh38
rs781259182 Chr.16: 75567995-75567995 on Build GRCh38
rs143894870 Chr.16: 75567988-75567988 on Build GRCh38
rs767586471 Chr.16: 75568262-75568262 on Build GRCh38
rs376524351 Chr.16: 75568252-75568252 on Build GRCh38
rs771872766 Chr.16: 75568054-75568054 on Build GRCh38
rs375396410 Chr.16: 75568006-75568006 on Build GRCh38
rs746385892 Chr.16: 75568077-75568077 on Build GRCh38
rs1031358414 Chr.16: 75568322-75568322 on Build GRCh38
rs887086043 Chr.16: 75568185-75568185 on Build GRCh38
rs748306555 Chr.16: 75567996-75567996 on Build GRCh38
rs141745406 Chr.16: 75568068-75568068 on Build GRCh38
rs1023490733 Chr.16: 75567973-75567973 on Build GRCh38
rs770213702 Chr.16: 75567989-75567989 on Build GRCh38
rs990073555 Chr.16: 75568364-75568364 on Build GRCh38
rs569363885 Chr.16: 75568042-75568042 on Build GRCh38
rs757319789 Chr.16: 75567986-75567986 on Build GRCh38
rs774133364 Chr.16: 75568003-75568003 on Build GRCh38
rs761962095 Chr.16: 75568253-75568253 on Build GRCh38
rs535218020 Chr.16: 75568266-75568266 on Build GRCh38
rs764362077 Chr.16: 75568032-75568032 on Build GRCh38
rs890906531 Chr.16: 75568275-75568275 on Build GRCh38
rs570092183 Chr.16: 75568009-75568009 on Build GRCh38
rs769043149 Chr.16: 75568131-75568131 on Build GRCh38
rs773314150 Chr.16: 75568014-75568014 on Build GRCh38
rs1022848726 Chr.16: 75568375-75568375 on Build GRCh38
rs779014166 Chr.16: 75568017-75568017 on Build GRCh38
rs147088769 Chr.16: 75568085-75568085 on Build GRCh38
rs766296843 Chr.16: 75568217-75568217 on Build GRCh38
rs200759741 Chr.16: 75568018-75568018 on Build GRCh38
rs146566773 Chr.16: 75568272-75568272 on Build GRCh38
rs754801051 Chr.16: 75568028-75568028 on Build GRCh38
rs35804034 Chr.16: 75568034-75568035 on Build GRCh38
rs941629800 Chr.16: 75568194-75568194 on Build GRCh38
rs151052580 Chr.16: 75568109-75568109 on Build GRCh38
rs572633192 Chr.16: 75568305-75568305 on Build GRCh38
rs759302311 Chr.16: 75568233-75568233 on Build GRCh38
rs17852684 Chr.16: 75568099-75568099 on Build GRCh38
rs11556291 Chr.16: 75568098-75568098 on Build GRCh38
rs558399222 Chr.16: 75568343-75568343 on Build GRCh38
rs11556292 Chr.16: 75568111-75568111 on Build GRCh38
rs11556297 Chr.16: 75568157-75568157 on Build GRCh38
rs776620797 Chr.16: 75568229-75568229 on Build GRCh38
rs934952364 Chr.16: 75568311-75568311 on Build GRCh38
rs772792179 Chr.16: 75568215-75568215 on Build GRCh38
rs764967250 Chr.16: 75568235-75568235 on Build GRCh38
rs1014848083 Chr.16: 75568354-75568354 on Build GRCh38
rs373239692 Chr.16: 75568180-75568180 on Build GRCh38
rs578254640 Chr.16: 75568349-75568349 on Build GRCh38
rs777501069 Chr.16: 75568361-75568361 on Build GRCh38
rs970137038 Chr.16: 75567977-75567977 on Build GRCh38
rs768419811 Chr.16: 75568304-75568304 on Build GRCh38
rs565593150 Chr.16: 75568025-75568025 on Build GRCh38
rs746764553 Chr.16: 75568174-75568174 on Build GRCh38
rs749670434 Chr.16: 75568110-75568110 on Build GRCh38
rs546777842 Chr.16: 75568319-75568319 on Build GRCh38
rs771730679 Chr.16: 75568209-75568209 on Build GRCh38
rs1043493323 Chr.16: 75568276-75568276 on Build GRCh38
rs766211680 Chr.16: 75568019-75568019 on Build GRCh38
rs1014386416 Chr.16: 75567970-75567970 on Build GRCh38
rs375289296 Chr.16: 75568004-75568004 on Build GRCh38
rs895997854 Chr.16: 75567966-75567966 on Build GRCh38
rs1031328987 Chr.16: 75568016-75568016 on Build GRCh38

View Full Product Details