Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 78424735-78425229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWOX
Entrez Gene ID
51741
Gene Name
WW domain containing oxidoreductase
Gene Aliases
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1
UniGene
Hs.461453
Gene Chromosome Location
Chr.16: 78099413-79212667 on Build GRCh38
Transcripts
NM_001291997.1 NM_016373.3 XM_011523101.2 XM_011523103.2 XM_011523104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2692609 c.731A>G p.Q244R Chr.16: 78424995-78424995 on Build GRCh38
5573535 c.319+1G>A p.? Chr.16: 78425056-78425056 on Build GRCh38
3691190 c.273C>T p.A91A Chr.16: 78425009-78425009 on Build GRCh38
6823588 c.167C>T p.T56I Chr.16: 78424902-78424902 on Build GRCh38
1159061 c.291G>A p.V97V Chr.16: 78425027-78425027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774003937 Chr.16: 78425090-78425090 on Build GRCh38
rs540446906 Chr.16: 78424763-78424763 on Build GRCh38
rs925097988 Chr.16: 78424767-78424767 on Build GRCh38
rs775895501 Chr.16: 78425031-78425031 on Build GRCh38
rs754238116 Chr.16: 78424856-78424857 on Build GRCh38
rs376709568 Chr.16: 78425050-78425050 on Build GRCh38
rs367599412 Chr.16: 78424920-78424920 on Build GRCh38
rs922761596 Chr.16: 78425116-78425116 on Build GRCh38
rs753602303 Chr.16: 78424902-78424902 on Build GRCh38
rs569529954 Chr.16: 78424960-78424960 on Build GRCh38
rs754689899 Chr.16: 78425070-78425070 on Build GRCh38
rs199628364 Chr.16: 78424953-78424953 on Build GRCh38
rs549596385 Chr.16: 78425170-78425170 on Build GRCh38
rs746406815 Chr.16: 78425082-78425082 on Build GRCh38
rs545932191 Chr.16: 78424823-78424823 on Build GRCh38
rs575080327 Chr.16: 78424979-78424979 on Build GRCh38
rs942917595 Chr.16: 78425089-78425089 on Build GRCh38
rs878855021 Chr.16: 78425043-78425043 on Build GRCh38
rs1007029734 Chr.16: 78424840-78424840 on Build GRCh38
rs139257288 Chr.16: 78424854-78424854 on Build GRCh38
rs932587405 Chr.16: 78425143-78425143 on Build GRCh38
rs747021033 Chr.16: 78424964-78424964 on Build GRCh38
rs767929766 Chr.16: 78424893-78424893 on Build GRCh38
rs547754277 Chr.16: 78425138-78425138 on Build GRCh38
rs939001505 Chr.16: 78425201-78425201 on Build GRCh38
rs1056085672 Chr.16: 78425208-78425208 on Build GRCh38
rs749917475 Chr.16: 78425097-78425097 on Build GRCh38
rs368928190 Chr.16: 78425013-78425013 on Build GRCh38
rs769589145 Chr.16: 78424975-78424975 on Build GRCh38
rs751007131 Chr.16: 78424793-78424793 on Build GRCh38
rs180692241 Chr.16: 78425088-78425088 on Build GRCh38
rs775807907 Chr.16: 78425087-78425087 on Build GRCh38
rs746194314 Chr.16: 78424835-78424835 on Build GRCh38
rs760577782 Chr.16: 78425103-78425103 on Build GRCh38
rs766371404 Chr.16: 78425107-78425107 on Build GRCh38
rs4130513 Chr.16: 78424853-78424853 on Build GRCh38
rs924675533 Chr.16: 78424948-78424948 on Build GRCh38
rs540386692 Chr.16: 78424985-78424985 on Build GRCh38
rs1051957011 Chr.16: 78425141-78425141 on Build GRCh38
rs547619401 Chr.16: 78424894-78424894 on Build GRCh38
rs146090457 Chr.16: 78425077-78425077 on Build GRCh38
rs370737224 Chr.16: 78424895-78424895 on Build GRCh38
rs763152209 Chr.16: 78424880-78424880 on Build GRCh38
rs896783193 Chr.16: 78425021-78425021 on Build GRCh38
rs757159030 Chr.16: 78424969-78424969 on Build GRCh38
rs779882158 Chr.16: 78424852-78424852 on Build GRCh38
rs759826716 Chr.16: 78424988-78424988 on Build GRCh38
rs866341813 Chr.16: 78424874-78424874 on Build GRCh38
rs777579075 Chr.16: 78424914-78424914 on Build GRCh38
rs372408512 Chr.16: 78424954-78424954 on Build GRCh38
rs75559202 Chr.16: 78425018-78425018 on Build GRCh38
rs982607423 Chr.16: 78425055-78425055 on Build GRCh38
rs765582297 Chr.16: 78424990-78424990 on Build GRCh38
rs762566029 Chr.16: 78425032-78425032 on Build GRCh38
rs757863128 Chr.16: 78424961-78424961 on Build GRCh38
rs377191055 Chr.16: 78424908-78424908 on Build GRCh38
rs757643012 Chr.16: 78425086-78425086 on Build GRCh38
rs773518778 Chr.16: 78425015-78425015 on Build GRCh38
rs200966505 Chr.16: 78425001-78425001 on Build GRCh38
rs768734917 Chr.16: 78424931-78424931 on Build GRCh38
rs779754174 Chr.16: 78425066-78425066 on Build GRCh38
rs767565166 Chr.16: 78424940-78424940 on Build GRCh38
rs191029309 Chr.16: 78424987-78424987 on Build GRCh38
rs755974419 Chr.16: 78424955-78424955 on Build GRCh38
rs1023123497 Chr.16: 78425073-78425073 on Build GRCh38
rs762427803 Chr.16: 78424888-78424888 on Build GRCh38
rs941156863 Chr.16: 78425012-78425012 on Build GRCh38
rs747172163 Chr.16: 78425076-78425076 on Build GRCh38
rs570658147 Chr.16: 78425144-78425144 on Build GRCh38
rs778327490 Chr.16: 78425120-78425120 on Build GRCh38
rs150811493 Chr.16: 78424801-78424801 on Build GRCh38
rs750905852 Chr.16: 78424942-78424942 on Build GRCh38
rs770774574 Chr.16: 78424922-78424922 on Build GRCh38
rs370129950 Chr.16: 78424861-78424861 on Build GRCh38
rs369715848 Chr.16: 78425024-78425024 on Build GRCh38
rs7202218 Chr.16: 78425123-78425123 on Build GRCh38
rs1023469089 Chr.16: 78425183-78425183 on Build GRCh38
rs1053373645 Chr.16: 78425158-78425158 on Build GRCh38
rs202006159 Chr.16: 78424899-78424899 on Build GRCh38
rs982171635 Chr.16: 78424930-78424930 on Build GRCh38
rs7201683 Chr.16: 78424910-78424910 on Build GRCh38
rs376279847 Chr.16: 78424882-78424882 on Build GRCh38
rs569753154 Chr.16: 78425177-78425177 on Build GRCh38
rs990864463 Chr.16: 78424770-78424770 on Build GRCh38
rs527595856 Chr.16: 78425129-78425129 on Build GRCh38
rs956038787 Chr.16: 78425115-78425115 on Build GRCh38
rs999623549 Chr.16: 78425153-78425153 on Build GRCh38
rs910158143 Chr.16: 78425086-78425086 on Build GRCh38
rs985286987 Chr.16: 78425176-78425176 on Build GRCh38
rs771068553 Chr.16: 78425022-78425022 on Build GRCh38
rs74860463 Chr.16: 78424877-78424877 on Build GRCh38
rs756074406 Chr.16: 78425060-78425060 on Build GRCh38
rs376040091 Chr.16: 78424937-78424937 on Build GRCh38
rs984097780 Chr.16: 78425083-78425083 on Build GRCh38
rs730882215 Chr.16: 78424869-78424869 on Build GRCh38
rs535446666 Chr.16: 78424963-78424963 on Build GRCh38
rs756495469 Chr.16: 78424843-78424843 on Build GRCh38
rs766952918 Chr.16: 78425059-78425059 on Build GRCh38
rs74618473 Chr.16: 78424802-78424802 on Build GRCh38
rs368401081 Chr.16: 78424944-78424944 on Build GRCh38
rs1036753245 Chr.16: 78425196-78425196 on Build GRCh38
rs1049479059 Chr.16: 78424792-78424792 on Build GRCh38
rs375934868 Chr.16: 78425011-78425011 on Build GRCh38
rs752354290 Chr.16: 78424994-78424994 on Build GRCh38
rs776409839 Chr.16: 78424923-78424923 on Build GRCh38
rs781010507 Chr.16: 78424970-78424970 on Build GRCh38
rs941197412 Chr.16: 78425194-78425194 on Build GRCh38
rs746893085 Chr.16: 78424915-78424915 on Build GRCh38
rs72549408 Chr.16: 78424933-78424933 on Build GRCh38
rs896915322 Chr.16: 78425200-78425200 on Build GRCh38
rs970976114 Chr.16: 78425186-78425186 on Build GRCh38
rs142806268 Chr.16: 78424977-78424977 on Build GRCh38
rs368070977 Chr.16: 78424981-78424981 on Build GRCh38
rs1023419687 Chr.16: 78425030-78425030 on Build GRCh38
rs756703833 Chr.16: 78425010-78425010 on Build GRCh38
rs761440135 Chr.16: 78425094-78425094 on Build GRCh38
rs185242276 Chr.16: 78424825-78424825 on Build GRCh38
rs770649734 Chr.16: 78424878-78424878 on Build GRCh38
rs756762196 Chr.16: 78425054-78425054 on Build GRCh38
rs763754438 Chr.16: 78425039-78425039 on Build GRCh38
rs753689854 Chr.16: 78425069-78425069 on Build GRCh38
rs765590833 Chr.16: 78425027-78425027 on Build GRCh38
rs766512901 Chr.16: 78424817-78424817 on Build GRCh38
rs761250135 Chr.16: 78424947-78424947 on Build GRCh38
rs745681407 Chr.16: 78424972-78424972 on Build GRCh38
rs186606524 Chr.16: 78425155-78425155 on Build GRCh38
rs745399177 Chr.16: 78425037-78425037 on Build GRCh38
rs1057517847 Chr.16: 78424982-78424982 on Build GRCh38
rs749277249 Chr.16: 78425009-78425009 on Build GRCh38
rs1032182347 Chr.16: 78425114-78425114 on Build GRCh38
rs760077691 Chr.16: 78425025-78425025 on Build GRCh38
rs996692455 Chr.16: 78424791-78424791 on Build GRCh38
rs936795819 Chr.16: 78424768-78424768 on Build GRCh38
rs978830011 Chr.16: 78425112-78425112 on Build GRCh38
rs900969040 Chr.16: 78424781-78424781 on Build GRCh38
rs1030035701 Chr.16: 78424998-78424998 on Build GRCh38

View Full Product Details