Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81869114-81869610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCG2
Entrez Gene ID
5336
Gene Name
phospholipase C gamma 2
Gene Aliases
APLAID, FCAS3, PLC-IV, PLC-gamma-2
UniGene
Hs.372303
Gene Chromosome Location
Chr.16: 81779258-81962693 on Build GRCh38
Transcripts
NM_002661.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6928451 c.487C>A p.L163I Chr.16: 81869221-81869221 on Build GRCh38
2693561 c.526A>G p.K176E Chr.16: 81869260-81869260 on Build GRCh38
974170 c.548T>C p.L183P Chr.16: 81869282-81869282 on Build GRCh38
435845 c.519C>A p.I173I Chr.16: 81869253-81869253 on Build GRCh38
974167 c.491G>A p.R164Q Chr.16: 81869225-81869225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563204700 Chr.16: 81869338-81869338 on Build GRCh38
rs763930840 Chr.16: 81869322-81869322 on Build GRCh38
rs751460917 Chr.16: 81869328-81869328 on Build GRCh38
rs571851487 Chr.16: 81869154-81869154 on Build GRCh38
rs759752251 Chr.16: 81869238-81869238 on Build GRCh38
rs534747933 Chr.16: 81869512-81869512 on Build GRCh38
rs4889419 Chr.16: 81869385-81869385 on Build GRCh38
rs766422892 Chr.16: 81869225-81869225 on Build GRCh38
rs1026003731 Chr.16: 81869414-81869414 on Build GRCh38
rs747767769 Chr.16: 81869155-81869155 on Build GRCh38
rs777295977 Chr.16: 81869220-81869220 on Build GRCh38
rs553968457 Chr.16: 81869180-81869180 on Build GRCh38
rs1007393418 Chr.16: 81869375-81869375 on Build GRCh38
rs115472157 Chr.16: 81869365-81869365 on Build GRCh38
rs1049147683 Chr.16: 81869407-81869407 on Build GRCh38
rs937478821 Chr.16: 81869566-81869566 on Build GRCh38
rs1010565795 Chr.16: 81869471-81869471 on Build GRCh38
rs200981336 Chr.16: 81869267-81869267 on Build GRCh38
rs1038926633 Chr.16: 81869380-81869380 on Build GRCh38
rs868812502 Chr.16: 81869384-81869384 on Build GRCh38
rs950494824 Chr.16: 81869424-81869424 on Build GRCh38
rs779774477 Chr.16: 81869275-81869275 on Build GRCh38
rs568110018 Chr.16: 81869393-81869393 on Build GRCh38
rs771371340 Chr.16: 81869349-81869349 on Build GRCh38
rs12445580 Chr.16: 81869192-81869192 on Build GRCh38
rs966448462 Chr.16: 81869484-81869484 on Build GRCh38
rs910069236 Chr.16: 81869323-81869323 on Build GRCh38
rs780892532 Chr.16: 81869190-81869190 on Build GRCh38
rs199650798 Chr.16: 81869335-81869335 on Build GRCh38
rs753974933 Chr.16: 81869236-81869236 on Build GRCh38
rs372619063 Chr.16: 81869314-81869314 on Build GRCh38
rs945170250 Chr.16: 81869214-81869214 on Build GRCh38
rs1033867500 Chr.16: 81869183-81869183 on Build GRCh38
rs901739877 Chr.16: 81869551-81869551 on Build GRCh38
rs772405739 Chr.16: 81869208-81869208 on Build GRCh38
rs142022471 Chr.16: 81869219-81869219 on Build GRCh38
rs758204350 Chr.16: 81869168-81869168 on Build GRCh38
rs117594647 Chr.16: 81869438-81869438 on Build GRCh38
rs41301799 Chr.16: 81869141-81869141 on Build GRCh38
rs913959412 Chr.16: 81869358-81869358 on Build GRCh38
rs759598100 Chr.16: 81869244-81869244 on Build GRCh38
rs754643031 Chr.16: 81869339-81869339 on Build GRCh38
rs544889842 Chr.16: 81869292-81869292 on Build GRCh38
rs965981959 Chr.16: 81869383-81869383 on Build GRCh38
rs925412822 Chr.16: 81869515-81869515 on Build GRCh38
rs757192083 Chr.16: 81869329-81869329 on Build GRCh38
rs138932132 Chr.16: 81869431-81869431 on Build GRCh38
rs56070867 Chr.16: 81869158-81869158 on Build GRCh38
rs950525775 Chr.16: 81869283-81869283 on Build GRCh38
rs921822245 Chr.16: 81869525-81869525 on Build GRCh38
rs564767670 Chr.16: 81869317-81869317 on Build GRCh38
rs373539123 Chr.16: 81869344-81869344 on Build GRCh38
rs961014939 Chr.16: 81869461-81869461 on Build GRCh38
rs778257517 Chr.16: 81869546-81869546 on Build GRCh38
rs150276286 Chr.16: 81869274-81869274 on Build GRCh38
rs905206926 Chr.16: 81869150-81869150 on Build GRCh38
rs200464512 Chr.16: 81869239-81869239 on Build GRCh38
rs908003585 Chr.16: 81869547-81869547 on Build GRCh38
rs1056428577 Chr.16: 81869469-81869469 on Build GRCh38
rs992548278 Chr.16: 81869470-81869470 on Build GRCh38
rs777145741 Chr.16: 81869350-81869350 on Build GRCh38
rs142152450 Chr.16: 81869451-81869451 on Build GRCh38
rs368907385 Chr.16: 81869306-81869306 on Build GRCh38
rs368147709 Chr.16: 81869199-81869199 on Build GRCh38
rs753723522 Chr.16: 81869445-81869445 on Build GRCh38
rs772451566 Chr.16: 81869281-81869281 on Build GRCh38
rs1022079896 Chr.16: 81869472-81869472 on Build GRCh38
rs371816221 Chr.16: 81869184-81869184 on Build GRCh38
rs903705959 Chr.16: 81869368-81869368 on Build GRCh38
rs974705417 Chr.16: 81869518-81869518 on Build GRCh38
rs759045625 Chr.16: 81869504-81869504 on Build GRCh38
rs571514350 Chr.16: 81869394-81869394 on Build GRCh38
rs185747646 Chr.16: 81869454-81869454 on Build GRCh38
rs559014966 Chr.16: 81869165-81869165 on Build GRCh38
rs761156413 Chr.16: 81869307-81869307 on Build GRCh38
rs570095697 Chr.16: 81869364-81869364 on Build GRCh38
rs767271565 Chr.16: 81869356-81869356 on Build GRCh38
rs542611829 Chr.16: 81869227-81869227 on Build GRCh38
rs4889420 Chr.16: 81869450-81869450 on Build GRCh38
rs375598204 Chr.16: 81869221-81869221 on Build GRCh38
rs945422569 Chr.16: 81869489-81869489 on Build GRCh38
rs758235707 Chr.16: 81869245-81869245 on Build GRCh38
rs780839015 Chr.16: 81869273-81869273 on Build GRCh38
rs1005982525 Chr.16: 81869427-81869427 on Build GRCh38
rs769635408 Chr.16: 81869196-81869196 on Build GRCh38
rs933271406 Chr.16: 81869565-81869565 on Build GRCh38
rs530733095 Chr.16: 81869357-81869357 on Build GRCh38
rs952075284 Chr.16: 81869537-81869537 on Build GRCh38
rs777536190 Chr.16: 81869250-81869250 on Build GRCh38
rs778557099 Chr.16: 81869163-81869163 on Build GRCh38
rs765328526 Chr.16: 81869316-81869316 on Build GRCh38
rs886113784 Chr.16: 81869581-81869581 on Build GRCh38
rs751515873 Chr.16: 81869255-81869255 on Build GRCh38
rs1038872886 Chr.16: 81869541-81869541 on Build GRCh38
rs146317930 Chr.16: 81869485-81869485 on Build GRCh38
rs576818181 Chr.16: 81869143-81869143 on Build GRCh38
rs1047819945 Chr.16: 81869576-81869576 on Build GRCh38
rs779469871 Chr.16: 81869337-81869337 on Build GRCh38
rs369350365 Chr.16: 81869336-81869336 on Build GRCh38
rs754452181 Chr.16: 81869449-81869449 on Build GRCh38
rs763128611 Chr.16: 81869321-81869321 on Build GRCh38
rs748418080 Chr.16: 81869276-81869276 on Build GRCh38
rs943199118 Chr.16: 81869534-81869534 on Build GRCh38
rs762248145 Chr.16: 81869205-81869205 on Build GRCh38
rs983337419 Chr.16: 81869298-81869298 on Build GRCh38
rs1046197367 Chr.16: 81869555-81869555 on Build GRCh38
rs551718964 Chr.16: 81869362-81869362 on Build GRCh38
rs747380823 Chr.16: 81869348-81869348 on Build GRCh38

View Full Product Details