Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81999626-82000130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDR42E1
Entrez Gene ID
93517
Gene Name
short chain dehydrogenase/reductase family 42E, member 1
Gene Aliases
HSPC105
UniGene
Hs.87779
Gene Chromosome Location
Chr.16: 81988855-82011488 on Build GRCh38
Transcripts
NM_145168.2 XM_005256257.3 XM_011523471.2 XM_011523472.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988693 c.646C>A p.P216T Chr.16: 81999647-81999647 on Build GRCh38
6145318 c.368C>T p.T123I Chr.16: 81999925-81999925 on Build GRCh38
3889063 c.451C>T p.H151Y Chr.16: 81999842-81999842 on Build GRCh38
5759548 c.265C>G p.Q89E Chr.16: 82000028-82000028 on Build GRCh38
3402512 c.259C>T p.R87W Chr.16: 82000034-82000034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542979442 Chr.16: 81999821-81999821 on Build GRCh38
rs749018288 Chr.16: 82000058-82000058 on Build GRCh38
rs369065439 Chr.16: 81999683-81999683 on Build GRCh38
rs531813448 Chr.16: 81999833-81999833 on Build GRCh38
rs188668118 Chr.16: 81999942-81999942 on Build GRCh38
rs200329125 Chr.16: 82000103-82000103 on Build GRCh38
rs766486778 Chr.16: 81999931-81999931 on Build GRCh38
rs757325623 Chr.16: 81999752-81999752 on Build GRCh38
rs377336713 Chr.16: 81999771-81999771 on Build GRCh38
rs757177952 Chr.16: 82000104-82000104 on Build GRCh38
rs767905284 Chr.16: 81999900-81999900 on Build GRCh38
rs16956174 Chr.16: 82000007-82000007 on Build GRCh38
rs377085444 Chr.16: 81999654-81999654 on Build GRCh38
rs767248528 Chr.16: 82000106-82000109 on Build GRCh38
rs536185013 Chr.16: 81999920-81999920 on Build GRCh38
rs755718224 Chr.16: 82000043-82000043 on Build GRCh38
rs374652788 Chr.16: 82000101-82000101 on Build GRCh38
rs372999295 Chr.16: 81999750-81999750 on Build GRCh38
rs769507054 Chr.16: 81999884-81999884 on Build GRCh38
rs760207708 Chr.16: 81999737-81999737 on Build GRCh38
rs762071018 Chr.16: 81999991-81999991 on Build GRCh38
rs751981686 Chr.16: 81999824-81999824 on Build GRCh38
rs768214139 Chr.16: 82000068-82000068 on Build GRCh38
rs748000244 Chr.16: 81999864-81999864 on Build GRCh38
rs770969365 Chr.16: 81999699-81999699 on Build GRCh38
rs577864776 Chr.16: 81999719-81999719 on Build GRCh38
rs542985854 Chr.16: 82000077-82000077 on Build GRCh38
rs761131995 Chr.16: 81999996-81999996 on Build GRCh38
rs754738263 Chr.16: 81999855-81999855 on Build GRCh38
rs914035439 Chr.16: 81999688-81999688 on Build GRCh38
rs764589474 Chr.16: 81999823-81999823 on Build GRCh38
rs147342330 Chr.16: 81999866-81999866 on Build GRCh38
rs772892529 Chr.16: 81999736-81999736 on Build GRCh38
rs202151516 Chr.16: 81999703-81999703 on Build GRCh38
rs769524016 Chr.16: 81999673-81999673 on Build GRCh38
rs767242601 Chr.16: 81999889-81999889 on Build GRCh38
rs779027858 Chr.16: 82000092-82000093 on Build GRCh38
rs777176586 Chr.16: 82000079-82000079 on Build GRCh38
rs368369528 Chr.16: 81999960-81999960 on Build GRCh38
rs201031405 Chr.16: 81999680-81999680 on Build GRCh38
rs74029334 Chr.16: 81999739-81999739 on Build GRCh38
rs370358690 Chr.16: 81999788-81999788 on Build GRCh38
rs371607120 Chr.16: 82000016-82000016 on Build GRCh38
rs200424203 Chr.16: 81999873-81999873 on Build GRCh38
rs79313103 Chr.16: 82000021-82000021 on Build GRCh38
rs765377203 Chr.16: 82000019-82000019 on Build GRCh38
rs768498391 Chr.16: 81999785-81999785 on Build GRCh38
rs975922917 Chr.16: 81999904-81999904 on Build GRCh38
rs776388358 Chr.16: 81999870-81999870 on Build GRCh38
rs747376831 Chr.16: 82000076-82000076 on Build GRCh38
rs748077114 Chr.16: 81999792-81999792 on Build GRCh38
rs781089298 Chr.16: 81999753-81999753 on Build GRCh38
rs764038782 Chr.16: 81999677-81999677 on Build GRCh38
rs777773930 Chr.16: 81999867-81999867 on Build GRCh38
rs373444132 Chr.16: 81999695-81999695 on Build GRCh38
rs377444731 Chr.16: 82000048-82000048 on Build GRCh38
rs777581352 Chr.16: 82000029-82000029 on Build GRCh38
rs758843072 Chr.16: 81999748-81999748 on Build GRCh38
rs1003705970 Chr.16: 81999798-81999798 on Build GRCh38
rs769262424 Chr.16: 81999981-81999981 on Build GRCh38
rs201095410 Chr.16: 81999727-81999727 on Build GRCh38
rs267604658 Chr.16: 81999725-81999725 on Build GRCh38
rs775458444 Chr.16: 82000096-82000096 on Build GRCh38
rs771131100 Chr.16: 81999656-81999656 on Build GRCh38
rs779541472 Chr.16: 82000054-82000054 on Build GRCh38
rs754047603 Chr.16: 81999991-81999992 on Build GRCh38
rs369956724 Chr.16: 82000064-82000064 on Build GRCh38
rs80065483 Chr.16: 82000033-82000033 on Build GRCh38
rs776781781 Chr.16: 81999659-81999659 on Build GRCh38
rs760278554 Chr.16: 81999805-81999805 on Build GRCh38
rs779462498 Chr.16: 81999687-81999687 on Build GRCh38
rs746979729 Chr.16: 81999955-81999955 on Build GRCh38
rs772536823 Chr.16: 81999994-81999994 on Build GRCh38
rs772550358 Chr.16: 81999803-81999803 on Build GRCh38
rs560725582 Chr.16: 81999768-81999768 on Build GRCh38
rs766986975 Chr.16: 81999733-81999733 on Build GRCh38
rs185391129 Chr.16: 81999857-81999857 on Build GRCh38
rs759358610 Chr.16: 81999663-81999663 on Build GRCh38
rs1013863358 Chr.16: 81999793-81999793 on Build GRCh38
rs958029792 Chr.16: 81999847-81999847 on Build GRCh38
rs749604812 Chr.16: 81999940-81999940 on Build GRCh38
rs767194959 Chr.16: 81999682-81999682 on Build GRCh38
rs757320810 Chr.16: 82000091-82000092 on Build GRCh38
rs908843293 Chr.16: 82000002-82000002 on Build GRCh38
rs749562730 Chr.16: 81999852-81999852 on Build GRCh38
rs772250009 Chr.16: 81999651-81999651 on Build GRCh38
rs141751383 Chr.16: 81999655-81999655 on Build GRCh38
rs866965730 Chr.16: 81999886-81999886 on Build GRCh38
rs112849482 Chr.16: 81999832-81999832 on Build GRCh38
rs375953968 Chr.16: 81999676-81999676 on Build GRCh38
rs761233284 Chr.16: 81999681-81999681 on Build GRCh38
rs1045212542 Chr.16: 81999770-81999770 on Build GRCh38
rs763198395 Chr.16: 82000097-82000097 on Build GRCh38
rs199769661 Chr.16: 81999650-81999650 on Build GRCh38
rs758928469 Chr.16: 81999693-81999693 on Build GRCh38
rs547630443 Chr.16: 81999868-81999868 on Build GRCh38
rs753145217 Chr.16: 81999690-81999690 on Build GRCh38
rs759669365 Chr.16: 82000018-82000018 on Build GRCh38
rs766155138 Chr.16: 81999738-81999738 on Build GRCh38
rs780258256 Chr.16: 81999772-81999772 on Build GRCh38
rs745382816 Chr.16: 81999978-81999978 on Build GRCh38
rs749077498 Chr.16: 81999784-81999784 on Build GRCh38
rs572676726 Chr.16: 82000034-82000034 on Build GRCh38
rs373873138 Chr.16: 81999820-81999820 on Build GRCh38
rs750996831 Chr.16: 81999919-81999919 on Build GRCh38
rs372075795 Chr.16: 81999969-81999969 on Build GRCh38
rs981202520 Chr.16: 81999653-81999653 on Build GRCh38
rs200345945 Chr.16: 82000089-82000089 on Build GRCh38
rs775215624 Chr.16: 81999717-81999717 on Build GRCh38
rs751419793 Chr.16: 82000031-82000031 on Build GRCh38
rs775371719 Chr.16: 81999674-81999674 on Build GRCh38
rs375434378 Chr.16: 81999988-81999988 on Build GRCh38
rs766066993 Chr.16: 81999806-81999806 on Build GRCh38
rs761373789 Chr.16: 81999732-81999732 on Build GRCh38
rs201686460 Chr.16: 81999895-81999895 on Build GRCh38
rs193269551 Chr.16: 81999952-81999952 on Build GRCh38
rs928988061 Chr.16: 81999746-81999746 on Build GRCh38
rs1009997370 Chr.16: 81999831-81999831 on Build GRCh38
rs139499034 Chr.16: 82000008-82000008 on Build GRCh38
rs201424944 Chr.16: 81999995-81999995 on Build GRCh38
rs201248467 Chr.16: 81999749-81999749 on Build GRCh38
rs746113267 Chr.16: 81999705-81999705 on Build GRCh38
rs144311554 Chr.16: 81999849-81999849 on Build GRCh38
rs554536374 Chr.16: 81999938-81999938 on Build GRCh38
rs374909800 Chr.16: 81999838-81999838 on Build GRCh38
rs374104197 Chr.16: 81999907-81999907 on Build GRCh38
rs772079715 Chr.16: 81999795-81999795 on Build GRCh38
rs755349804 Chr.16: 81999684-81999684 on Build GRCh38
rs199633870 Chr.16: 82000095-82000095 on Build GRCh38
rs919169416 Chr.16: 81999741-81999741 on Build GRCh38

View Full Product Details