Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87831431-87831932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A5
Entrez Gene ID
8140
Gene Name
solute carrier family 7 member 5
Gene Aliases
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1
UniGene
Hs.513797
Gene Chromosome Location
Chr.16: 87830022-87869499 on Build GRCh38
Transcripts
NM_003486.5 XM_006721286.3

SNP Detail

SNP ID Chromosome Location
rs113538348 Chr.16: 87831507-87831507 on Build GRCh38
rs563829290 Chr.16: 87831874-87831874 on Build GRCh38
rs113089978 Chr.16: 87831491-87831491 on Build GRCh38
rs368908903 Chr.16: 87831457-87831457 on Build GRCh38
rs961142925 Chr.16: 87831909-87831909 on Build GRCh38
rs141994153 Chr.16: 87831618-87831618 on Build GRCh38
rs528909236 Chr.16: 87831884-87831884 on Build GRCh38
rs911611773 Chr.16: 87831836-87831836 on Build GRCh38
rs1060257 Chr.16: 87831688-87831688 on Build GRCh38
rs1033657476 Chr.16: 87831731-87831731 on Build GRCh38
rs976578842 Chr.16: 87831747-87831747 on Build GRCh38
rs796357152 Chr.16: 87831482-87831482 on Build GRCh38
rs553239962 Chr.16: 87831663-87831663 on Build GRCh38
rs1015195744 Chr.16: 87831907-87831907 on Build GRCh38
rs563932925 Chr.16: 87831795-87831795 on Build GRCh38
rs932597971 Chr.16: 87831768-87831768 on Build GRCh38
rs1038718622 Chr.16: 87831621-87831621 on Build GRCh38
rs531029077 Chr.16: 87831797-87831797 on Build GRCh38
rs553047967 Chr.16: 87831643-87831643 on Build GRCh38
rs796558699 Chr.16: 87831518-87831519 on Build GRCh38
rs1057068357 Chr.16: 87831859-87831859 on Build GRCh38
rs920887078 Chr.16: 87831789-87831789 on Build GRCh38
rs964686234 Chr.16: 87831469-87831469 on Build GRCh38
rs201848498 Chr.16: 87831669-87831669 on Build GRCh38
rs542776237 Chr.16: 87831824-87831824 on Build GRCh38
rs5818648 Chr.16: 87831663-87831664 on Build GRCh38
rs376915929 Chr.16: 87831584-87831584 on Build GRCh38
rs970861279 Chr.16: 87831713-87831713 on Build GRCh38
rs869243139 Chr.16: 87831668-87831669 on Build GRCh38
rs1032936774 Chr.16: 87831652-87831652 on Build GRCh38
rs569850454 Chr.16: 87831564-87831564 on Build GRCh38
rs565967507 Chr.16: 87831492-87831492 on Build GRCh38
rs752758436 Chr.16: 87831785-87831785 on Build GRCh38
rs973588578 Chr.16: 87831759-87831759 on Build GRCh38
rs557974979 Chr.16: 87831525-87831525 on Build GRCh38
rs397811145 Chr.16: 87831667-87831668 on Build GRCh38
rs548369517 Chr.16: 87831453-87831453 on Build GRCh38
rs545358826 Chr.16: 87831777-87831777 on Build GRCh38
rs952270031 Chr.16: 87831889-87831890 on Build GRCh38
rs55936694 Chr.16: 87831595-87831595 on Build GRCh38
rs1023091465 Chr.16: 87831682-87831682 on Build GRCh38
rs938242015 Chr.16: 87831565-87831565 on Build GRCh38
rs920731137 Chr.16: 87831574-87831574 on Build GRCh38
rs539347773 Chr.16: 87831498-87831498 on Build GRCh38
rs386385353 Chr.16: 87831665-87831666 on Build GRCh38
rs895313817 Chr.16: 87831871-87831871 on Build GRCh38
rs386385352 Chr.16: 87831664-87831665 on Build GRCh38
rs528874509 Chr.16: 87831468-87831468 on Build GRCh38
rs1017928698 Chr.16: 87831462-87831462 on Build GRCh38
rs767648007 Chr.16: 87831748-87831748 on Build GRCh38
rs959374483 Chr.16: 87831735-87831735 on Build GRCh38
rs973639294 Chr.16: 87831567-87831567 on Build GRCh38
rs34684838 Chr.16: 87831516-87831517 on Build GRCh38
rs922450681 Chr.16: 87831753-87831753 on Build GRCh38
rs915105408 Chr.16: 87831801-87831801 on Build GRCh38
rs1003614080 Chr.16: 87831849-87831849 on Build GRCh38
rs146380761 Chr.16: 87831619-87831619 on Build GRCh38
rs893801484 Chr.16: 87831594-87831594 on Build GRCh38
rs1032820616 Chr.16: 87831485-87831485 on Build GRCh38
rs186229967 Chr.16: 87831847-87831847 on Build GRCh38
rs373243616 Chr.16: 87831638-87831638 on Build GRCh38
rs150027728 Chr.16: 87831902-87831902 on Build GRCh38
rs1039205997 Chr.16: 87831858-87831858 on Build GRCh38
rs1010112124 Chr.16: 87831683-87831683 on Build GRCh38
rs982623876 Chr.16: 87831719-87831719 on Build GRCh38
rs959918920 Chr.16: 87831664-87831664 on Build GRCh38
rs567627965 Chr.16: 87831470-87831470 on Build GRCh38
rs374667114 Chr.16: 87831666-87831667 on Build GRCh38
rs971132920 Chr.16: 87831508-87831508 on Build GRCh38

View Full Product Details