Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 2689618-2690113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUH
Entrez Gene ID
23277
Gene Name
clustered mitochondria homolog
Gene Aliases
CLU1, KIAA0664
UniGene
Hs.22616
Gene Chromosome Location
Chr.17: 2689386-2711633 on Build GRCh38
Transcripts
NM_015229.3 XM_011523768.2

SNP Detail

SNP ID Chromosome Location
rs918467075 Chr.17: 2689703-2689703 on Build GRCh38
rs892731504 Chr.17: 2689955-2689955 on Build GRCh38
rs570337445 Chr.17: 2689661-2689661 on Build GRCh38
rs77218430 Chr.17: 2689807-2689807 on Build GRCh38
rs1011091678 Chr.17: 2689792-2689792 on Build GRCh38
rs938620937 Chr.17: 2689660-2689660 on Build GRCh38
rs1052506599 Chr.17: 2689933-2689933 on Build GRCh38
rs765109005 Chr.17: 2689913-2689913 on Build GRCh38
rs373387658 Chr.17: 2690063-2690063 on Build GRCh38
rs1013035182 Chr.17: 2689684-2689684 on Build GRCh38
rs541732418 Chr.17: 2689804-2689804 on Build GRCh38
rs913473798 Chr.17: 2689956-2689956 on Build GRCh38
rs975771062 Chr.17: 2690009-2690009 on Build GRCh38
rs549860611 Chr.17: 2689646-2689646 on Build GRCh38
rs998426163 Chr.17: 2689754-2689754 on Build GRCh38
rs918530640 Chr.17: 2689866-2689866 on Build GRCh38
rs1002338979 Chr.17: 2689996-2689996 on Build GRCh38
rs1031257756 Chr.17: 2689779-2689779 on Build GRCh38
rs924313641 Chr.17: 2690027-2690027 on Build GRCh38
rs929829415 Chr.17: 2689707-2689707 on Build GRCh38
rs1057040929 Chr.17: 2689666-2689666 on Build GRCh38
rs961628489 Chr.17: 2689746-2689746 on Build GRCh38
rs909702839 Chr.17: 2689880-2689880 on Build GRCh38
rs897404344 Chr.17: 2689937-2689937 on Build GRCh38
rs564609366 Chr.17: 2689988-2689988 on Build GRCh38
rs376735587 Chr.17: 2689879-2689879 on Build GRCh38
rs927313770 Chr.17: 2690038-2690038 on Build GRCh38
rs924491489 Chr.17: 2689822-2689822 on Build GRCh38
rs934615420 Chr.17: 2689827-2689827 on Build GRCh38
rs1044458517 Chr.17: 2689882-2689882 on Build GRCh38
rs1006584014 Chr.17: 2689722-2689722 on Build GRCh38
rs757528394 Chr.17: 2690000-2690000 on Build GRCh38
rs1054208480 Chr.17: 2689862-2689862 on Build GRCh38
rs960758637 Chr.17: 2690012-2690012 on Build GRCh38
rs751767227 Chr.17: 2689763-2689763 on Build GRCh38
rs981440106 Chr.17: 2690032-2690032 on Build GRCh38
rs1039365701 Chr.17: 2689723-2689723 on Build GRCh38
rs1017320700 Chr.17: 2689999-2690001 on Build GRCh38
rs755294429 Chr.17: 2689719-2689719 on Build GRCh38
rs929855196 Chr.17: 2689872-2689872 on Build GRCh38
rs1035106643 Chr.17: 2689999-2689999 on Build GRCh38
rs1037076805 Chr.17: 2689934-2689934 on Build GRCh38
rs539426185 Chr.17: 2689798-2689798 on Build GRCh38
rs960706174 Chr.17: 2689816-2689816 on Build GRCh38
rs575408590 Chr.17: 2690033-2690033 on Build GRCh38
rs914504112 Chr.17: 2690040-2690040 on Build GRCh38
rs1044776418 Chr.17: 2689706-2689706 on Build GRCh38
rs150583066 Chr.17: 2689681-2689681 on Build GRCh38
rs561543039 Chr.17: 2689763-2689770 on Build GRCh38
rs868372330 Chr.17: 2689938-2689938 on Build GRCh38
rs972081553 Chr.17: 2690020-2690020 on Build GRCh38
rs965710932 Chr.17: 2689805-2689805 on Build GRCh38
rs371049211 Chr.17: 2690087-2690087 on Build GRCh38
rs781658524 Chr.17: 2689724-2689724 on Build GRCh38
rs74821358 Chr.17: 2689796-2689796 on Build GRCh38
rs893216949 Chr.17: 2689662-2689662 on Build GRCh38
rs370203235 Chr.17: 2689908-2689908 on Build GRCh38
rs937333937 Chr.17: 2690042-2690042 on Build GRCh38
rs951355429 Chr.17: 2690047-2690047 on Build GRCh38
rs972435680 Chr.17: 2689850-2689850 on Build GRCh38
rs1044623791 Chr.17: 2690064-2690064 on Build GRCh38
rs192615788 Chr.17: 2690080-2690080 on Build GRCh38
rs907179266 Chr.17: 2689708-2689708 on Build GRCh38
rs368746842 Chr.17: 2689964-2689964 on Build GRCh38
rs535717804 Chr.17: 2689668-2689668 on Build GRCh38
rs138691123 Chr.17: 2689795-2689795 on Build GRCh38
rs942490486 Chr.17: 2689894-2689894 on Build GRCh38
rs995671750 Chr.17: 2689758-2689758 on Build GRCh38
rs927936907 Chr.17: 2689648-2689648 on Build GRCh38
rs1048254843 Chr.17: 2689874-2689874 on Build GRCh38
rs910310588 Chr.17: 2689803-2689803 on Build GRCh38
rs1022484568 Chr.17: 2689973-2689973 on Build GRCh38
rs533230279 Chr.17: 2690029-2690029 on Build GRCh38
rs1048682893 Chr.17: 2689714-2689714 on Build GRCh38
rs1054219000 Chr.17: 2689647-2689647 on Build GRCh38
rs555849352 Chr.17: 2689806-2689806 on Build GRCh38
rs905352780 Chr.17: 2689980-2689980 on Build GRCh38
rs1039419360 Chr.17: 2689907-2689907 on Build GRCh38
rs572309306 Chr.17: 2689928-2689928 on Build GRCh38
rs984439383 Chr.17: 2690048-2690048 on Build GRCh38
rs534874099 Chr.17: 2689782-2689782 on Build GRCh38
rs1026722160 Chr.17: 2690034-2690034 on Build GRCh38
rs565963136 Chr.17: 2689720-2689720 on Build GRCh38

View Full Product Details