Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 20155853-20156366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPECC1
Entrez Gene ID
92521
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases
CYTSB, HCMOGT-1, HCMOGT1, NSP
UniGene
Hs.431045
Gene Chromosome Location
Chr.17: 20009301-20319026 on Build GRCh38
Transcripts
NM_001033553.2 NM_001033554.2 NM_001033555.2 NM_001243438.1 NM_001243439.1 NM_152904.4 XM_005256860.2 XM_011524075.1 XM_017025317.1 XM_017025318.1 XM_017025319.1 XM_017025320.1 XM_017025321.1 XM_017025322.1 XM_017025323.1 XM_017025324.1 XM_017025325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6788512 c.9C>A p.N3K Chr.17: 20156205-20156205 on Build GRCh38
6788500 c.5G>A p.G2D Chr.17: 20156201-20156201 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762007554 Chr.17: 20156225-20156225 on Build GRCh38
rs941602642 Chr.17: 20156077-20156077 on Build GRCh38
rs200072578 Chr.17: 20156283-20156284 on Build GRCh38
rs1005770736 Chr.17: 20155947-20155947 on Build GRCh38
rs551873946 Chr.17: 20155886-20155886 on Build GRCh38
rs961698950 Chr.17: 20155987-20155987 on Build GRCh38
rs931025092 Chr.17: 20156308-20156308 on Build GRCh38
rs1015654452 Chr.17: 20155889-20155889 on Build GRCh38
rs752646896 Chr.17: 20156277-20156278 on Build GRCh38
rs898270160 Chr.17: 20156227-20156227 on Build GRCh38
rs1015580687 Chr.17: 20155957-20155957 on Build GRCh38
rs971698458 Chr.17: 20156324-20156324 on Build GRCh38
rs568249533 Chr.17: 20156278-20156279 on Build GRCh38
rs769539232 Chr.17: 20156209-20156209 on Build GRCh38
rs1055197700 Chr.17: 20156065-20156065 on Build GRCh38
rs1036835200 Chr.17: 20156294-20156294 on Build GRCh38
rs62066869 Chr.17: 20156029-20156029 on Build GRCh38
rs3833861 Chr.17: 20156279-20156280 on Build GRCh38
rs373920002 Chr.17: 20156043-20156043 on Build GRCh38
rs576583123 Chr.17: 20156278-20156278 on Build GRCh38
rs565188289 Chr.17: 20155940-20155940 on Build GRCh38
rs896763594 Chr.17: 20156338-20156338 on Build GRCh38
rs759300181 Chr.17: 20156274-20156274 on Build GRCh38
rs981207763 Chr.17: 20156038-20156038 on Build GRCh38
rs1032861713 Chr.17: 20156021-20156021 on Build GRCh38
rs1020119294 Chr.17: 20156303-20156303 on Build GRCh38
rs957247126 Chr.17: 20156022-20156022 on Build GRCh38
rs767504341 Chr.17: 20156233-20156233 on Build GRCh38
rs897359410 Chr.17: 20156245-20156245 on Build GRCh38
rs777123957 Chr.17: 20156150-20156150 on Build GRCh38
rs1044033268 Chr.17: 20156028-20156028 on Build GRCh38
rs898340104 Chr.17: 20156296-20156296 on Build GRCh38
rs1015353128 Chr.17: 20156064-20156064 on Build GRCh38
rs989165563 Chr.17: 20156027-20156027 on Build GRCh38
rs118085302 Chr.17: 20156270-20156270 on Build GRCh38
rs937096226 Chr.17: 20156059-20156059 on Build GRCh38
rs985249751 Chr.17: 20156018-20156018 on Build GRCh38
rs1052532986 Chr.17: 20156012-20156012 on Build GRCh38
rs892968277 Chr.17: 20156286-20156286 on Build GRCh38
rs1035267950 Chr.17: 20156282-20156283 on Build GRCh38
rs527784255 Chr.17: 20156334-20156334 on Build GRCh38
rs774293627 Chr.17: 20156221-20156221 on Build GRCh38
rs1055229426 Chr.17: 20155919-20155919 on Build GRCh38
rs1050889797 Chr.17: 20156041-20156041 on Build GRCh38
rs866722645 Chr.17: 20156228-20156228 on Build GRCh38
rs1004331574 Chr.17: 20156062-20156062 on Build GRCh38
rs1002361006 Chr.17: 20155912-20155912 on Build GRCh38
rs1020655391 Chr.17: 20156146-20156146 on Build GRCh38
rs778611123 Chr.17: 20155899-20155899 on Build GRCh38
rs56000460 Chr.17: 20156285-20156286 on Build GRCh38
rs955139690 Chr.17: 20155879-20155879 on Build GRCh38
rs889849529 Chr.17: 20156090-20156090 on Build GRCh38
rs547804228 Chr.17: 20156008-20156008 on Build GRCh38
rs905527050 Chr.17: 20156332-20156332 on Build GRCh38
rs990146878 Chr.17: 20156257-20156257 on Build GRCh38
rs745525829 Chr.17: 20156174-20156174 on Build GRCh38
rs868567435 Chr.17: 20156199-20156199 on Build GRCh38
rs568894066 Chr.17: 20156134-20156135 on Build GRCh38
rs766800848 Chr.17: 20156266-20156266 on Build GRCh38
rs988744728 Chr.17: 20156143-20156143 on Build GRCh38
rs1002375646 Chr.17: 20156280-20156280 on Build GRCh38
rs937078304 Chr.17: 20156180-20156180 on Build GRCh38
rs545231314 Chr.17: 20156284-20156284 on Build GRCh38
rs554124645 Chr.17: 20156244-20156244 on Build GRCh38
rs761004829 Chr.17: 20156264-20156264 on Build GRCh38
rs888660095 Chr.17: 20155930-20155930 on Build GRCh38
rs558625330 Chr.17: 20156080-20156080 on Build GRCh38
rs867163729 Chr.17: 20156119-20156119 on Build GRCh38
rs906699222 Chr.17: 20155907-20155907 on Build GRCh38
rs574030351 Chr.17: 20156246-20156246 on Build GRCh38
rs749205547 Chr.17: 20156216-20156216 on Build GRCh38
rs890921891 Chr.17: 20156207-20156207 on Build GRCh38
rs556553696 Chr.17: 20156271-20156271 on Build GRCh38
rs567593630 Chr.17: 20156010-20156010 on Build GRCh38
rs910050511 Chr.17: 20156215-20156215 on Build GRCh38
rs969583286 Chr.17: 20156068-20156068 on Build GRCh38
rs538405581 Chr.17: 20156070-20156070 on Build GRCh38
rs767315538 Chr.17: 20156273-20156274 on Build GRCh38
rs957217837 Chr.17: 20156137-20156137 on Build GRCh38
rs866609762 Chr.17: 20156328-20156328 on Build GRCh38
rs527697019 Chr.17: 20155975-20155975 on Build GRCh38
rs780767220 Chr.17: 20156162-20156162 on Build GRCh38
rs1010450686 Chr.17: 20156299-20156299 on Build GRCh38
rs558898635 Chr.17: 20156284-20156285 on Build GRCh38
rs1056624920 Chr.17: 20156335-20156335 on Build GRCh38
rs534360862 Chr.17: 20156103-20156103 on Build GRCh38
rs988624973 Chr.17: 20156118-20156118 on Build GRCh38
rs980931884 Chr.17: 20155972-20155972 on Build GRCh38
rs746567616 Chr.17: 20156154-20156154 on Build GRCh38
rs934277387 Chr.17: 20156254-20156254 on Build GRCh38
rs534740086 Chr.17: 20156031-20156031 on Build GRCh38
rs565426048 Chr.17: 20156083-20156083 on Build GRCh38
rs753159799 Chr.17: 20156283-20156283 on Build GRCh38
rs1029664716 Chr.17: 20156089-20156089 on Build GRCh38
rs998277800 Chr.17: 20155991-20155991 on Build GRCh38
rs369357234 Chr.17: 20155904-20155904 on Build GRCh38
rs1027847689 Chr.17: 20156066-20156066 on Build GRCh38
rs756853865 Chr.17: 20156158-20156158 on Build GRCh38
rs1029400909 Chr.17: 20155877-20155877 on Build GRCh38
rs775289926 Chr.17: 20156210-20156210 on Build GRCh38
rs1032833992 Chr.17: 20156121-20156121 on Build GRCh38
rs998558288 Chr.17: 20156086-20156086 on Build GRCh38
rs1002806060 Chr.17: 20156330-20156330 on Build GRCh38
rs768681628 Chr.17: 20156220-20156220 on Build GRCh38
rs182777885 Chr.17: 20156037-20156037 on Build GRCh38
rs981475893 Chr.17: 20156161-20156161 on Build GRCh38

View Full Product Details