Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 22523760-22524307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTRNR2L1
Entrez Gene ID
100462977
Gene Name
MT-RNR2-like 1
Gene Aliases
HN1
UniGene
Hs.740185
Gene Chromosome Location
Chr.17: 22523111-22524665 on Build GRCh38
Transcripts
NM_001190452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243409 c.35C>T p.S12L Chr.17: 22524096-22524096 on Build GRCh38
6556240 c.51C>T p.D17D Chr.17: 22524112-22524112 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558008894 Chr.17: 22523999-22523999 on Build GRCh38
rs942591590 Chr.17: 22524130-22524130 on Build GRCh38
rs866571427 Chr.17: 22524159-22524159 on Build GRCh38
rs538071799 Chr.17: 22524224-22524224 on Build GRCh38
rs370276110 Chr.17: 22524233-22524233 on Build GRCh38
rs531851338 Chr.17: 22523840-22523840 on Build GRCh38
rs990865409 Chr.17: 22523842-22523842 on Build GRCh38
rs550428945 Chr.17: 22523898-22523898 on Build GRCh38
rs915529963 Chr.17: 22524112-22524112 on Build GRCh38
rs539930091 Chr.17: 22523995-22523995 on Build GRCh38
rs1020496524 Chr.17: 22523969-22523969 on Build GRCh38
rs75326150 Chr.17: 22524160-22524160 on Build GRCh38
rs1056307052 Chr.17: 22523953-22523953 on Build GRCh38
rs967536004 Chr.17: 22523973-22523973 on Build GRCh38
rs960675428 Chr.17: 22524269-22524269 on Build GRCh38
rs746097827 Chr.17: 22524110-22524110 on Build GRCh38
rs796553954 Chr.17: 22524135-22524137 on Build GRCh38
rs77383618 Chr.17: 22524137-22524137 on Build GRCh38
rs773794263 Chr.17: 22523919-22523919 on Build GRCh38
rs191739489 Chr.17: 22524003-22524003 on Build GRCh38
rs566152723 Chr.17: 22523994-22523994 on Build GRCh38
rs1036252559 Chr.17: 22524225-22524225 on Build GRCh38
rs1013578102 Chr.17: 22524230-22524230 on Build GRCh38
rs115274490 Chr.17: 22523909-22523909 on Build GRCh38
rs76080437 Chr.17: 22524017-22524017 on Build GRCh38
rs547787289 Chr.17: 22523908-22523908 on Build GRCh38
rs76850145 Chr.17: 22524058-22524058 on Build GRCh38
rs17052231 Chr.17: 22524001-22524001 on Build GRCh38
rs1011653192 Chr.17: 22523966-22523966 on Build GRCh38
rs377409306 Chr.17: 22524260-22524260 on Build GRCh38
rs200333123 Chr.17: 22524277-22524278 on Build GRCh38
rs373391928 Chr.17: 22524197-22524197 on Build GRCh38
rs76639367 Chr.17: 22524096-22524096 on Build GRCh38
rs200832978 Chr.17: 22524135-22524135 on Build GRCh38
rs891002178 Chr.17: 22523920-22523920 on Build GRCh38
rs893132541 Chr.17: 22524155-22524155 on Build GRCh38
rs78540355 Chr.17: 22523790-22523790 on Build GRCh38
rs1026427428 Chr.17: 22523805-22523805 on Build GRCh38
rs552234490 Chr.17: 22524201-22524201 on Build GRCh38
rs967686853 Chr.17: 22524118-22524118 on Build GRCh38
rs537670913 Chr.17: 22524002-22524002 on Build GRCh38
rs994881236 Chr.17: 22523976-22523976 on Build GRCh38
rs763326287 Chr.17: 22524175-22524175 on Build GRCh38
rs74404314 Chr.17: 22523952-22523952 on Build GRCh38
rs912096939 Chr.17: 22523846-22523846 on Build GRCh38
rs995213191 Chr.17: 22524179-22524179 on Build GRCh38
rs770922305 Chr.17: 22524075-22524075 on Build GRCh38
rs564074737 Chr.17: 22524081-22524081 on Build GRCh38
rs369868628 Chr.17: 22524207-22524207 on Build GRCh38
rs574344912 Chr.17: 22524238-22524238 on Build GRCh38
rs928842405 Chr.17: 22523833-22523833 on Build GRCh38
rs374883733 Chr.17: 22524279-22524279 on Build GRCh38
rs79405849 Chr.17: 22523789-22523789 on Build GRCh38
rs764140064 Chr.17: 22524222-22524222 on Build GRCh38
rs895034191 Chr.17: 22523958-22523958 on Build GRCh38
rs1012188129 Chr.17: 22523959-22523959 on Build GRCh38
rs78850120 Chr.17: 22524176-22524176 on Build GRCh38
rs370428096 Chr.17: 22524278-22524278 on Build GRCh38
rs371463546 Chr.17: 22524188-22524188 on Build GRCh38
rs183519861 Chr.17: 22524048-22524048 on Build GRCh38
rs1016883310 Chr.17: 22523972-22523972 on Build GRCh38
rs893246911 Chr.17: 22523946-22523946 on Build GRCh38
rs1026777940 Chr.17: 22523980-22523980 on Build GRCh38
rs369109548 Chr.17: 22524195-22524195 on Build GRCh38
rs912498677 Chr.17: 22524128-22524128 on Build GRCh38
rs374673395 Chr.17: 22524258-22524258 on Build GRCh38
rs201540164 Chr.17: 22523872-22523872 on Build GRCh38
rs914891910 Chr.17: 22523899-22523899 on Build GRCh38
rs959190706 Chr.17: 22524009-22524009 on Build GRCh38
rs950838609 Chr.17: 22523997-22523997 on Build GRCh38
rs773306031 Chr.17: 22524016-22524016 on Build GRCh38
rs939126676 Chr.17: 22524129-22524129 on Build GRCh38
rs373383042 Chr.17: 22524220-22524220 on Build GRCh38
rs941097000 Chr.17: 22523883-22523883 on Build GRCh38
rs918297870 Chr.17: 22524084-22524084 on Build GRCh38
rs566068826 Chr.17: 22524147-22524147 on Build GRCh38
rs368915468 Chr.17: 22524187-22524187 on Build GRCh38
rs962358058 Chr.17: 22524255-22524255 on Build GRCh38
rs767288212 Chr.17: 22524272-22524272 on Build GRCh38
rs531914703 Chr.17: 22524119-22524119 on Build GRCh38
rs377757852 Chr.17: 22524227-22524227 on Build GRCh38
rs80040932 Chr.17: 22524131-22524131 on Build GRCh38
rs1042525859 Chr.17: 22523913-22523913 on Build GRCh38
rs112345695 Chr.17: 22523785-22523785 on Build GRCh38
rs148742901 Chr.17: 22524014-22524014 on Build GRCh38
rs775663638 Chr.17: 22524174-22524174 on Build GRCh38
rs374751737 Chr.17: 22524192-22524192 on Build GRCh38
rs78617575 Chr.17: 22524054-22524054 on Build GRCh38
rs1039703036 Chr.17: 22523928-22523928 on Build GRCh38
rs368330834 Chr.17: 22524281-22524281 on Build GRCh38
rs535515665 Chr.17: 22523825-22523825 on Build GRCh38
rs1023297317 Chr.17: 22524241-22524241 on Build GRCh38
rs968334534 Chr.17: 22524254-22524254 on Build GRCh38
rs187584759 Chr.17: 22524051-22524051 on Build GRCh38
rs990527170 Chr.17: 22524013-22524013 on Build GRCh38
rs73982945 Chr.17: 22524202-22524202 on Build GRCh38
rs1006904554 Chr.17: 22523922-22523922 on Build GRCh38
rs967650216 Chr.17: 22523801-22523801 on Build GRCh38
rs750161680 Chr.17: 22524273-22524273 on Build GRCh38
rs376680415 Chr.17: 22524198-22524198 on Build GRCh38
rs973791414 Chr.17: 22524031-22524031 on Build GRCh38
rs552449656 Chr.17: 22523810-22523810 on Build GRCh38
rs770071031 Chr.17: 22524162-22524162 on Build GRCh38
rs978131157 Chr.17: 22524259-22524259 on Build GRCh38
rs550447422 Chr.17: 22523877-22523877 on Build GRCh38
rs200609542 Chr.17: 22523930-22523930 on Build GRCh38
rs77503166 Chr.17: 22524141-22524141 on Build GRCh38
rs62051411 Chr.17: 22524265-22524265 on Build GRCh38
rs199549973 Chr.17: 22523874-22523874 on Build GRCh38
rs775954090 Chr.17: 22524169-22524169 on Build GRCh38
rs916352578 Chr.17: 22523844-22523844 on Build GRCh38
rs538926233 Chr.17: 22524138-22524140 on Build GRCh38

View Full Product Details