Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44082464-44082968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC5
Entrez Gene ID
10014
Gene Name
histone deacetylase 5
Gene Aliases
HD5, NY-CO-9
UniGene
Hs.438782
Gene Chromosome Location
Chr.17: 44076753-44123646 on Build GRCh38
Transcripts
NM_001015053.1 NM_005474.4 XM_005256905.1 XM_005256906.4 XM_011524149.1 XM_011524150.1 XM_017023988.1 XM_017023989.1 XM_017023990.1 XM_017023991.1 XM_017023992.1 XM_017023993.1 XM_017023994.1 XM_017023995.1 XM_017023996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5513315 c.2500G>A p.A834T Chr.17: 44082784-44082784 on Build GRCh38
4414644 c.2546C>G p.A849G Chr.17: 44082646-44082646 on Build GRCh38
7318118 c.2526C>T p.F842F Chr.17: 44082666-44082666 on Build GRCh38
1383596 c.2594T>C p.L865P Chr.17: 44082598-44082598 on Build GRCh38
979857 c.2501C>T p.A834V Chr.17: 44082783-44082783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754129277 Chr.17: 44082697-44082697 on Build GRCh38
rs768139493 Chr.17: 44082666-44082666 on Build GRCh38
rs763408264 Chr.17: 44082505-44082505 on Build GRCh38
rs372426132 Chr.17: 44082715-44082715 on Build GRCh38
rs377430985 Chr.17: 44082596-44082596 on Build GRCh38
rs112293573 Chr.17: 44082763-44082763 on Build GRCh38
rs781493044 Chr.17: 44082552-44082552 on Build GRCh38
rs773900836 Chr.17: 44082737-44082737 on Build GRCh38
rs766485100 Chr.17: 44082695-44082695 on Build GRCh38
rs971009496 Chr.17: 44082904-44082904 on Build GRCh38
rs768540618 Chr.17: 44082593-44082593 on Build GRCh38
rs200821940 Chr.17: 44082548-44082548 on Build GRCh38
rs758211798 Chr.17: 44082788-44082788 on Build GRCh38
rs750663250 Chr.17: 44082558-44082558 on Build GRCh38
rs757385670 Chr.17: 44082542-44082542 on Build GRCh38
rs766106081 Chr.17: 44082564-44082564 on Build GRCh38
rs371154124 Chr.17: 44082617-44082617 on Build GRCh38
rs1018136273 Chr.17: 44082882-44082882 on Build GRCh38
rs767621428 Chr.17: 44082694-44082696 on Build GRCh38
rs140165224 Chr.17: 44082621-44082621 on Build GRCh38
rs771514429 Chr.17: 44082873-44082873 on Build GRCh38
rs764459679 Chr.17: 44082518-44082518 on Build GRCh38
rs764563550 Chr.17: 44082637-44082637 on Build GRCh38
rs780351740 Chr.17: 44082565-44082565 on Build GRCh38
rs377542774 Chr.17: 44082738-44082738 on Build GRCh38
rs753327311 Chr.17: 44082719-44082719 on Build GRCh38
rs756236594 Chr.17: 44082683-44082683 on Build GRCh38
rs752165844 Chr.17: 44082529-44082529 on Build GRCh38
rs778736380 Chr.17: 44082704-44082704 on Build GRCh38
rs1027299624 Chr.17: 44082852-44082852 on Build GRCh38
rs369492491 Chr.17: 44082759-44082759 on Build GRCh38
rs200454255 Chr.17: 44082870-44082870 on Build GRCh38
rs374385731 Chr.17: 44082726-44082726 on Build GRCh38
rs751334636 Chr.17: 44082888-44082888 on Build GRCh38
rs143887884 Chr.17: 44082654-44082654 on Build GRCh38
rs754724677 Chr.17: 44082698-44082698 on Build GRCh38
rs1042265000 Chr.17: 44082681-44082681 on Build GRCh38
rs1028155562 Chr.17: 44082939-44082939 on Build GRCh38
rs374793409 Chr.17: 44082757-44082757 on Build GRCh38
rs376791446 Chr.17: 44082861-44082861 on Build GRCh38
rs199938552 Chr.17: 44082606-44082606 on Build GRCh38
rs373580384 Chr.17: 44082785-44082785 on Build GRCh38
rs373488804 Chr.17: 44082503-44082503 on Build GRCh38
rs985273735 Chr.17: 44082509-44082509 on Build GRCh38
rs774690985 Chr.17: 44082509-44082511 on Build GRCh38
rs777721425 Chr.17: 44082716-44082716 on Build GRCh38
rs575483176 Chr.17: 44082883-44082883 on Build GRCh38
rs760623534 Chr.17: 44082933-44082933 on Build GRCh38
rs762470534 Chr.17: 44082639-44082639 on Build GRCh38
rs770450250 Chr.17: 44082613-44082613 on Build GRCh38
rs371805085 Chr.17: 44082731-44082731 on Build GRCh38
rs929335531 Chr.17: 44082632-44082632 on Build GRCh38
rs570767798 Chr.17: 44082718-44082718 on Build GRCh38
rs545786430 Chr.17: 44082914-44082914 on Build GRCh38
rs113536871 Chr.17: 44082916-44082916 on Build GRCh38
rs915239382 Chr.17: 44082645-44082645 on Build GRCh38
rs201199523 Chr.17: 44082782-44082782 on Build GRCh38
rs774996601 Chr.17: 44082638-44082638 on Build GRCh38
rs973873221 Chr.17: 44082927-44082927 on Build GRCh38
rs188695612 Chr.17: 44082876-44082876 on Build GRCh38
rs144350170 Chr.17: 44082612-44082612 on Build GRCh38
rs202158617 Chr.17: 44082734-44082734 on Build GRCh38
rs1047683777 Chr.17: 44082692-44082692 on Build GRCh38
rs768086137 Chr.17: 44082736-44082736 on Build GRCh38
rs370764559 Chr.17: 44082569-44082569 on Build GRCh38
rs777666412 Chr.17: 44082808-44082808 on Build GRCh38
rs574625831 Chr.17: 44082844-44082844 on Build GRCh38
rs776120499 Chr.17: 44082720-44082720 on Build GRCh38
rs747949588 Chr.17: 44082714-44082714 on Build GRCh38
rs955284223 Chr.17: 44082506-44082506 on Build GRCh38
rs750487566 Chr.17: 44082680-44082680 on Build GRCh38
rs766546958 Chr.17: 44082750-44082750 on Build GRCh38
rs772653767 Chr.17: 44082611-44082611 on Build GRCh38
rs780815611 Chr.17: 44082862-44082862 on Build GRCh38
rs748150021 Chr.17: 44082597-44082597 on Build GRCh38
rs759606962 Chr.17: 44082533-44082538 on Build GRCh38

View Full Product Details