Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32894786-32895291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM54
Entrez Gene ID
113452
Gene Name
transmembrane protein 54
Gene Aliases
BCLP, CAC-1, CAC1
UniGene
Hs.534521
Gene Chromosome Location
Chr.1: 32894595-32901435 on Build GRCh38
Transcripts
NM_033504.2 XM_005270408.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4825470 c.668G>C p.*223S Chr.1: 32894806-32894806 on Build GRCh38
3773493 c.646A>T p.S216C Chr.1: 32894828-32894828 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746007733 Chr.1: 32894849-32894849 on Build GRCh38
rs746083236 Chr.1: 32894906-32894907 on Build GRCh38
rs144983176 Chr.1: 32895262-32895262 on Build GRCh38
rs772336341 Chr.1: 32894925-32894931 on Build GRCh38
rs765800514 Chr.1: 32894922-32894922 on Build GRCh38
rs747413698 Chr.1: 32895258-32895258 on Build GRCh38
rs769711526 Chr.1: 32894834-32894834 on Build GRCh38
rs774573271 Chr.1: 32894848-32894848 on Build GRCh38
rs768373550 Chr.1: 32894827-32894827 on Build GRCh38
rs565411544 Chr.1: 32895083-32895083 on Build GRCh38
rs1045058277 Chr.1: 32895053-32895053 on Build GRCh38
rs987063684 Chr.1: 32894875-32894875 on Build GRCh38
rs907705778 Chr.1: 32895001-32895001 on Build GRCh38
rs190204816 Chr.1: 32895223-32895223 on Build GRCh38
rs892633449 Chr.1: 32895144-32895144 on Build GRCh38
rs570652899 Chr.1: 32895230-32895230 on Build GRCh38
rs150537161 Chr.1: 32894811-32894811 on Build GRCh38
rs896429005 Chr.1: 32895051-32895051 on Build GRCh38
rs1030006856 Chr.1: 32894863-32894863 on Build GRCh38
rs147308790 Chr.1: 32894828-32894828 on Build GRCh38
rs578112733 Chr.1: 32895014-32895014 on Build GRCh38
rs1056306231 Chr.1: 32895040-32895040 on Build GRCh38
rs527445029 Chr.1: 32894825-32894838 on Build GRCh38
rs971885507 Chr.1: 32895237-32895237 on Build GRCh38
rs912858943 Chr.1: 32894876-32894876 on Build GRCh38
rs780372929 Chr.1: 32894928-32894930 on Build GRCh38
rs758184497 Chr.1: 32895011-32895011 on Build GRCh38
rs941830725 Chr.1: 32894938-32894938 on Build GRCh38
rs974436056 Chr.1: 32894940-32894940 on Build GRCh38
rs772914609 Chr.1: 32895070-32895070 on Build GRCh38
rs60869063 Chr.1: 32895143-32895143 on Build GRCh38
rs79707050 Chr.1: 32894961-32894961 on Build GRCh38
rs754546282 Chr.1: 32895219-32895219 on Build GRCh38
rs539478097 Chr.1: 32895052-32895052 on Build GRCh38
rs750148973 Chr.1: 32894906-32894906 on Build GRCh38
rs1028668292 Chr.1: 32895242-32895242 on Build GRCh38
rs140899806 Chr.1: 32894851-32894851 on Build GRCh38
rs527625041 Chr.1: 32894979-32894979 on Build GRCh38
rs148509739 Chr.1: 32894873-32894873 on Build GRCh38
rs868272561 Chr.1: 32894946-32894946 on Build GRCh38
rs997224151 Chr.1: 32895121-32895121 on Build GRCh38
rs957142456 Chr.1: 32894857-32894857 on Build GRCh38
rs539821033 Chr.1: 32895221-32895221 on Build GRCh38
rs573870806 Chr.1: 32895235-32895235 on Build GRCh38
rs1008836634 Chr.1: 32895147-32895147 on Build GRCh38
rs984994241 Chr.1: 32894969-32894969 on Build GRCh38
rs757227956 Chr.1: 32894819-32894820 on Build GRCh38
rs564281898 Chr.1: 32894868-32894868 on Build GRCh38
rs968131742 Chr.1: 32894820-32894820 on Build GRCh38
rs567930124 Chr.1: 32894874-32894874 on Build GRCh38
rs986085014 Chr.1: 32894881-32894881 on Build GRCh38
rs1000960075 Chr.1: 32895089-32895089 on Build GRCh38
rs775898295 Chr.1: 32894853-32894853 on Build GRCh38
rs377069821 Chr.1: 32895259-32895259 on Build GRCh38
rs750916916 Chr.1: 32894923-32894923 on Build GRCh38
rs780997711 Chr.1: 32894830-32894830 on Build GRCh38
rs538194697 Chr.1: 32894884-32894884 on Build GRCh38
rs757951992 Chr.1: 32895256-32895256 on Build GRCh38
rs903267777 Chr.1: 32894953-32894969 on Build GRCh38
rs199652031 Chr.1: 32894861-32894861 on Build GRCh38
rs953087772 Chr.1: 32895188-32895188 on Build GRCh38
rs764390268 Chr.1: 32894862-32894862 on Build GRCh38
rs373374840 Chr.1: 32895264-32895264 on Build GRCh38
rs879412084 Chr.1: 32894976-32894976 on Build GRCh38
rs975713107 Chr.1: 32894860-32894860 on Build GRCh38
rs1019866890 Chr.1: 32895154-32895154 on Build GRCh38

View Full Product Details