Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34983675-34984169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM35B
Entrez Gene ID
100506144
Gene Name
transmembrane protein 35B
Gene Aliases
ZMYM6NB
UniGene
Hs.533986
Gene Chromosome Location
Chr.1: 34981526-34985347 on Build GRCh38
Transcripts
NM_001195156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5214472 c.2163A>G p.A721A Chr.1: 34983931-34983931 on Build GRCh38
5566735 c.2159G>A p.R720H Chr.1: 34983935-34983935 on Build GRCh38
7202391 c.2162C>T p.A721V Chr.1: 34983932-34983932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044394344 Chr.1: 34983980-34983980 on Build GRCh38
rs769534297 Chr.1: 34983870-34983870 on Build GRCh38
rs78787188 Chr.1: 34983727-34983727 on Build GRCh38
rs947339412 Chr.1: 34983814-34983814 on Build GRCh38
rs746553898 Chr.1: 34983780-34983780 on Build GRCh38
rs914277171 Chr.1: 34984115-34984115 on Build GRCh38
rs991106584 Chr.1: 34984111-34984111 on Build GRCh38
rs114821631 Chr.1: 34983846-34983846 on Build GRCh38
rs542062160 Chr.1: 34983935-34983935 on Build GRCh38
rs1049726208 Chr.1: 34984051-34984051 on Build GRCh38
rs981570555 Chr.1: 34984032-34984032 on Build GRCh38
rs939083056 Chr.1: 34983914-34983914 on Build GRCh38
rs745682345 Chr.1: 34983793-34983793 on Build GRCh38
rs773073312 Chr.1: 34983908-34983908 on Build GRCh38
rs746726093 Chr.1: 34983998-34983998 on Build GRCh38
rs1018706521 Chr.1: 34984091-34984091 on Build GRCh38
rs772063692 Chr.1: 34983789-34983789 on Build GRCh38
rs962949415 Chr.1: 34983960-34983960 on Build GRCh38
rs539253999 Chr.1: 34984016-34984016 on Build GRCh38
rs958729414 Chr.1: 34984075-34984075 on Build GRCh38
rs554381153 Chr.1: 34984034-34984034 on Build GRCh38
rs752263252 Chr.1: 34984118-34984118 on Build GRCh38
rs760883742 Chr.1: 34983913-34983913 on Build GRCh38
rs760151728 Chr.1: 34983952-34983952 on Build GRCh38
rs377495374 Chr.1: 34983912-34983912 on Build GRCh38
rs1006876675 Chr.1: 34983936-34983936 on Build GRCh38
rs995799050 Chr.1: 34983965-34983965 on Build GRCh38
rs768108057 Chr.1: 34983796-34983796 on Build GRCh38
rs763380711 Chr.1: 34983741-34983741 on Build GRCh38
rs1056620945 Chr.1: 34983927-34983927 on Build GRCh38
rs891172382 Chr.1: 34984065-34984065 on Build GRCh38
rs951204006 Chr.1: 34983708-34983708 on Build GRCh38
rs947265742 Chr.1: 34984116-34984116 on Build GRCh38
rs1041623511 Chr.1: 34984124-34984124 on Build GRCh38
rs924408939 Chr.1: 34984132-34984132 on Build GRCh38
rs550206674 Chr.1: 34983828-34983828 on Build GRCh38
rs766448611 Chr.1: 34983911-34983911 on Build GRCh38
rs951587532 Chr.1: 34984000-34984000 on Build GRCh38
rs766870501 Chr.1: 34984014-34984014 on Build GRCh38
rs1051200301 Chr.1: 34983926-34983926 on Build GRCh38
rs1055614015 Chr.1: 34983766-34983766 on Build GRCh38
rs903025878 Chr.1: 34983869-34983869 on Build GRCh38
rs762768073 Chr.1: 34983910-34983910 on Build GRCh38
rs566256524 Chr.1: 34984101-34984101 on Build GRCh38
rs928255436 Chr.1: 34983728-34983728 on Build GRCh38
rs867167105 Chr.1: 34983833-34983833 on Build GRCh38
rs928423024 Chr.1: 34984056-34984056 on Build GRCh38
rs914084333 Chr.1: 34983962-34983962 on Build GRCh38
rs997291492 Chr.1: 34984007-34984007 on Build GRCh38

View Full Product Details