Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50460547-50461055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF2
Entrez Gene ID
80221
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
ACSMW, AVYV493
UniGene
Hs.288959
Gene Chromosome Location
Chr.17: 50426158-50474845 on Build GRCh38
Transcripts
NM_001288968.1 NM_001288969.1 NM_001288970.1 NM_001288971.1 NM_001288972.1 NM_025149.5 XM_006722110.3 XM_011525294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966975 c.228G>A p.Q76Q Chr.17: 50460776-50460776 on Build GRCh38
3665233 c.212G>A p.S71N Chr.17: 50460760-50460760 on Build GRCh38
4658971 c.214A>G p.K72E Chr.17: 50460762-50460762 on Build GRCh38
3519082 c.276C>T p.V92V Chr.17: 50460824-50460824 on Build GRCh38
4130383 c.129T>A p.S43R Chr.17: 50460677-50460677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766108906 Chr.17: 50460734-50460734 on Build GRCh38
rs779726036 Chr.17: 50460665-50460665 on Build GRCh38
rs199772678 Chr.17: 50460706-50460706 on Build GRCh38
rs769868081 Chr.17: 50460694-50460694 on Build GRCh38
rs560075310 Chr.17: 50460591-50460591 on Build GRCh38
rs775675070 Chr.17: 50460780-50460780 on Build GRCh38
rs541683291 Chr.17: 50460729-50460729 on Build GRCh38
rs746036854 Chr.17: 50460839-50460839 on Build GRCh38
rs1020324543 Chr.17: 50460810-50460810 on Build GRCh38
rs778806401 Chr.17: 50460861-50460861 on Build GRCh38
rs749411233 Chr.17: 50460709-50460709 on Build GRCh38
rs778333909 Chr.17: 50460661-50460661 on Build GRCh38
rs17856448 Chr.17: 50460772-50460772 on Build GRCh38
rs777182644 Chr.17: 50460804-50460804 on Build GRCh38
rs563110320 Chr.17: 50461001-50461001 on Build GRCh38
rs754666432 Chr.17: 50460742-50460742 on Build GRCh38
rs1038008553 Chr.17: 50461029-50461029 on Build GRCh38
rs140242443 Chr.17: 50460713-50460713 on Build GRCh38
rs933712581 Chr.17: 50460641-50460641 on Build GRCh38
rs781188170 Chr.17: 50460693-50460693 on Build GRCh38
rs761119663 Chr.17: 50460727-50460727 on Build GRCh38
rs201438196 Chr.17: 50460718-50460718 on Build GRCh38
rs900939040 Chr.17: 50460606-50460606 on Build GRCh38
rs756075797 Chr.17: 50460759-50460759 on Build GRCh38
rs764566310 Chr.17: 50460801-50460801 on Build GRCh38
rs774495027 Chr.17: 50460714-50460714 on Build GRCh38
rs941027238 Chr.17: 50461028-50461028 on Build GRCh38
rs762514280 Chr.17: 50460884-50460884 on Build GRCh38
rs934655254 Chr.17: 50460613-50460613 on Build GRCh38
rs879457609 Chr.17: 50460707-50460707 on Build GRCh38
rs754538592 Chr.17: 50460821-50460821 on Build GRCh38
rs928685221 Chr.17: 50460595-50460595 on Build GRCh38
rs1039834198 Chr.17: 50460598-50460598 on Build GRCh38
rs768055812 Chr.17: 50460655-50460655 on Build GRCh38
rs766731338 Chr.17: 50460628-50460628 on Build GRCh38
rs752740105 Chr.17: 50460571-50460571 on Build GRCh38
rs777847636 Chr.17: 50460840-50460840 on Build GRCh38
rs746018585 Chr.17: 50460776-50460776 on Build GRCh38
rs765744649 Chr.17: 50460811-50460811 on Build GRCh38
rs757956148 Chr.17: 50460735-50460735 on Build GRCh38
rs751239668 Chr.17: 50460741-50460741 on Build GRCh38
rs537528442 Chr.17: 50460775-50460775 on Build GRCh38
rs749364182 Chr.17: 50460763-50460763 on Build GRCh38
rs757231514 Chr.17: 50460857-50460857 on Build GRCh38
rs926173739 Chr.17: 50460577-50460577 on Build GRCh38
rs771199340 Chr.17: 50460771-50460771 on Build GRCh38
rs866369340 Chr.17: 50460792-50460792 on Build GRCh38
rs886815057 Chr.17: 50461025-50461026 on Build GRCh38
rs762450696 Chr.17: 50460805-50460805 on Build GRCh38
rs947481843 Chr.17: 50461030-50461030 on Build GRCh38
rs988877903 Chr.17: 50460614-50460614 on Build GRCh38
rs144020265 Chr.17: 50460722-50460722 on Build GRCh38
rs762563268 Chr.17: 50460667-50460667 on Build GRCh38
rs550962440 Chr.17: 50460728-50460728 on Build GRCh38
rs369904885 Chr.17: 50460689-50460689 on Build GRCh38
rs753209086 Chr.17: 50460658-50460658 on Build GRCh38
rs758210033 Chr.17: 50460778-50460778 on Build GRCh38
rs780369980 Chr.17: 50460887-50460887 on Build GRCh38
rs1043543929 Chr.17: 50460705-50460705 on Build GRCh38
rs866194893 Chr.17: 50460679-50460679 on Build GRCh38
rs975564480 Chr.17: 50460966-50460966 on Build GRCh38
rs746800710 Chr.17: 50460667-50460667 on Build GRCh38
rs947445943 Chr.17: 50460691-50460691 on Build GRCh38
rs777626653 Chr.17: 50460782-50460782 on Build GRCh38
rs117744567 Chr.17: 50460996-50460996 on Build GRCh38
rs756617568 Chr.17: 50460660-50460660 on Build GRCh38
rs776988366 Chr.17: 50460924-50460924 on Build GRCh38
rs146454487 Chr.17: 50460825-50460825 on Build GRCh38
rs749953094 Chr.17: 50460662-50460662 on Build GRCh38
rs756377882 Chr.17: 50460984-50460984 on Build GRCh38
rs200747876 Chr.17: 50460675-50460675 on Build GRCh38
rs970336445 Chr.17: 50460584-50460584 on Build GRCh38
rs753761055 Chr.17: 50460852-50460852 on Build GRCh38
rs549006469 Chr.17: 50460645-50460645 on Build GRCh38
rs759938793 Chr.17: 50460715-50460715 on Build GRCh38
rs370674405 Chr.17: 50460892-50460892 on Build GRCh38
rs777898840 Chr.17: 50460760-50460760 on Build GRCh38
rs769077684 Chr.17: 50460921-50460921 on Build GRCh38
rs529216314 Chr.17: 50460632-50460632 on Build GRCh38
rs767231964 Chr.17: 50460824-50460824 on Build GRCh38
rs55706027 Chr.17: 50460998-50460998 on Build GRCh38
rs758002982 Chr.17: 50460664-50460664 on Build GRCh38
rs766045803 Chr.17: 50460737-50460737 on Build GRCh38
rs751095750 Chr.17: 50460820-50460820 on Build GRCh38
rs182248172 Chr.17: 50460756-50460756 on Build GRCh38

View Full Product Details