Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37682728-37683234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf109
Entrez Gene ID
54955
Gene Name
chromosome 1 open reading frame 109
Gene Aliases
-
UniGene
Hs.272673
Gene Chromosome Location
Chr.1: 37681570-37692336 on Build GRCh38
Transcripts
NM_001303030.1 NM_001303031.1 NM_017850.2 XM_005270975.4 XM_005270979.1 XM_005270980.1 XM_005270981.4 XM_005270982.1 XM_011541637.1 XM_011541638.1 XM_011541639.1 XM_011541640.1 XM_011541641.1 XM_011541642.1 XM_011541643.1 XM_011541646.2 XM_017001544.1 XM_017001545.1 XM_017001546.1 XM_017001547.1 XM_017001548.1 XM_017001549.1 XM_017001550.1 XM_017001551.1 XM_017001552.1 XM_017001553.1 XM_017001554.1 XM_017001555.1 XM_017001556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527629 c.593C>T p.S198F Chr.1: 37683142-37683142 on Build GRCh38
5380898 c.579C>T p.I193I Chr.1: 37683156-37683156 on Build GRCh38
7351556 c.575-1G>T p.? Chr.1: 37683161-37683161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374234641 Chr.1: 37683111-37683111 on Build GRCh38
rs1038093426 Chr.1: 37683015-37683015 on Build GRCh38
rs1031835108 Chr.1: 37683112-37683112 on Build GRCh38
rs540214611 Chr.1: 37682835-37682835 on Build GRCh38
rs553787824 Chr.1: 37682971-37682971 on Build GRCh38
rs533734506 Chr.1: 37682778-37682778 on Build GRCh38
rs779248512 Chr.1: 37683163-37683163 on Build GRCh38
rs966863893 Chr.1: 37682908-37682908 on Build GRCh38
rs575576456 Chr.1: 37683067-37683067 on Build GRCh38
rs948345307 Chr.1: 37683046-37683046 on Build GRCh38
rs749556908 Chr.1: 37682940-37682940 on Build GRCh38
rs555460300 Chr.1: 37682821-37682821 on Build GRCh38
rs933849559 Chr.1: 37682892-37682892 on Build GRCh38
rs139692993 Chr.1: 37682862-37682862 on Build GRCh38
rs781477657 Chr.1: 37683122-37683122 on Build GRCh38
rs80108999 Chr.1: 37683202-37683202 on Build GRCh38
rs375986196 Chr.1: 37683187-37683187 on Build GRCh38
rs878881016 Chr.1: 37682897-37682897 on Build GRCh38
rs146821670 Chr.1: 37683113-37683113 on Build GRCh38
rs939788936 Chr.1: 37682997-37682997 on Build GRCh38
rs574705596 Chr.1: 37682802-37682802 on Build GRCh38
rs529361208 Chr.1: 37683205-37683205 on Build GRCh38
rs775982245 Chr.1: 37683195-37683195 on Build GRCh38
rs370918377 Chr.1: 37683097-37683097 on Build GRCh38
rs765247078 Chr.1: 37683210-37683210 on Build GRCh38
rs780948989 Chr.1: 37682848-37682848 on Build GRCh38
rs143559169 Chr.1: 37683012-37683012 on Build GRCh38
rs1040190778 Chr.1: 37682787-37682787 on Build GRCh38
rs1025064537 Chr.1: 37682890-37682890 on Build GRCh38
rs564874909 Chr.1: 37682847-37682847 on Build GRCh38
rs776151968 Chr.1: 37683045-37683045 on Build GRCh38
rs746722539 Chr.1: 37683200-37683200 on Build GRCh38
rs879117176 Chr.1: 37683061-37683061 on Build GRCh38
rs761175871 Chr.1: 37683204-37683204 on Build GRCh38
rs372472705 Chr.1: 37683118-37683118 on Build GRCh38
rs756917079 Chr.1: 37682833-37682834 on Build GRCh38
rs555052239 Chr.1: 37682757-37682757 on Build GRCh38
rs749352194 Chr.1: 37683152-37683152 on Build GRCh38
rs141474570 Chr.1: 37683161-37683161 on Build GRCh38
rs774139218 Chr.1: 37683104-37683104 on Build GRCh38
rs111962243 Chr.1: 37683176-37683176 on Build GRCh38
rs759408696 Chr.1: 37683136-37683136 on Build GRCh38
rs975322700 Chr.1: 37682859-37682859 on Build GRCh38
rs760274537 Chr.1: 37683096-37683096 on Build GRCh38
rs201746884 Chr.1: 37683190-37683190 on Build GRCh38
rs986644567 Chr.1: 37682931-37682931 on Build GRCh38
rs562244764 Chr.1: 37683203-37683203 on Build GRCh38
rs777874924 Chr.1: 37683145-37683145 on Build GRCh38
rs771787576 Chr.1: 37683088-37683088 on Build GRCh38
rs1013793853 Chr.1: 37682887-37682887 on Build GRCh38
rs964655988 Chr.1: 37682855-37682855 on Build GRCh38
rs929345051 Chr.1: 37682832-37682832 on Build GRCh38
rs770496172 Chr.1: 37682849-37682849 on Build GRCh38
rs750250750 Chr.1: 37683114-37683114 on Build GRCh38
rs684260 Chr.1: 37683093-37683093 on Build GRCh38
rs752977175 Chr.1: 37683131-37683131 on Build GRCh38
rs756327464 Chr.1: 37683134-37683134 on Build GRCh38
rs567227620 Chr.1: 37682824-37682824 on Build GRCh38

View Full Product Details