Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79733350-79733849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP7
Entrez Gene ID
339221
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Gene Aliases
ALK-SMase, E-NPP 7, NPP-7, NPP7
UniGene
Hs.114084
Gene Chromosome Location
Chr.17: 79730919-79742926 on Build GRCh38
Transcripts
NM_178543.4 XM_011524737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4070301 c.342C>T p.G114G Chr.17: 79733596-79733596 on Build GRCh38
4886736 c.365G>T p.G122V Chr.17: 79733619-79733619 on Build GRCh38
3680602 c.306C>T p.T102T Chr.17: 79733560-79733560 on Build GRCh38
6397033 c.274G>T p.G92W Chr.17: 79733528-79733528 on Build GRCh38
7252308 c.330C>T p.H110H Chr.17: 79733584-79733584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368328037 Chr.17: 79733672-79733672 on Build GRCh38
rs145931726 Chr.17: 79733624-79733624 on Build GRCh38
rs200905387 Chr.17: 79733608-79733608 on Build GRCh38
rs201200066 Chr.17: 79733493-79733493 on Build GRCh38
rs201934331 Chr.17: 79733623-79733623 on Build GRCh38
rs974599689 Chr.17: 79733425-79733425 on Build GRCh38
rs782707558 Chr.17: 79733538-79733538 on Build GRCh38
rs782534221 Chr.17: 79733490-79733490 on Build GRCh38
rs978348808 Chr.17: 79733768-79733768 on Build GRCh38
rs782068681 Chr.17: 79733652-79733652 on Build GRCh38
rs200178880 Chr.17: 79733691-79733691 on Build GRCh38
rs188386728 Chr.17: 79733479-79733479 on Build GRCh38
rs574897072 Chr.17: 79733391-79733391 on Build GRCh38
rs781800266 Chr.17: 79733682-79733682 on Build GRCh38
rs59356172 Chr.17: 79733371-79733371 on Build GRCh38
rs142356062 Chr.17: 79733589-79733589 on Build GRCh38
rs943152806 Chr.17: 79733387-79733387 on Build GRCh38
rs576866254 Chr.17: 79733809-79733809 on Build GRCh38
rs544115844 Chr.17: 79733815-79733815 on Build GRCh38
rs782586363 Chr.17: 79733508-79733508 on Build GRCh38
rs782411863 Chr.17: 79733686-79733686 on Build GRCh38
rs898628146 Chr.17: 79733653-79733653 on Build GRCh38
rs371802382 Chr.17: 79733401-79733401 on Build GRCh38
rs782214637 Chr.17: 79733516-79733516 on Build GRCh38
rs113286331 Chr.17: 79733484-79733484 on Build GRCh38
rs782001647 Chr.17: 79733480-79733480 on Build GRCh38
rs79403711 Chr.17: 79733451-79733451 on Build GRCh38
rs782155263 Chr.17: 79733668-79733668 on Build GRCh38
rs782243955 Chr.17: 79733684-79733684 on Build GRCh38
rs534325591 Chr.17: 79733574-79733574 on Build GRCh38
rs12453180 Chr.17: 79733655-79733655 on Build GRCh38
rs11150740 Chr.17: 79733694-79733694 on Build GRCh38
rs967846459 Chr.17: 79733767-79733767 on Build GRCh38
rs782738635 Chr.17: 79733485-79733485 on Build GRCh38
rs782312167 Chr.17: 79733617-79733617 on Build GRCh38
rs782543703 Chr.17: 79733674-79733674 on Build GRCh38
rs201722006 Chr.17: 79733478-79733478 on Build GRCh38
rs765929853 Chr.17: 79733590-79733590 on Build GRCh38
rs572470530 Chr.17: 79733656-79733656 on Build GRCh38
rs199658107 Chr.17: 79733618-79733618 on Build GRCh38
rs1018846936 Chr.17: 79733531-79733531 on Build GRCh38
rs782150305 Chr.17: 79733642-79733642 on Build GRCh38
rs1022230444 Chr.17: 79733737-79733737 on Build GRCh38
rs781955212 Chr.17: 79733695-79733695 on Build GRCh38
rs911803464 Chr.17: 79733375-79733375 on Build GRCh38
rs782411680 Chr.17: 79733468-79733468 on Build GRCh38
rs781950270 Chr.17: 79733541-79733541 on Build GRCh38
rs782268065 Chr.17: 79733465-79733465 on Build GRCh38
rs556541327 Chr.17: 79733381-79733381 on Build GRCh38
rs1008217530 Chr.17: 79733673-79733673 on Build GRCh38
rs964655057 Chr.17: 79733547-79733547 on Build GRCh38
rs914984004 Chr.17: 79733450-79733450 on Build GRCh38
rs782642834 Chr.17: 79733458-79733458 on Build GRCh38
rs542262808 Chr.17: 79733429-79733429 on Build GRCh38
rs782437623 Chr.17: 79733507-79733507 on Build GRCh38
rs180944528 Chr.17: 79733481-79733481 on Build GRCh38
rs996057330 Chr.17: 79733710-79733710 on Build GRCh38
rs782332512 Chr.17: 79733457-79733457 on Build GRCh38
rs782439906 Chr.17: 79733643-79733643 on Build GRCh38
rs782006820 Chr.17: 79733640-79733640 on Build GRCh38
rs781859150 Chr.17: 79733486-79733486 on Build GRCh38
rs574138520 Chr.17: 79733519-79733519 on Build GRCh38
rs78658284 Chr.17: 79733584-79733584 on Build GRCh38
rs782120928 Chr.17: 79733660-79733660 on Build GRCh38
rs782093477 Chr.17: 79733546-79733546 on Build GRCh38
rs34685219 Chr.17: 79733814-79733814 on Build GRCh38
rs201000413 Chr.17: 79733518-79733518 on Build GRCh38
rs200647393 Chr.17: 79733560-79733560 on Build GRCh38
rs782183600 Chr.17: 79733692-79733692 on Build GRCh38
rs782045800 Chr.17: 79733528-79733528 on Build GRCh38
rs535733167 Chr.17: 79733648-79733648 on Build GRCh38
rs782776361 Chr.17: 79733559-79733559 on Build GRCh38
rs375922133 Chr.17: 79733373-79733373 on Build GRCh38
rs782679869 Chr.17: 79733688-79733688 on Build GRCh38
rs960766075 Chr.17: 79733818-79733818 on Build GRCh38
rs782527401 Chr.17: 79733591-79733591 on Build GRCh38
rs755529885 Chr.17: 79733683-79733683 on Build GRCh38
rs138305176 Chr.17: 79733566-79733566 on Build GRCh38
rs571036557 Chr.17: 79733585-79733585 on Build GRCh38
rs1042074871 Chr.17: 79733456-79733456 on Build GRCh38
rs375672909 Chr.17: 79733667-79733667 on Build GRCh38
rs752032407 Chr.17: 79733676-79733676 on Build GRCh38
rs782302021 Chr.17: 79733504-79733504 on Build GRCh38
rs782287883 Chr.17: 79733611-79733611 on Build GRCh38

View Full Product Details