Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81511811-81512310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTG1
Entrez Gene ID
71
Gene Name
actin gamma 1
Gene Aliases
ACT, ACTG, BRWS2, DFNA20, DFNA26, HEL-176
UniGene
Hs.514581
Gene Chromosome Location
Chr.17: 81509971-81512866 on Build GRCh38
Transcripts
NM_001199954.1 NM_001614.3 NR_037688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6021703 c.238G>A p.D80N Chr.17: 81512028-81512028 on Build GRCh38
4139949 c.136G>A p.G46S Chr.17: 81512130-81512130 on Build GRCh38
6048728 c.209_210insC p.I71fs*2 Chr.17: 81512056-81512057 on Build GRCh38
5577944 c.324C>T p.A108A Chr.17: 81511942-81511942 on Build GRCh38
6082553 c.136G>T p.G46C Chr.17: 81512130-81512130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11549193 Chr.17: 81512250-81512250 on Build GRCh38
rs782500482 Chr.17: 81512026-81512026 on Build GRCh38
rs782763504 Chr.17: 81512214-81512214 on Build GRCh38
rs538822936 Chr.17: 81512037-81512037 on Build GRCh38
rs782059003 Chr.17: 81511936-81511936 on Build GRCh38
rs11549238 Chr.17: 81512256-81512256 on Build GRCh38
rs202223445 Chr.17: 81512207-81512207 on Build GRCh38
rs782796849 Chr.17: 81512219-81512219 on Build GRCh38
rs28999111 Chr.17: 81512000-81512000 on Build GRCh38
rs201275526 Chr.17: 81512056-81512056 on Build GRCh38
rs782817444 Chr.17: 81512008-81512008 on Build GRCh38
rs202103644 Chr.17: 81511908-81511908 on Build GRCh38
rs373618606 Chr.17: 81512148-81512148 on Build GRCh38
rs374907377 Chr.17: 81512128-81512128 on Build GRCh38
rs781831072 Chr.17: 81512023-81512023 on Build GRCh38
rs781870017 Chr.17: 81511854-81511854 on Build GRCh38
rs11549230 Chr.17: 81512274-81512274 on Build GRCh38
rs562047291 Chr.17: 81512241-81512241 on Build GRCh38
rs370149113 Chr.17: 81511873-81511873 on Build GRCh38
rs770592789 Chr.17: 81512189-81512189 on Build GRCh38
rs782205549 Chr.17: 81511896-81511896 on Build GRCh38
rs11549188 Chr.17: 81511968-81511968 on Build GRCh38
rs782047199 Chr.17: 81512053-81512053 on Build GRCh38
rs374327916 Chr.17: 81512159-81512159 on Build GRCh38
rs115307446 Chr.17: 81512156-81512156 on Build GRCh38
rs11549229 Chr.17: 81511962-81511962 on Build GRCh38
rs202038633 Chr.17: 81511847-81511847 on Build GRCh38
rs372370199 Chr.17: 81511948-81511948 on Build GRCh38
rs727502884 Chr.17: 81511933-81511933 on Build GRCh38
rs781910131 Chr.17: 81512171-81512171 on Build GRCh38
rs11549231 Chr.17: 81512260-81512260 on Build GRCh38
rs782351704 Chr.17: 81512201-81512201 on Build GRCh38
rs11549174 Chr.17: 81512235-81512235 on Build GRCh38
rs199979742 Chr.17: 81511872-81511872 on Build GRCh38
rs11549220 Chr.17: 81512089-81512089 on Build GRCh38
rs781861355 Chr.17: 81512216-81512216 on Build GRCh38
rs199955399 Chr.17: 81512259-81512259 on Build GRCh38
rs116329164 Chr.17: 81511898-81511898 on Build GRCh38
rs376850595 Chr.17: 81512151-81512151 on Build GRCh38
rs386799895 Chr.17: 81511844-81511846 on Build GRCh38
rs782658400 Chr.17: 81511871-81511871 on Build GRCh38
rs782542290 Chr.17: 81511859-81511859 on Build GRCh38
rs782204140 Chr.17: 81512039-81512039 on Build GRCh38
rs369910719 Chr.17: 81512195-81512195 on Build GRCh38
rs144338558 Chr.17: 81512086-81512086 on Build GRCh38
rs782028455 Chr.17: 81512051-81512051 on Build GRCh38
rs9910792 Chr.17: 81511890-81511890 on Build GRCh38
rs147001458 Chr.17: 81512011-81512011 on Build GRCh38
rs11549214 Chr.17: 81512124-81512124 on Build GRCh38
rs782566714 Chr.17: 81512141-81512141 on Build GRCh38
rs532474677 Chr.17: 81511841-81511841 on Build GRCh38
rs11549199 Chr.17: 81512090-81512090 on Build GRCh38
rs201043115 Chr.17: 81511879-81511879 on Build GRCh38
rs782713618 Chr.17: 81512110-81512110 on Build GRCh38
rs782510294 Chr.17: 81511868-81511868 on Build GRCh38
rs11549197 Chr.17: 81512140-81512140 on Build GRCh38
rs536366580 Chr.17: 81512062-81512062 on Build GRCh38
rs569289825 Chr.17: 81511843-81511843 on Build GRCh38
rs140152735 Chr.17: 81512076-81512076 on Build GRCh38
rs11549181 Chr.17: 81511943-81511943 on Build GRCh38
rs782608331 Chr.17: 81512147-81512147 on Build GRCh38
rs782800711 Chr.17: 81511861-81511861 on Build GRCh38
rs146402466 Chr.17: 81512101-81512101 on Build GRCh38
rs11549176 Chr.17: 81511931-81511931 on Build GRCh38
rs149057480 Chr.17: 81511921-81511921 on Build GRCh38
rs782798449 Chr.17: 81512168-81512168 on Build GRCh38
rs11549185 Chr.17: 81512045-81512045 on Build GRCh38
rs200292222 Chr.17: 81511927-81511927 on Build GRCh38
rs782370155 Chr.17: 81512158-81512158 on Build GRCh38
rs11549245 Chr.17: 81512077-81512077 on Build GRCh38
rs781938714 Chr.17: 81511848-81511848 on Build GRCh38
rs138240892 Chr.17: 81512074-81512074 on Build GRCh38
rs183867515 Chr.17: 81512206-81512206 on Build GRCh38
rs782481421 Chr.17: 81512146-81512146 on Build GRCh38
rs782388764 Chr.17: 81511986-81511986 on Build GRCh38
rs11549209 Chr.17: 81511996-81511996 on Build GRCh38
rs1057518673 Chr.17: 81512237-81512237 on Build GRCh38
rs201279208 Chr.17: 81512150-81512150 on Build GRCh38
rs782197648 Chr.17: 81511963-81511963 on Build GRCh38
rs376965053 Chr.17: 81511862-81511862 on Build GRCh38
rs566543337 Chr.17: 81511990-81511990 on Build GRCh38
rs782297602 Chr.17: 81511905-81511905 on Build GRCh38
rs782338610 Chr.17: 81511877-81511877 on Build GRCh38
rs781955025 Chr.17: 81511863-81511863 on Build GRCh38
rs782108009 Chr.17: 81512270-81512270 on Build GRCh38
rs11549172 Chr.17: 81511961-81511961 on Build GRCh38
rs782730692 Chr.17: 81512173-81512173 on Build GRCh38
rs782183612 Chr.17: 81511895-81511895 on Build GRCh38
rs782569269 Chr.17: 81511960-81511960 on Build GRCh38
rs371967814 Chr.17: 81512278-81512278 on Build GRCh38
rs377741704 Chr.17: 81512180-81512180 on Build GRCh38
rs782137311 Chr.17: 81512217-81512217 on Build GRCh38
rs782056092 Chr.17: 81512109-81512109 on Build GRCh38
rs11549228 Chr.17: 81512277-81512277 on Build GRCh38
rs782556001 Chr.17: 81511839-81511839 on Build GRCh38
rs781879811 Chr.17: 81512190-81512190 on Build GRCh38
rs782549602 Chr.17: 81511956-81511956 on Build GRCh38
rs782275620 Chr.17: 81512244-81512244 on Build GRCh38
rs782512698 Chr.17: 81512233-81512233 on Build GRCh38
rs201951886 Chr.17: 81512172-81512172 on Build GRCh38
rs139517777 Chr.17: 81512283-81512283 on Build GRCh38
rs139059365 Chr.17: 81512014-81512014 on Build GRCh38
rs368299537 Chr.17: 81512098-81512098 on Build GRCh38
rs11549239 Chr.17: 81511966-81511966 on Build GRCh38
rs11549224 Chr.17: 81512067-81512067 on Build GRCh38
rs782198284 Chr.17: 81511874-81511874 on Build GRCh38
rs782044225 Chr.17: 81511886-81511886 on Build GRCh38
rs782100140 Chr.17: 81512211-81512211 on Build GRCh38
rs536476533 Chr.17: 81512224-81512224 on Build GRCh38
rs781886356 Chr.17: 81512181-81512182 on Build GRCh38
rs782754511 Chr.17: 81512065-81512065 on Build GRCh38
rs571472114 Chr.17: 81512174-81512174 on Build GRCh38
rs140398667 Chr.17: 81511999-81511999 on Build GRCh38
rs550758256 Chr.17: 81512203-81512203 on Build GRCh38
rs782271399 Chr.17: 81511837-81511837 on Build GRCh38
rs782540819 Chr.17: 81511893-81511893 on Build GRCh38
rs11549243 Chr.17: 81512267-81512267 on Build GRCh38
rs782460937 Chr.17: 81512280-81512280 on Build GRCh38
rs372586603 Chr.17: 81512095-81512095 on Build GRCh38
rs11549244 Chr.17: 81512242-81512242 on Build GRCh38
rs281875325 Chr.17: 81511907-81511907 on Build GRCh38
rs782227962 Chr.17: 81511897-81511897 on Build GRCh38
rs200000684 Chr.17: 81511894-81511894 on Build GRCh38
rs551034127 Chr.17: 81511842-81511842 on Build GRCh38
rs148632011 Chr.17: 81511838-81511838 on Build GRCh38
rs148759022 Chr.17: 81512087-81512087 on Build GRCh38
rs11549216 Chr.17: 81512105-81512105 on Build GRCh38
rs782617619 Chr.17: 81511975-81511975 on Build GRCh38
rs782594584 Chr.17: 81512017-81512017 on Build GRCh38
rs782193841 Chr.17: 81511840-81511840 on Build GRCh38
rs9912409 Chr.17: 81511845-81511845 on Build GRCh38
rs782249707 Chr.17: 81512047-81512047 on Build GRCh38
rs782660034 Chr.17: 81512149-81512149 on Build GRCh38
rs782654999 Chr.17: 81512084-81512084 on Build GRCh38
rs782350069 Chr.17: 81511918-81511918 on Build GRCh38
rs577515884 Chr.17: 81512117-81512117 on Build GRCh38
rs11549232 Chr.17: 81512268-81512268 on Build GRCh38
rs75355043 Chr.17: 81511846-81511846 on Build GRCh38
rs552858239 Chr.17: 81511864-81511864 on Build GRCh38
rs782205858 Chr.17: 81512092-81512092 on Build GRCh38
rs367960707 Chr.17: 81512209-81512209 on Build GRCh38
rs147032031 Chr.17: 81512068-81512068 on Build GRCh38
rs200210656 Chr.17: 81512129-81512129 on Build GRCh38
rs149682185 Chr.17: 81512289-81512289 on Build GRCh38
rs11549236 Chr.17: 81512236-81512236 on Build GRCh38
rs11549233 Chr.17: 81512064-81512064 on Build GRCh38
rs202094234 Chr.17: 81511951-81511951 on Build GRCh38
rs886041280 Chr.17: 81512135-81512135 on Build GRCh38
rs200772616 Chr.17: 81511915-81511915 on Build GRCh38
rs202102245 Chr.17: 81512264-81512264 on Build GRCh38
rs553335992 Chr.17: 81512032-81512032 on Build GRCh38
rs782815041 Chr.17: 81512113-81512113 on Build GRCh38
rs782399347 Chr.17: 81512265-81512265 on Build GRCh38
rs142131810 Chr.17: 81511887-81511887 on Build GRCh38
rs782119646 Chr.17: 81512253-81512253 on Build GRCh38
rs782397522 Chr.17: 81512050-81512050 on Build GRCh38
rs782129674 Chr.17: 81512166-81512166 on Build GRCh38
rs782504245 Chr.17: 81512082-81512082 on Build GRCh38
rs143028649 Chr.17: 81512038-81512038 on Build GRCh38
rs104894544 Chr.17: 81511913-81511913 on Build GRCh38
rs782127437 Chr.17: 81512200-81512200 on Build GRCh38
rs267606630 Chr.17: 81511912-81511912 on Build GRCh38
rs9912403 Chr.17: 81511844-81511844 on Build GRCh38
rs782132566 Chr.17: 81511883-81511883 on Build GRCh38
rs548138575 Chr.17: 81511955-81511955 on Build GRCh38
rs375450454 Chr.17: 81512035-81512035 on Build GRCh38
rs370205158 Chr.17: 81512202-81512202 on Build GRCh38
rs782165228 Chr.17: 81511987-81511987 on Build GRCh38
rs782606686 Chr.17: 81512227-81512227 on Build GRCh38
rs139751304 Chr.17: 81512107-81512107 on Build GRCh38
rs782768875 Chr.17: 81512273-81512273 on Build GRCh38
rs782757560 Chr.17: 81511891-81511891 on Build GRCh38
rs373830909 Chr.17: 81511851-81511851 on Build GRCh38
rs550936698 Chr.17: 81511942-81511942 on Build GRCh38
rs782205195 Chr.17: 81512196-81512196 on Build GRCh38
rs782473240 Chr.17: 81512075-81512075 on Build GRCh38
rs782142072 Chr.17: 81512005-81512005 on Build GRCh38
rs782033812 Chr.17: 81511884-81511884 on Build GRCh38
rs782777443 Chr.17: 81512122-81512122 on Build GRCh38
rs781953399 Chr.17: 81512060-81512060 on Build GRCh38
rs368844224 Chr.17: 81512044-81512044 on Build GRCh38
rs782049770 Chr.17: 81511845-81511846 on Build GRCh38
rs782810052 Chr.17: 81511866-81511867 on Build GRCh38
rs199905564 Chr.17: 81511900-81511900 on Build GRCh38
rs143798951 Chr.17: 81512145-81512145 on Build GRCh38
rs782236244 Chr.17: 81511981-81511981 on Build GRCh38
rs782345089 Chr.17: 81512041-81512041 on Build GRCh38
rs532725688 Chr.17: 81511930-81511930 on Build GRCh38
rs375159565 Chr.17: 81512269-81512269 on Build GRCh38

View Full Product Details