Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26017168-26017667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SS18
Entrez Gene ID
6760
Gene Name
SS18, nBAF chromatin remodeling complex subunit
Gene Aliases
SSXT, SYT
UniGene
Hs.129261
Gene Chromosome Location
Chr.18: 26016253-26091217 on Build GRCh38
Transcripts
NM_001007559.2 NM_001308201.1 NM_005637.3 XM_006722527.2 XM_011526145.1 XM_011526147.2 XM_011526148.2 XM_011526149.2 XM_011526150.2 XM_011526151.2 XM_011526152.2

SNP Detail

SNP ID Chromosome Location
rs914658082 Chr.18: 26017216-26017216 on Build GRCh38
rs1057235063 Chr.18: 26017358-26017358 on Build GRCh38
rs868439784 Chr.18: 26017377-26017377 on Build GRCh38
rs376581468 Chr.18: 26017274-26017274 on Build GRCh38
rs894602351 Chr.18: 26017371-26017371 on Build GRCh38
rs747267910 Chr.18: 26017205-26017205 on Build GRCh38
rs547028654 Chr.18: 26017509-26017509 on Build GRCh38
rs551175314 Chr.18: 26017243-26017243 on Build GRCh38
rs947396760 Chr.18: 26017222-26017222 on Build GRCh38
rs887518406 Chr.18: 26017445-26017445 on Build GRCh38
rs533432087 Chr.18: 26017479-26017479 on Build GRCh38
rs938914874 Chr.18: 26017296-26017296 on Build GRCh38
rs531170675 Chr.18: 26017229-26017229 on Build GRCh38
rs562236203 Chr.18: 26017195-26017195 on Build GRCh38
rs1045257002 Chr.18: 26017402-26017402 on Build GRCh38
rs535996469 Chr.18: 26017467-26017471 on Build GRCh38
rs189662043 Chr.18: 26017316-26017316 on Build GRCh38
rs1030129995 Chr.18: 26017190-26017190 on Build GRCh38
rs552995006 Chr.18: 26017398-26017398 on Build GRCh38
rs761877169 Chr.18: 26017388-26017388 on Build GRCh38
rs866465261 Chr.18: 26017608-26017608 on Build GRCh38

View Full Product Details