Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26032293-26032829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SS18
Entrez Gene ID
6760
Gene Name
SS18, nBAF chromatin remodeling complex subunit
Gene Aliases
SSXT, SYT
UniGene
Hs.129261
Gene Chromosome Location
Chr.18: 26016253-26091217 on Build GRCh38
Transcripts
NM_001007559.2 NM_001308201.1 NM_005637.3 XM_006722527.2 XM_011526145.1 XM_011526147.2 XM_011526148.2 XM_011526149.2 XM_011526150.2 XM_011526151.2 XM_011526152.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672655 c.1217A>C p.Y406S Chr.18: 26032412-26032412 on Build GRCh38
6148983 c.1167A>G p.G389G Chr.18: 26032462-26032462 on Build GRCh38
2808065 c.1149A>T p.Q383H Chr.18: 26032480-26032480 on Build GRCh38
96215 c.1116A>G p.P372P Chr.18: 26032513-26032513 on Build GRCh38
1193194 c.1165G>A p.G389R Chr.18: 26032464-26032464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979306360 Chr.18: 26032693-26032693 on Build GRCh38
rs760942449 Chr.18: 26032438-26032438 on Build GRCh38
rs887329322 Chr.18: 26032432-26032432 on Build GRCh38
rs761293756 Chr.18: 26032571-26032571 on Build GRCh38
rs527332079 Chr.18: 26032360-26032360 on Build GRCh38
rs61737050 Chr.18: 26032520-26032520 on Build GRCh38
rs773560125 Chr.18: 26032568-26032568 on Build GRCh38
rs754179506 Chr.18: 26032349-26032349 on Build GRCh38
rs200720553 Chr.18: 26032384-26032384 on Build GRCh38
rs564838173 Chr.18: 26032330-26032330 on Build GRCh38
rs989882329 Chr.18: 26032745-26032745 on Build GRCh38
rs529695041 Chr.18: 26032472-26032472 on Build GRCh38
rs754213987 Chr.18: 26032574-26032574 on Build GRCh38
rs373793671 Chr.18: 26032354-26032354 on Build GRCh38
rs769701366 Chr.18: 26032371-26032371 on Build GRCh38
rs567310634 Chr.18: 26032459-26032459 on Build GRCh38
rs534076761 Chr.18: 26032782-26032782 on Build GRCh38
rs769458598 Chr.18: 26032496-26032496 on Build GRCh38
rs150400353 Chr.18: 26032577-26032577 on Build GRCh38
rs779777593 Chr.18: 26032504-26032504 on Build GRCh38
rs574533870 Chr.18: 26032765-26032765 on Build GRCh38
rs773812449 Chr.18: 26032417-26032417 on Build GRCh38
rs757924493 Chr.18: 26032565-26032567 on Build GRCh38
rs372907379 Chr.18: 26032427-26032427 on Build GRCh38
rs749690456 Chr.18: 26032530-26032530 on Build GRCh38
rs762646011 Chr.18: 26032381-26032381 on Build GRCh38
rs946624503 Chr.18: 26032650-26032650 on Build GRCh38
rs910494091 Chr.18: 26032720-26032720 on Build GRCh38
rs758380832 Chr.18: 26032582-26032582 on Build GRCh38
rs774008004 Chr.18: 26032545-26032545 on Build GRCh38
rs878971864 Chr.18: 26032519-26032519 on Build GRCh38
rs771680846 Chr.18: 26032564-26032564 on Build GRCh38
rs779306175 Chr.18: 26032468-26032468 on Build GRCh38
rs116165792 Chr.18: 26032756-26032756 on Build GRCh38
rs200000768 Chr.18: 26032558-26032558 on Build GRCh38
rs960745390 Chr.18: 26032604-26032604 on Build GRCh38
rs749115830 Chr.18: 26032509-26032509 on Build GRCh38
rs1028762506 Chr.18: 26032736-26032736 on Build GRCh38
rs764609036 Chr.18: 26032462-26032467 on Build GRCh38
rs376179585 Chr.18: 26032366-26032366 on Build GRCh38
rs770948013 Chr.18: 26032355-26032355 on Build GRCh38
rs776088570 Chr.18: 26032373-26032373 on Build GRCh38
rs887113468 Chr.18: 26032613-26032613 on Build GRCh38
rs774977693 Chr.18: 26032379-26032379 on Build GRCh38
rs753022509 Chr.18: 26032581-26032581 on Build GRCh38
rs1047036888 Chr.18: 26032805-26032805 on Build GRCh38
rs901225886 Chr.18: 26032771-26032771 on Build GRCh38
rs934073948 Chr.18: 26032773-26032773 on Build GRCh38
rs767854254 Chr.18: 26032433-26032433 on Build GRCh38
rs761773975 Chr.18: 26032748-26032749 on Build GRCh38
rs969525809 Chr.18: 26032561-26032561 on Build GRCh38
rs755200495 Chr.18: 26032466-26032466 on Build GRCh38
rs758514205 Chr.18: 26032475-26032475 on Build GRCh38
rs774768749 Chr.18: 26032687-26032687 on Build GRCh38
rs981377969 Chr.18: 26032425-26032425 on Build GRCh38
rs1017224835 Chr.18: 26032552-26032552 on Build GRCh38
rs755077606 Chr.18: 26032350-26032350 on Build GRCh38
rs780974644 Chr.18: 26032483-26032483 on Build GRCh38
rs900002415 Chr.18: 26032706-26032706 on Build GRCh38
rs138978468 Chr.18: 26032610-26032610 on Build GRCh38
rs931326105 Chr.18: 26032597-26032597 on Build GRCh38
rs997003501 Chr.18: 26032715-26032715 on Build GRCh38
rs763528676 Chr.18: 26032511-26032511 on Build GRCh38
rs766806675 Chr.18: 26032455-26032455 on Build GRCh38
rs879246927 Chr.18: 26032329-26032329 on Build GRCh38
rs957138351 Chr.18: 26032744-26032744 on Build GRCh38
rs943203972 Chr.18: 26032727-26032727 on Build GRCh38
rs553840946 Chr.18: 26032801-26032801 on Build GRCh38
rs1008931281 Chr.18: 26032637-26032638 on Build GRCh38
rs777479507 Chr.18: 26032571-26032572 on Build GRCh38
rs752271134 Chr.18: 26032552-26032553 on Build GRCh38
rs745862153 Chr.18: 26032489-26032489 on Build GRCh38
rs913576326 Chr.18: 26032621-26032621 on Build GRCh38
rs765493479 Chr.18: 26032578-26032578 on Build GRCh38
rs77644251 Chr.18: 26032435-26032435 on Build GRCh38

View Full Product Details