Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4292188-4292683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMIGD2
Entrez Gene ID
126259
Gene Name
transmembrane and immunoglobulin domain containing 2
Gene Aliases
CD28H, IGPR-1, IGPR1
UniGene
Hs.263928
Gene Chromosome Location
Chr.19: 4292227-4302439 on Build GRCh38
Transcripts
NM_001169126.1 NM_001308232.1 NM_144615.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5543547 c.774G>A p.M258I Chr.19: 4292674-4292674 on Build GRCh38
3823245 c.836T>G p.V279G Chr.19: 4292612-4292612 on Build GRCh38
6404452 c.778A>G p.R260G Chr.19: 4292670-4292670 on Build GRCh38
6882259 c.767T>A p.V256D Chr.19: 4292681-4292681 on Build GRCh38
3287823 c.766G>A p.V256I Chr.19: 4292682-4292682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762446672 Chr.19: 4292553-4292553 on Build GRCh38
rs866325366 Chr.19: 4292372-4292372 on Build GRCh38
rs1001048562 Chr.19: 4292234-4292234 on Build GRCh38
rs754451058 Chr.19: 4292621-4292621 on Build GRCh38
rs927308320 Chr.19: 4292641-4292641 on Build GRCh38
rs936810024 Chr.19: 4292422-4292422 on Build GRCh38
rs1053311361 Chr.19: 4292413-4292413 on Build GRCh38
rs913459916 Chr.19: 4292425-4292425 on Build GRCh38
rs888327915 Chr.19: 4292510-4292510 on Build GRCh38
rs781460284 Chr.19: 4292524-4292524 on Build GRCh38
rs983990426 Chr.19: 4292330-4292330 on Build GRCh38
rs547076686 Chr.19: 4292595-4292595 on Build GRCh38
rs994114378 Chr.19: 4292304-4292304 on Build GRCh38
rs892195746 Chr.19: 4292427-4292427 on Build GRCh38
rs759229071 Chr.19: 4292571-4292571 on Build GRCh38
rs745975486 Chr.19: 4292635-4292635 on Build GRCh38
rs918008423 Chr.19: 4292545-4292545 on Build GRCh38
rs148323916 Chr.19: 4292212-4292212 on Build GRCh38
rs564860685 Chr.19: 4292539-4292539 on Build GRCh38
rs965278835 Chr.19: 4292278-4292278 on Build GRCh38
rs765950132 Chr.19: 4292516-4292516 on Build GRCh38
rs183765076 Chr.19: 4292381-4292381 on Build GRCh38
rs940816115 Chr.19: 4292360-4292360 on Build GRCh38
rs372626161 Chr.19: 4292542-4292542 on Build GRCh38
rs747831163 Chr.19: 4292638-4292638 on Build GRCh38
rs57181230 Chr.19: 4292483-4292483 on Build GRCh38
rs781042986 Chr.19: 4292351-4292351 on Build GRCh38
rs980755638 Chr.19: 4292570-4292570 on Build GRCh38
rs375170109 Chr.19: 4292558-4292558 on Build GRCh38
rs937329613 Chr.19: 4292642-4292642 on Build GRCh38
rs910984821 Chr.19: 4292341-4292341 on Build GRCh38
rs375143619 Chr.19: 4292312-4292312 on Build GRCh38
rs952376799 Chr.19: 4292323-4292323 on Build GRCh38
rs1025569681 Chr.19: 4292490-4292490 on Build GRCh38
rs191819438 Chr.19: 4292512-4292512 on Build GRCh38
rs769816708 Chr.19: 4292488-4292488 on Build GRCh38
rs755323981 Chr.19: 4292517-4292517 on Build GRCh38
rs917956989 Chr.19: 4292379-4292379 on Build GRCh38
rs544151147 Chr.19: 4292552-4292552 on Build GRCh38
rs746139243 Chr.19: 4292541-4292541 on Build GRCh38
rs751193393 Chr.19: 4292564-4292564 on Build GRCh38
rs975364538 Chr.19: 4292280-4292280 on Build GRCh38
rs144403458 Chr.19: 4292636-4292636 on Build GRCh38
rs758313911 Chr.19: 4292598-4292601 on Build GRCh38
rs961245305 Chr.19: 4292459-4292459 on Build GRCh38
rs972061997 Chr.19: 4292480-4292480 on Build GRCh38
rs754923565 Chr.19: 4292305-4292305 on Build GRCh38
rs187046549 Chr.19: 4292428-4292428 on Build GRCh38
rs952627335 Chr.19: 4292322-4292322 on Build GRCh38
rs776247956 Chr.19: 4292543-4292543 on Build GRCh38
rs905854673 Chr.19: 4292449-4292449 on Build GRCh38
rs146297078 Chr.19: 4292436-4292436 on Build GRCh38
rs778134043 Chr.19: 4292614-4292614 on Build GRCh38
rs996530399 Chr.19: 4292476-4292476 on Build GRCh38
rs188609479 Chr.19: 4292247-4292247 on Build GRCh38
rs764978222 Chr.19: 4292525-4292545 on Build GRCh38
rs767233750 Chr.19: 4292576-4292576 on Build GRCh38
rs150052833 Chr.19: 4292624-4292624 on Build GRCh38
rs905901626 Chr.19: 4292262-4292262 on Build GRCh38
rs757387712 Chr.19: 4292622-4292622 on Build GRCh38
rs947660484 Chr.19: 4292610-4292610 on Build GRCh38
rs748660471 Chr.19: 4292534-4292534 on Build GRCh38
rs769697450 Chr.19: 4292640-4292640 on Build GRCh38
rs538603568 Chr.19: 4292245-4292245 on Build GRCh38
rs933476145 Chr.19: 4292411-4292411 on Build GRCh38
rs6510800 Chr.19: 4292365-4292365 on Build GRCh38
rs776909037 Chr.19: 4292225-4292225 on Build GRCh38
rs959856921 Chr.19: 4292286-4292286 on Build GRCh38
rs139426380 Chr.19: 4292460-4292460 on Build GRCh38
rs1001383505 Chr.19: 4292275-4292275 on Build GRCh38
rs1053980306 Chr.19: 4292423-4292423 on Build GRCh38
rs559542387 Chr.19: 4292569-4292570 on Build GRCh38
rs748937434 Chr.19: 4292544-4292544 on Build GRCh38
rs765025905 Chr.19: 4292546-4292546 on Build GRCh38
rs892232790 Chr.19: 4292256-4292256 on Build GRCh38
rs572970606 Chr.19: 4292489-4292489 on Build GRCh38
rs778369732 Chr.19: 4292538-4292538 on Build GRCh38
rs905457580 Chr.19: 4292399-4292399 on Build GRCh38
rs866612319 Chr.19: 4292471-4292471 on Build GRCh38
rs369334813 Chr.19: 4292656-4292656 on Build GRCh38
rs923466781 Chr.19: 4292380-4292380 on Build GRCh38
rs984154525 Chr.19: 4292327-4292327 on Build GRCh38
rs1045256271 Chr.19: 4292391-4292391 on Build GRCh38
rs192962526 Chr.19: 4292277-4292277 on Build GRCh38
rs888932 Chr.19: 4292600-4292600 on Build GRCh38
rs1009627896 Chr.19: 4292273-4292273 on Build GRCh38
rs200584828 Chr.19: 4292584-4292584 on Build GRCh38

View Full Product Details