Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11805928-11806430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF491
Entrez Gene ID
126069
Gene Name
zinc finger protein 491
Gene Aliases
-
UniGene
Hs.631634
Gene Chromosome Location
Chr.19: 11798509-11809622 on Build GRCh38
Transcripts
NM_152356.3 XM_005259730.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
258468 c.300G>T p.E100D Chr.19: 11806253-11806253 on Build GRCh38
6476591 c.323G>T p.C108F Chr.19: 11806276-11806276 on Build GRCh38
2149625 c.239G>A p.R80H Chr.19: 11806192-11806192 on Build GRCh38
4442060 c.261C>A p.S87R Chr.19: 11806214-11806214 on Build GRCh38
2814846 c.65C>T p.P22L Chr.19: 11806018-11806018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373373297 Chr.19: 11806315-11806315 on Build GRCh38
rs1032225800 Chr.19: 11806006-11806006 on Build GRCh38
rs373842039 Chr.19: 11806018-11806018 on Build GRCh38
rs762390668 Chr.19: 11806144-11806154 on Build GRCh38
rs375847840 Chr.19: 11806351-11806351 on Build GRCh38
rs780333499 Chr.19: 11806399-11806399 on Build GRCh38
rs61736273 Chr.19: 11806049-11806049 on Build GRCh38
rs144399257 Chr.19: 11806309-11806309 on Build GRCh38
rs773603988 Chr.19: 11806062-11806062 on Build GRCh38
rs564679078 Chr.19: 11806052-11806052 on Build GRCh38
rs139676501 Chr.19: 11806257-11806257 on Build GRCh38
rs755884504 Chr.19: 11806014-11806014 on Build GRCh38
rs376441208 Chr.19: 11805989-11805989 on Build GRCh38
rs759537385 Chr.19: 11806092-11806092 on Build GRCh38
rs751684697 Chr.19: 11805968-11805968 on Build GRCh38
rs373050538 Chr.19: 11806102-11806102 on Build GRCh38
rs367695952 Chr.19: 11806019-11806019 on Build GRCh38
rs745921472 Chr.19: 11806195-11806195 on Build GRCh38
rs773440550 Chr.19: 11805957-11805957 on Build GRCh38
rs202016817 Chr.19: 11806121-11806121 on Build GRCh38
rs111570520 Chr.19: 11806360-11806360 on Build GRCh38
rs150644754 Chr.19: 11806242-11806242 on Build GRCh38
rs892921069 Chr.19: 11806093-11806093 on Build GRCh38
rs141870350 Chr.19: 11806099-11806099 on Build GRCh38
rs990740097 Chr.19: 11806234-11806234 on Build GRCh38
rs552135465 Chr.19: 11806249-11806249 on Build GRCh38
rs760518312 Chr.19: 11806215-11806215 on Build GRCh38
rs187146401 Chr.19: 11806125-11806125 on Build GRCh38
rs772015572 Chr.19: 11806201-11806201 on Build GRCh38
rs55951313 Chr.19: 11806165-11806165 on Build GRCh38
rs758106680 Chr.19: 11806244-11806244 on Build GRCh38
rs998461869 Chr.19: 11806337-11806337 on Build GRCh38
rs200219612 Chr.19: 11806224-11806224 on Build GRCh38
rs766412552 Chr.19: 11805965-11805965 on Build GRCh38
rs764137663 Chr.19: 11806188-11806188 on Build GRCh38
rs752332928 Chr.19: 11806250-11806250 on Build GRCh38
rs778256006 Chr.19: 11806261-11806261 on Build GRCh38
rs764938776 Chr.19: 11806231-11806231 on Build GRCh38
rs370911780 Chr.19: 11806013-11806013 on Build GRCh38
rs145101992 Chr.19: 11806294-11806294 on Build GRCh38
rs1009791827 Chr.19: 11806035-11806035 on Build GRCh38
rs375933257 Chr.19: 11806192-11806192 on Build GRCh38
rs777511243 Chr.19: 11806351-11806352 on Build GRCh38
rs761664949 Chr.19: 11806316-11806316 on Build GRCh38
rs139779307 Chr.19: 11806308-11806308 on Build GRCh38
rs150364675 Chr.19: 11806216-11806216 on Build GRCh38
rs369000509 Chr.19: 11806245-11806245 on Build GRCh38
rs771199541 Chr.19: 11806080-11806080 on Build GRCh38
rs763961665 Chr.19: 11806138-11806138 on Build GRCh38
rs756690835 Chr.19: 11806185-11806185 on Build GRCh38
rs140117479 Chr.19: 11806130-11806130 on Build GRCh38
rs758254455 Chr.19: 11806265-11806267 on Build GRCh38
rs369776367 Chr.19: 11806172-11806172 on Build GRCh38
rs754444202 Chr.19: 11806190-11806191 on Build GRCh38
rs778715617 Chr.19: 11806152-11806152 on Build GRCh38
rs764812480 Chr.19: 11806192-11806193 on Build GRCh38
rs61736266 Chr.19: 11806075-11806075 on Build GRCh38
rs771073180 Chr.19: 11806191-11806191 on Build GRCh38
rs910099462 Chr.19: 11806162-11806162 on Build GRCh38
rs961355559 Chr.19: 11806394-11806394 on Build GRCh38
rs550928147 Chr.19: 11806171-11806171 on Build GRCh38
rs367903717 Chr.19: 11806083-11806083 on Build GRCh38
rs370864375 Chr.19: 11806205-11806205 on Build GRCh38
rs146593862 Chr.19: 11806318-11806318 on Build GRCh38
rs905954543 Chr.19: 11806225-11806225 on Build GRCh38
rs746057286 Chr.19: 11806300-11806300 on Build GRCh38
rs753746417 Chr.19: 11806145-11806145 on Build GRCh38
rs762977499 Chr.19: 11805959-11805959 on Build GRCh38
rs757021144 Chr.19: 11806026-11806026 on Build GRCh38
rs370609725 Chr.19: 11806324-11806324 on Build GRCh38
rs964952574 Chr.19: 11806165-11806165 on Build GRCh38
rs112159359 Chr.19: 11806404-11806404 on Build GRCh38
rs147204893 Chr.19: 11805984-11805984 on Build GRCh38
rs766119539 Chr.19: 11806325-11806325 on Build GRCh38
rs909291937 Chr.19: 11806209-11806209 on Build GRCh38
rs769143597 Chr.19: 11806046-11806046 on Build GRCh38
rs199900139 Chr.19: 11806173-11806173 on Build GRCh38
rs983730667 Chr.19: 11806369-11806369 on Build GRCh38
rs770082337 Chr.19: 11806058-11806058 on Build GRCh38
rs750965752 Chr.19: 11806329-11806329 on Build GRCh38
rs753321510 Chr.19: 11806186-11806187 on Build GRCh38
rs901004820 Chr.19: 11806311-11806311 on Build GRCh38
rs749894950 Chr.19: 11806169-11806169 on Build GRCh38
rs1016570625 Chr.19: 11806383-11806383 on Build GRCh38
rs778556059 Chr.19: 11806027-11806027 on Build GRCh38
rs953707050 Chr.19: 11806112-11806112 on Build GRCh38
rs1027996364 Chr.19: 11806354-11806354 on Build GRCh38
rs376445091 Chr.19: 11806147-11806147 on Build GRCh38
rs768021239 Chr.19: 11806147-11806148 on Build GRCh38
rs758480930 Chr.19: 11806396-11806396 on Build GRCh38
rs761717969 Chr.19: 11806230-11806230 on Build GRCh38
rs149778854 Chr.19: 11806278-11806278 on Build GRCh38
rs546146723 Chr.19: 11806045-11806045 on Build GRCh38
rs748492769 Chr.19: 11806056-11806056 on Build GRCh38
rs775041321 Chr.19: 11806015-11806015 on Build GRCh38
rs376696358 Chr.19: 11806385-11806385 on Build GRCh38
rs768660008 Chr.19: 11806312-11806312 on Build GRCh38
rs779132232 Chr.19: 11806370-11806370 on Build GRCh38

View Full Product Details