Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39980379-39980883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC4
Entrez Gene ID
5704
Gene Name
proteasome 26S subunit, ATPase 4
Gene Aliases
MIP224, RPT3, S6, TBP-7, TBP7
UniGene
Hs.211594
Gene Chromosome Location
Chr.19: 39971005-39981764 on Build GRCh38
Transcripts
NM_006503.3 NM_153001.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
996511 c.1107G>T p.K369N Chr.19: 39980681-39980681 on Build GRCh38
3533962 c.1013G>A p.R338H Chr.19: 39980380-39980380 on Build GRCh38
6757761 c.1097G>A p.R366Q Chr.19: 39980671-39980671 on Build GRCh38
3378935 c.1016G>A p.R339H Chr.19: 39980383-39980383 on Build GRCh38
4078106 c.1123A>G p.I375V Chr.19: 39980697-39980697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922982561 Chr.19: 39980820-39980820 on Build GRCh38
rs747867144 Chr.19: 39980429-39980429 on Build GRCh38
rs543898500 Chr.19: 39980548-39980548 on Build GRCh38
rs994378295 Chr.19: 39980567-39980567 on Build GRCh38
rs370960680 Chr.19: 39980606-39980606 on Build GRCh38
rs370164553 Chr.19: 39980654-39980654 on Build GRCh38
rs762211596 Chr.19: 39980724-39980724 on Build GRCh38
rs370201221 Chr.19: 39980487-39980487 on Build GRCh38
rs754407940 Chr.19: 39980502-39980502 on Build GRCh38
rs771058353 Chr.19: 39980466-39980466 on Build GRCh38
rs935028710 Chr.19: 39980743-39980743 on Build GRCh38
rs199789009 Chr.19: 39980475-39980475 on Build GRCh38
rs537338646 Chr.19: 39980795-39980796 on Build GRCh38
rs777706805 Chr.19: 39980617-39980617 on Build GRCh38
rs1127090 Chr.19: 39980699-39980699 on Build GRCh38
rs898643847 Chr.19: 39980566-39980566 on Build GRCh38
rs377606492 Chr.19: 39980496-39980496 on Build GRCh38
rs577380773 Chr.19: 39980610-39980610 on Build GRCh38
rs776963907 Chr.19: 39980723-39980723 on Build GRCh38
rs755796252 Chr.19: 39980614-39980614 on Build GRCh38
rs762917882 Chr.19: 39980454-39980454 on Build GRCh38
rs373501281 Chr.19: 39980486-39980486 on Build GRCh38
rs771995511 Chr.19: 39980677-39980677 on Build GRCh38
rs750884187 Chr.19: 39980741-39980741 on Build GRCh38
rs932982401 Chr.19: 39980507-39980507 on Build GRCh38
rs1047171651 Chr.19: 39980572-39980572 on Build GRCh38
rs376616888 Chr.19: 39980750-39980750 on Build GRCh38
rs752820559 Chr.19: 39980482-39980482 on Build GRCh38
rs541193657 Chr.19: 39980612-39980612 on Build GRCh38
rs530558521 Chr.19: 39980557-39980557 on Build GRCh38
rs752262509 Chr.19: 39980508-39980508 on Build GRCh38
rs370953179 Chr.19: 39980505-39980505 on Build GRCh38
rs956737059 Chr.19: 39980782-39980782 on Build GRCh38
rs1050052235 Chr.19: 39980518-39980518 on Build GRCh38
rs759585971 Chr.19: 39980474-39980474 on Build GRCh38
rs145486997 Chr.19: 39980484-39980484 on Build GRCh38
rs986620384 Chr.19: 39980591-39980591 on Build GRCh38
rs745847777 Chr.19: 39980667-39980667 on Build GRCh38
rs954798717 Chr.19: 39980585-39980586 on Build GRCh38
rs778186660 Chr.19: 39980799-39980799 on Build GRCh38
rs774469012 Chr.19: 39980657-39980657 on Build GRCh38
rs749155602 Chr.19: 39980639-39980639 on Build GRCh38
rs911439270 Chr.19: 39980520-39980520 on Build GRCh38
rs746262816 Chr.19: 39980793-39980793 on Build GRCh38
rs773158269 Chr.19: 39980435-39980435 on Build GRCh38
rs765788323 Chr.19: 39980497-39980497 on Build GRCh38
rs765624734 Chr.19: 39980738-39980738 on Build GRCh38
rs760738061 Chr.19: 39980697-39980697 on Build GRCh38
rs201585070 Chr.19: 39980745-39980745 on Build GRCh38
rs571180845 Chr.19: 39980555-39980555 on Build GRCh38
rs749062570 Chr.19: 39980768-39980768 on Build GRCh38
rs563289926 Chr.19: 39980821-39980821 on Build GRCh38
rs1013555443 Chr.19: 39980655-39980655 on Build GRCh38
rs924908320 Chr.19: 39980732-39980732 on Build GRCh38
rs771569020 Chr.19: 39980744-39980744 on Build GRCh38
rs755708927 Chr.19: 39980761-39980761 on Build GRCh38
rs942592059 Chr.19: 39980678-39980678 on Build GRCh38
rs777406855 Chr.19: 39980680-39980680 on Build GRCh38
rs758938422 Chr.19: 39980782-39980783 on Build GRCh38
rs201100542 Chr.19: 39980751-39980751 on Build GRCh38
rs376905637 Chr.19: 39980430-39980430 on Build GRCh38
rs780750036 Chr.19: 39980405-39980405 on Build GRCh38
rs73545950 Chr.19: 39980558-39980558 on Build GRCh38
rs774561164 Chr.19: 39980471-39980471 on Build GRCh38
rs751957256 Chr.19: 39980855-39980856 on Build GRCh38
rs112260376 Chr.19: 39980827-39980827 on Build GRCh38
rs777619101 Chr.19: 39980766-39980766 on Build GRCh38
rs751042109 Chr.19: 39980499-39980499 on Build GRCh38

View Full Product Details