Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44217554-44218091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF227
Entrez Gene ID
7770
Gene Name
zinc finger protein 227
Gene Aliases
-
UniGene
Hs.298089
Gene Chromosome Location
Chr.19: 44207527-44237268 on Build GRCh38
Transcripts
NM_001289166.1 NM_001289167.1 NM_001289168.1 NM_001289169.1 NM_001289170.1 NM_001289171.1 NM_001289172.1 NM_001289173.1 NM_182490.2 XM_005259232.2 XM_006723371.1 XM_011527289.2 XM_011527290.1 XM_017027266.1 XM_017027267.1 XM_017027268.1 XM_017027269.1 XM_017027270.1 XM_017027271.1 XM_017027272.1 XM_017027273.1 XM_017027274.1 XM_017027275.1 XM_017027276.1 XM_017027277.1 XM_017027279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588987 c.6A>G p.P2P Chr.19: 44217798-44217798 on Build GRCh38
1612381 c.54G>C p.K18N Chr.19: 44217846-44217846 on Build GRCh38
6317639 c.60+1G>T p.? Chr.19: 44217853-44217853 on Build GRCh38
361204 c.26C>T p.P9L Chr.19: 44217818-44217818 on Build GRCh38
997857 c.42G>A p.E14E Chr.19: 44217834-44217834 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573845374 Chr.19: 44217736-44217736 on Build GRCh38
rs755558756 Chr.19: 44217764-44217764 on Build GRCh38
rs958812826 Chr.19: 44218020-44218020 on Build GRCh38
rs753388922 Chr.19: 44217784-44217784 on Build GRCh38
rs967379312 Chr.19: 44217788-44217788 on Build GRCh38
rs759434814 Chr.19: 44217759-44217759 on Build GRCh38
rs191327173 Chr.19: 44217737-44217737 on Build GRCh38
rs754496175 Chr.19: 44217769-44217777 on Build GRCh38
rs377128532 Chr.19: 44217951-44217951 on Build GRCh38
rs35459284 Chr.19: 44217615-44217616 on Build GRCh38
rs894854950 Chr.19: 44218016-44218016 on Build GRCh38
rs746311059 Chr.19: 44217599-44217599 on Build GRCh38
rs772681862 Chr.19: 44217605-44217605 on Build GRCh38
rs141285343 Chr.19: 44217826-44217826 on Build GRCh38
rs778601307 Chr.19: 44218026-44218027 on Build GRCh38
rs138167873 Chr.19: 44217950-44217951 on Build GRCh38
rs1020257697 Chr.19: 44217712-44217712 on Build GRCh38
rs761412870 Chr.19: 44217882-44217882 on Build GRCh38
rs763544516 Chr.19: 44217868-44217868 on Build GRCh38
rs552615043 Chr.19: 44218052-44218052 on Build GRCh38
rs775336910 Chr.19: 44217863-44217863 on Build GRCh38
rs772201856 Chr.19: 44217680-44217680 on Build GRCh38
rs1026778832 Chr.19: 44218029-44218029 on Build GRCh38
rs763738125 Chr.19: 44217773-44217773 on Build GRCh38
rs139871578 Chr.19: 44217807-44217807 on Build GRCh38
rs759358804 Chr.19: 44217856-44217856 on Build GRCh38
rs759523314 Chr.19: 44217958-44217958 on Build GRCh38
rs991326138 Chr.19: 44217824-44217824 on Build GRCh38
rs908483176 Chr.19: 44218060-44218060 on Build GRCh38
rs867824196 Chr.19: 44217794-44217794 on Build GRCh38
rs969338646 Chr.19: 44217709-44217709 on Build GRCh38
rs767441279 Chr.19: 44217760-44217760 on Build GRCh38
rs779504160 Chr.19: 44217596-44217596 on Build GRCh38
rs202201777 Chr.19: 44217833-44217833 on Build GRCh38
rs773423584 Chr.19: 44217707-44217707 on Build GRCh38
rs200848109 Chr.19: 44217814-44217814 on Build GRCh38
rs535079341 Chr.19: 44217630-44217630 on Build GRCh38
rs190402067 Chr.19: 44218051-44218051 on Build GRCh38
rs780651036 Chr.19: 44217813-44217813 on Build GRCh38
rs570300729 Chr.19: 44217984-44217984 on Build GRCh38
rs186745834 Chr.19: 44217975-44217975 on Build GRCh38
rs774085128 Chr.19: 44217840-44217840 on Build GRCh38
rs932420017 Chr.19: 44217936-44217936 on Build GRCh38
rs774178625 Chr.19: 44217756-44217756 on Build GRCh38
rs143075857 Chr.19: 44217715-44217715 on Build GRCh38
rs757492690 Chr.19: 44217591-44217591 on Build GRCh38
rs747073871 Chr.19: 44217616-44217616 on Build GRCh38
rs183858428 Chr.19: 44217761-44217761 on Build GRCh38
rs151172615 Chr.19: 44217799-44217799 on Build GRCh38
rs375660767 Chr.19: 44217873-44217873 on Build GRCh38
rs764937893 Chr.19: 44217892-44217892 on Build GRCh38
rs906356653 Chr.19: 44217623-44217623 on Build GRCh38
rs189094722 Chr.19: 44217608-44217608 on Build GRCh38
rs765129435 Chr.19: 44218015-44218015 on Build GRCh38
rs772006979 Chr.19: 44217857-44217857 on Build GRCh38
rs989523887 Chr.19: 44217638-44217638 on Build GRCh38
rs769015554 Chr.19: 44217620-44217620 on Build GRCh38
rs982664191 Chr.19: 44217877-44217878 on Build GRCh38
rs181290314 Chr.19: 44217945-44217945 on Build GRCh38
rs977702125 Chr.19: 44217716-44217716 on Build GRCh38
rs921003100 Chr.19: 44217935-44217935 on Build GRCh38
rs7254458 Chr.19: 44217906-44217906 on Build GRCh38
rs770029398 Chr.19: 44217835-44217835 on Build GRCh38
rs766043166 Chr.19: 44217635-44217635 on Build GRCh38
rs188525370 Chr.19: 44217866-44217866 on Build GRCh38
rs750767093 Chr.19: 44217903-44217903 on Build GRCh38
rs200101180 Chr.19: 44217832-44217832 on Build GRCh38
rs1036130863 Chr.19: 44218018-44218018 on Build GRCh38
rs1002858332 Chr.19: 44218017-44218018 on Build GRCh38
rs760575272 Chr.19: 44217963-44217964 on Build GRCh38
rs1036936225 Chr.19: 44217942-44217942 on Build GRCh38
rs1042005586 Chr.19: 44217968-44217968 on Build GRCh38
rs547250618 Chr.19: 44217576-44217576 on Build GRCh38
rs757661352 Chr.19: 44217785-44217785 on Build GRCh38
rs768817642 Chr.19: 44217816-44217816 on Build GRCh38
rs116380402 Chr.19: 44217804-44217804 on Build GRCh38
rs1041166817 Chr.19: 44217922-44217922 on Build GRCh38
rs781500103 Chr.19: 44217819-44217819 on Build GRCh38
rs908181371 Chr.19: 44217894-44217894 on Build GRCh38

View Full Product Details