Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44800180-44800670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLC
Entrez Gene ID
23624
Gene Name
Cbl proto-oncogene C
Gene Aliases
CBL-3, CBL-SL, RNF57
UniGene
Hs.466907
Gene Chromosome Location
Chr.19: 44777855-44800646 on Build GRCh38
Transcripts
NM_001130852.1 NM_012116.3 XM_011526688.2 XM_011526689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620920 c.1394C>T p.A465V Chr.19: 44800412-44800412 on Build GRCh38
5815759 c.1406A>T p.Q469L Chr.19: 44800424-44800424 on Build GRCh38
6085373 c.1391C>G p.A464G Chr.19: 44800409-44800409 on Build GRCh38
5815758 c.1406A>T p.Q469L Chr.19: 44800424-44800424 on Build GRCh38
3103396 c.1400G>T p.G467V Chr.19: 44800418-44800418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775629187 Chr.19: 44800442-44800442 on Build GRCh38
rs771571423 Chr.19: 44800236-44800236 on Build GRCh38
rs752358314 Chr.19: 44800485-44800485 on Build GRCh38
rs781728391 Chr.19: 44800401-44800401 on Build GRCh38
rs983026382 Chr.19: 44800250-44800250 on Build GRCh38
rs776401872 Chr.19: 44800354-44800354 on Build GRCh38
rs754656011 Chr.19: 44800398-44800398 on Build GRCh38
rs761598020 Chr.19: 44800444-44800444 on Build GRCh38
rs187556678 Chr.19: 44800204-44800204 on Build GRCh38
rs916654543 Chr.19: 44800268-44800268 on Build GRCh38
rs201244009 Chr.19: 44800478-44800487 on Build GRCh38
rs1011305311 Chr.19: 44800413-44800413 on Build GRCh38
rs927041508 Chr.19: 44800331-44800331 on Build GRCh38
rs770969313 Chr.19: 44800426-44800426 on Build GRCh38
rs753128512 Chr.19: 44800403-44800403 on Build GRCh38
rs922756154 Chr.19: 44800237-44800237 on Build GRCh38
rs61750956 Chr.19: 44800382-44800382 on Build GRCh38
rs907571088 Chr.19: 44800300-44800300 on Build GRCh38
rs765170387 Chr.19: 44800366-44800366 on Build GRCh38
rs769537341 Chr.19: 44800454-44800454 on Build GRCh38
rs73572010 Chr.19: 44800437-44800437 on Build GRCh38
rs940730497 Chr.19: 44800578-44800578 on Build GRCh38
rs150486523 Chr.19: 44800608-44800608 on Build GRCh38
rs754467378 Chr.19: 44800371-44800371 on Build GRCh38
rs761472600 Chr.19: 44800577-44800577 on Build GRCh38
rs201089304 Chr.19: 44800339-44800339 on Build GRCh38
rs779464880 Chr.19: 44800395-44800403 on Build GRCh38
rs576435380 Chr.19: 44800446-44800446 on Build GRCh38
rs759707965 Chr.19: 44800276-44800276 on Build GRCh38
rs752951359 Chr.19: 44800468-44800468 on Build GRCh38
rs931886470 Chr.19: 44800594-44800594 on Build GRCh38
rs1050232407 Chr.19: 44800391-44800391 on Build GRCh38
rs765846775 Chr.19: 44800464-44800464 on Build GRCh38
rs758912772 Chr.19: 44800476-44800476 on Build GRCh38
rs756573147 Chr.19: 44800486-44800486 on Build GRCh38
rs200015142 Chr.19: 44800392-44800392 on Build GRCh38
rs902931578 Chr.19: 44800607-44800607 on Build GRCh38
rs991403675 Chr.19: 44800542-44800542 on Build GRCh38
rs572533436 Chr.19: 44800431-44800431 on Build GRCh38
rs1035328648 Chr.19: 44800625-44800625 on Build GRCh38
rs962918335 Chr.19: 44800532-44800532 on Build GRCh38
rs892990800 Chr.19: 44800598-44800598 on Build GRCh38
rs768379559 Chr.19: 44800344-44800344 on Build GRCh38
rs1041520350 Chr.19: 44800606-44800606 on Build GRCh38
rs568844963 Chr.19: 44800231-44800231 on Build GRCh38
rs1017208827 Chr.19: 44800530-44800530 on Build GRCh38
rs762400110 Chr.19: 44800385-44800385 on Build GRCh38
rs780639506 Chr.19: 44800492-44800501 on Build GRCh38
rs1040352749 Chr.19: 44800580-44800580 on Build GRCh38
rs774463936 Chr.19: 44800341-44800341 on Build GRCh38
rs373596478 Chr.19: 44800490-44800499 on Build GRCh38
rs749384545 Chr.19: 44800335-44800335 on Build GRCh38
rs976551980 Chr.19: 44800279-44800280 on Build GRCh38
rs537974510 Chr.19: 44800248-44800248 on Build GRCh38
rs749756713 Chr.19: 44800422-44800422 on Build GRCh38
rs973066408 Chr.19: 44800533-44800533 on Build GRCh38
rs746139762 Chr.19: 44800432-44800432 on Build GRCh38
rs370308752 Chr.19: 44800338-44800338 on Build GRCh38
rs756481545 Chr.19: 44800408-44800408 on Build GRCh38
rs984864837 Chr.19: 44800558-44800558 on Build GRCh38
rs777240869 Chr.19: 44800247-44800247 on Build GRCh38
rs964132189 Chr.19: 44800203-44800203 on Build GRCh38
rs767284522 Chr.19: 44800482-44800482 on Build GRCh38
rs773798441 Chr.19: 44800333-44800333 on Build GRCh38
rs904426529 Chr.19: 44800412-44800412 on Build GRCh38
rs1010137624 Chr.19: 44800377-44800377 on Build GRCh38
rs991000324 Chr.19: 44800262-44800262 on Build GRCh38
rs748913 Chr.19: 44800321-44800321 on Build GRCh38
rs778263593 Chr.19: 44800490-44800490 on Build GRCh38
rs565125133 Chr.19: 44800604-44800604 on Build GRCh38
rs751067514 Chr.19: 44800471-44800471 on Build GRCh38
rs917093481 Chr.19: 44800645-44800645 on Build GRCh38
rs376412498 Chr.19: 44800463-44800463 on Build GRCh38
rs923186221 Chr.19: 44800586-44800586 on Build GRCh38
rs951758708 Chr.19: 44800249-44800249 on Build GRCh38
rs961261053 Chr.19: 44800642-44800642 on Build GRCh38
rs141594020 Chr.19: 44800355-44800355 on Build GRCh38
rs373295347 Chr.19: 44800343-44800343 on Build GRCh38
rs773153802 Chr.19: 44800455-44800455 on Build GRCh38
rs952247176 Chr.19: 44800547-44800547 on Build GRCh38
rs983057339 Chr.19: 44800488-44800488 on Build GRCh38
rs796350918 Chr.19: 44800267-44800267 on Build GRCh38
rs755514112 Chr.19: 44800372-44800372 on Build GRCh38
rs193110123 Chr.19: 44800543-44800543 on Build GRCh38
rs941693771 Chr.19: 44800303-44800303 on Build GRCh38
rs765864698 Chr.19: 44800386-44800386 on Build GRCh38

View Full Product Details