Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50359658-50360167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPSA
Entrez Gene ID
9476
Gene Name
napsin A aspartic peptidase
Gene Aliases
KAP, Kdap, NAP1, NAPA, SNAPA
UniGene
Hs.512843
Gene Chromosome Location
Chr.19: 50358472-50369331 on Build GRCh38
Transcripts
NM_004851.2 XM_011527540.1 XM_017027512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2754425 c.767C>T p.A256V Chr.19: 50359764-50359764 on Build GRCh38
3757061 c.765T>C p.P255P Chr.19: 50359766-50359766 on Build GRCh38
1395434 c.675T>C p.P225P Chr.19: 50359856-50359856 on Build GRCh38
5525285 c.764C>T p.P255L Chr.19: 50359767-50359767 on Build GRCh38
336106 c.709G>A p.G237S Chr.19: 50359822-50359822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753918630 Chr.19: 50359868-50359868 on Build GRCh38
rs971456434 Chr.19: 50360043-50360043 on Build GRCh38
rs375232742 Chr.19: 50359848-50359848 on Build GRCh38
rs638294 Chr.19: 50359766-50359766 on Build GRCh38
rs528716175 Chr.19: 50359889-50359889 on Build GRCh38
rs372372402 Chr.19: 50359741-50359741 on Build GRCh38
rs773891178 Chr.19: 50360016-50360016 on Build GRCh38
rs749829491 Chr.19: 50360007-50360007 on Build GRCh38
rs634713 Chr.19: 50359928-50359928 on Build GRCh38
rs748131931 Chr.19: 50359727-50359727 on Build GRCh38
rs560915904 Chr.19: 50359887-50359887 on Build GRCh38
rs769339253 Chr.19: 50359913-50359913 on Build GRCh38
rs774523248 Chr.19: 50359820-50359821 on Build GRCh38
rs756132672 Chr.19: 50359800-50359800 on Build GRCh38
rs569170963 Chr.19: 50360095-50360095 on Build GRCh38
rs73932468 Chr.19: 50360023-50360023 on Build GRCh38
rs750124264 Chr.19: 50359840-50359840 on Build GRCh38
rs140131070 Chr.19: 50359772-50359772 on Build GRCh38
rs148925155 Chr.19: 50359812-50359812 on Build GRCh38
rs374700636 Chr.19: 50359809-50359809 on Build GRCh38
rs771942751 Chr.19: 50359816-50359816 on Build GRCh38
rs772647545 Chr.19: 50359861-50359861 on Build GRCh38
rs915355633 Chr.19: 50360135-50360135 on Build GRCh38
rs759389353 Chr.19: 50359751-50359751 on Build GRCh38
rs115708794 Chr.19: 50359817-50359817 on Build GRCh38
rs779968249 Chr.19: 50359882-50359882 on Build GRCh38
rs151042743 Chr.19: 50359820-50359820 on Build GRCh38
rs1029141197 Chr.19: 50360120-50360120 on Build GRCh38
rs781517212 Chr.19: 50359796-50359796 on Build GRCh38
rs144934721 Chr.19: 50359878-50359878 on Build GRCh38
rs369695611 Chr.19: 50359710-50359710 on Build GRCh38
rs954790181 Chr.19: 50360132-50360132 on Build GRCh38
rs200649376 Chr.19: 50359844-50359844 on Build GRCh38
rs989738713 Chr.19: 50360107-50360107 on Build GRCh38
rs764825762 Chr.19: 50359837-50359837 on Build GRCh38
rs539333263 Chr.19: 50359825-50359830 on Build GRCh38
rs1027419004 Chr.19: 50359990-50359990 on Build GRCh38
rs763697359 Chr.19: 50359679-50359679 on Build GRCh38
rs142041611 Chr.19: 50360018-50360018 on Build GRCh38
rs758116701 Chr.19: 50359881-50359881 on Build GRCh38
rs751816692 Chr.19: 50359787-50359787 on Build GRCh38
rs775870283 Chr.19: 50360057-50360057 on Build GRCh38
rs963476318 Chr.19: 50359836-50359836 on Build GRCh38
rs146758370 Chr.19: 50360080-50360080 on Build GRCh38
rs773704054 Chr.19: 50359880-50359881 on Build GRCh38
rs376453232 Chr.19: 50359744-50359744 on Build GRCh38
rs748819177 Chr.19: 50359783-50359783 on Build GRCh38
rs1043277141 Chr.19: 50359746-50359746 on Build GRCh38
rs778116703 Chr.19: 50359735-50359735 on Build GRCh38
rs771854245 Chr.19: 50359758-50359758 on Build GRCh38
rs761524390 Chr.19: 50359784-50359784 on Build GRCh38
rs201819380 Chr.19: 50359843-50359843 on Build GRCh38
rs916987252 Chr.19: 50359996-50359996 on Build GRCh38
rs778435688 Chr.19: 50359858-50359858 on Build GRCh38
rs867897359 Chr.19: 50360042-50360042 on Build GRCh38
rs551311595 Chr.19: 50359976-50359976 on Build GRCh38
rs755377702 Chr.19: 50359718-50359718 on Build GRCh38
rs143898038 Chr.19: 50359773-50359773 on Build GRCh38
rs753531339 Chr.19: 50359684-50359684 on Build GRCh38
rs756620162 Chr.19: 50359855-50359855 on Build GRCh38
rs769953944 Chr.19: 50359732-50359732 on Build GRCh38
rs1009015582 Chr.19: 50360138-50360139 on Build GRCh38
rs199991360 Chr.19: 50359875-50359875 on Build GRCh38
rs994659684 Chr.19: 50359767-50359767 on Build GRCh38
rs764878935 Chr.19: 50359785-50359785 on Build GRCh38
rs377299154 Chr.19: 50359811-50359811 on Build GRCh38
rs774222573 Chr.19: 50359750-50359750 on Build GRCh38
rs747514253 Chr.19: 50359716-50359716 on Build GRCh38
rs140973133 Chr.19: 50359821-50359821 on Build GRCh38
rs2305505 Chr.19: 50359977-50359977 on Build GRCh38
rs753293037 Chr.19: 50359797-50359797 on Build GRCh38
rs371222764 Chr.19: 50359803-50359803 on Build GRCh38
rs1055667378 Chr.19: 50359991-50359991 on Build GRCh38
rs759794042 Chr.19: 50359841-50359841 on Build GRCh38
rs1014834075 Chr.19: 50359737-50359737 on Build GRCh38
rs770554778 Chr.19: 50360004-50360004 on Build GRCh38
rs1031358770 Chr.19: 50360070-50360070 on Build GRCh38
rs547179258 Chr.19: 50359897-50359897 on Build GRCh38
rs758740532 Chr.19: 50359705-50359705 on Build GRCh38
rs760724883 Chr.19: 50359818-50359818 on Build GRCh38
rs370408939 Chr.19: 50360112-50360112 on Build GRCh38
rs925860859 Chr.19: 50359988-50359988 on Build GRCh38
rs146890854 Chr.19: 50359793-50359793 on Build GRCh38
rs774467450 Chr.19: 50360066-50360066 on Build GRCh38
rs757443248 Chr.19: 50359869-50359869 on Build GRCh38
rs776576866 Chr.19: 50359896-50359896 on Build GRCh38
rs978664781 Chr.19: 50359908-50359908 on Build GRCh38
rs749576537 Chr.19: 50359740-50359740 on Build GRCh38
rs781730739 Chr.19: 50359722-50359722 on Build GRCh38

View Full Product Details