Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57605596-57606104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF530
Entrez Gene ID
348327
Gene Name
zinc finger protein 530
Gene Aliases
-
UniGene
Hs.662177
Gene Chromosome Location
Chr.19: 57599877-57612722 on Build GRCh38
Transcripts
NM_001321981.1 NM_020880.4 NR_135923.1 NR_135924.1 XM_006723194.1 XM_011526919.2 XM_011526921.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143041 c.173G>C p.G58A Chr.19: 57605698-57605698 on Build GRCh38
2972778 c.374G>A p.R125K Chr.19: 57605899-57605899 on Build GRCh38
6208406 c.224T>C p.L75P Chr.19: 57605749-57605749 on Build GRCh38
4744154 c.535G>A p.A179T Chr.19: 57606060-57606060 on Build GRCh38
6394595 c.248C>A p.A83E Chr.19: 57605773-57605773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763259758 Chr.19: 57606011-57606011 on Build GRCh38
rs772431565 Chr.19: 57605967-57605967 on Build GRCh38
rs529420510 Chr.19: 57605732-57605732 on Build GRCh38
rs777013515 Chr.19: 57606063-57606063 on Build GRCh38
rs747308582 Chr.19: 57606059-57606059 on Build GRCh38
rs111675758 Chr.19: 57605746-57605746 on Build GRCh38
rs962300167 Chr.19: 57605975-57605975 on Build GRCh38
rs777683891 Chr.19: 57605717-57605717 on Build GRCh38
rs774857046 Chr.19: 57605772-57605772 on Build GRCh38
rs193202079 Chr.19: 57605982-57605982 on Build GRCh38
rs112840530 Chr.19: 57605902-57605902 on Build GRCh38
rs989637539 Chr.19: 57605697-57605697 on Build GRCh38
rs752967418 Chr.19: 57606010-57606010 on Build GRCh38
rs774737119 Chr.19: 57605797-57605797 on Build GRCh38
rs143267609 Chr.19: 57606044-57606044 on Build GRCh38
rs147794417 Chr.19: 57605901-57605901 on Build GRCh38
rs774684277 Chr.19: 57605664-57605664 on Build GRCh38
rs990059289 Chr.19: 57605993-57605993 on Build GRCh38
rs904270279 Chr.19: 57605869-57605869 on Build GRCh38
rs754096419 Chr.19: 57605703-57605703 on Build GRCh38
rs762197497 Chr.19: 57605812-57605812 on Build GRCh38
rs777975824 Chr.19: 57605672-57605676 on Build GRCh38
rs199647158 Chr.19: 57605648-57605648 on Build GRCh38
rs2360543 Chr.19: 57605855-57605855 on Build GRCh38
rs978040990 Chr.19: 57605950-57605950 on Build GRCh38
rs766074276 Chr.19: 57605839-57605839 on Build GRCh38
rs953162052 Chr.19: 57605922-57605922 on Build GRCh38
rs763114530 Chr.19: 57605854-57605854 on Build GRCh38
rs745738113 Chr.19: 57605733-57605733 on Build GRCh38
rs144915290 Chr.19: 57605783-57605783 on Build GRCh38
rs777403523 Chr.19: 57605874-57605874 on Build GRCh38
rs745880838 Chr.19: 57605863-57605863 on Build GRCh38
rs200925051 Chr.19: 57606042-57606042 on Build GRCh38
rs368341724 Chr.19: 57605656-57605656 on Build GRCh38
rs147119609 Chr.19: 57606050-57606050 on Build GRCh38
rs774016352 Chr.19: 57605939-57605939 on Build GRCh38
rs770062904 Chr.19: 57606072-57606072 on Build GRCh38
rs578006238 Chr.19: 57605699-57605699 on Build GRCh38
rs566890006 Chr.19: 57606032-57606032 on Build GRCh38
rs938603191 Chr.19: 57605780-57605780 on Build GRCh38
rs750163145 Chr.19: 57606024-57606024 on Build GRCh38
rs766551119 Chr.19: 57605821-57605821 on Build GRCh38
rs771614565 Chr.19: 57605791-57605792 on Build GRCh38
rs775695836 Chr.19: 57606074-57606074 on Build GRCh38
rs952718991 Chr.19: 57606006-57606006 on Build GRCh38
rs775218011 Chr.19: 57605652-57605652 on Build GRCh38
rs747742922 Chr.19: 57605949-57605949 on Build GRCh38
rs534331833 Chr.19: 57605655-57605655 on Build GRCh38
rs148196119 Chr.19: 57605969-57605969 on Build GRCh38
rs145898267 Chr.19: 57605857-57605857 on Build GRCh38
rs771290670 Chr.19: 57606061-57606061 on Build GRCh38
rs149878564 Chr.19: 57605779-57605779 on Build GRCh38
rs146709323 Chr.19: 57606005-57606005 on Build GRCh38
rs17855076 Chr.19: 57605895-57605895 on Build GRCh38
rs897045236 Chr.19: 57605799-57605799 on Build GRCh38
rs111634535 Chr.19: 57606045-57606045 on Build GRCh38
rs571846434 Chr.19: 57606001-57606001 on Build GRCh38
rs767476735 Chr.19: 57606017-57606017 on Build GRCh38
rs776815496 Chr.19: 57605992-57605992 on Build GRCh38
rs761306270 Chr.19: 57605985-57605985 on Build GRCh38
rs1026778909 Chr.19: 57605848-57605848 on Build GRCh38
rs9677004 Chr.19: 57605715-57605715 on Build GRCh38
rs768170721 Chr.19: 57605934-57605934 on Build GRCh38
rs765528225 Chr.19: 57605831-57605831 on Build GRCh38
rs756242579 Chr.19: 57605695-57605695 on Build GRCh38
rs755280328 Chr.19: 57605827-57605827 on Build GRCh38
rs755991314 Chr.19: 57605874-57605874 on Build GRCh38
rs115480393 Chr.19: 57606036-57606036 on Build GRCh38
rs371280063 Chr.19: 57605819-57605819 on Build GRCh38
rs968483491 Chr.19: 57605624-57605624 on Build GRCh38
rs531189449 Chr.19: 57606014-57606014 on Build GRCh38
rs767823307 Chr.19: 57605817-57605817 on Build GRCh38
rs151225498 Chr.19: 57605731-57605731 on Build GRCh38
rs188498612 Chr.19: 57605628-57605628 on Build GRCh38
rs931433966 Chr.19: 57605778-57605778 on Build GRCh38
rs753852953 Chr.19: 57605825-57605825 on Build GRCh38
rs779663871 Chr.19: 57605885-57605885 on Build GRCh38
rs750890623 Chr.19: 57605693-57605693 on Build GRCh38
rs760016676 Chr.19: 57606069-57606069 on Build GRCh38
rs767979429 Chr.19: 57605684-57605684 on Build GRCh38
rs1041994025 Chr.19: 57606071-57606071 on Build GRCh38
rs11883343 Chr.19: 57606031-57606031 on Build GRCh38
rs185138735 Chr.19: 57606058-57606058 on Build GRCh38
rs746781559 Chr.19: 57605724-57605724 on Build GRCh38
rs9676252 Chr.19: 57605654-57605654 on Build GRCh38
rs1019604612 Chr.19: 57605893-57605893 on Build GRCh38

View Full Product Details