Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24021554-24022044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD2B
Entrez Gene ID
388931
Gene Name
major facilitator superfamily domain containing 2B
Gene Aliases
-
UniGene
Hs.407482
Gene Chromosome Location
Chr.2: 24009022-24026775 on Build GRCh38
Transcripts
NM_001080473.1 XM_005264301.4 XM_017004063.1 XM_017004064.1 XM_017004065.1 XM_017004066.1 XM_017004067.1 XM_017004068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1406786 c.698T>C p.I233T Chr.2: 24021664-24021664 on Build GRCh38
4093098 c.847T>C p.Y283H Chr.2: 24021923-24021923 on Build GRCh38
4093100 c.859G>A p.V287M Chr.2: 24021935-24021935 on Build GRCh38
5658392 c.878T>C p.I293T Chr.2: 24021954-24021954 on Build GRCh38
1406788 c.753A>G p.L251L Chr.2: 24021719-24021719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745916974 Chr.2: 24021969-24021969 on Build GRCh38
rs756789096 Chr.2: 24021781-24021781 on Build GRCh38
rs947251289 Chr.2: 24021898-24021898 on Build GRCh38
rs202230526 Chr.2: 24021674-24021674 on Build GRCh38
rs764398621 Chr.2: 24021859-24021859 on Build GRCh38
rs772156990 Chr.2: 24021838-24021838 on Build GRCh38
rs368779478 Chr.2: 24021834-24021834 on Build GRCh38
rs942475768 Chr.2: 24021940-24021942 on Build GRCh38
rs915233311 Chr.2: 24021963-24021963 on Build GRCh38
rs555738056 Chr.2: 24021863-24021863 on Build GRCh38
rs368952967 Chr.2: 24021907-24021907 on Build GRCh38
rs745343369 Chr.2: 24021883-24021883 on Build GRCh38
rs553736501 Chr.2: 24021995-24021995 on Build GRCh38
rs771901950 Chr.2: 24021721-24021721 on Build GRCh38
rs747126152 Chr.2: 24021655-24021655 on Build GRCh38
rs532843606 Chr.2: 24021725-24021725 on Build GRCh38
rs193121648 Chr.2: 24021599-24021599 on Build GRCh38
rs766529144 Chr.2: 24021735-24021735 on Build GRCh38
rs748157251 Chr.2: 24021925-24021925 on Build GRCh38
rs955959473 Chr.2: 24021872-24021872 on Build GRCh38
rs775601550 Chr.2: 24021987-24021987 on Build GRCh38
rs951009603 Chr.2: 24021578-24021578 on Build GRCh38
rs970163497 Chr.2: 24021797-24021797 on Build GRCh38
rs770569916 Chr.2: 24021605-24021605 on Build GRCh38
rs777971493 Chr.2: 24021680-24021702 on Build GRCh38
rs746748488 Chr.2: 24021816-24021816 on Build GRCh38
rs767986355 Chr.2: 24021740-24021740 on Build GRCh38
rs367681797 Chr.2: 24021653-24021653 on Build GRCh38
rs769853015 Chr.2: 24021928-24021928 on Build GRCh38
rs751272669 Chr.2: 24022013-24022013 on Build GRCh38
rs938635850 Chr.2: 24021841-24021841 on Build GRCh38
rs760817247 Chr.2: 24021988-24021988 on Build GRCh38
rs764558486 Chr.2: 24021762-24021762 on Build GRCh38
rs201522941 Chr.2: 24021675-24021675 on Build GRCh38
rs778325478 Chr.2: 24021786-24021786 on Build GRCh38
rs185798712 Chr.2: 24021817-24021817 on Build GRCh38
rs760461163 Chr.2: 24021638-24021638 on Build GRCh38
rs768709238 Chr.2: 24021732-24021732 on Build GRCh38
rs753833077 Chr.2: 24021971-24021971 on Build GRCh38
rs764858377 Chr.2: 24021882-24021882 on Build GRCh38
rs748769226 Chr.2: 24021690-24021690 on Build GRCh38
rs115334231 Chr.2: 24021733-24021733 on Build GRCh38
rs200363363 Chr.2: 24021842-24021842 on Build GRCh38
rs754345361 Chr.2: 24021871-24021871 on Build GRCh38
rs745468231 Chr.2: 24021719-24021719 on Build GRCh38
rs763148373 Chr.2: 24021734-24021734 on Build GRCh38
rs762402340 Chr.2: 24022000-24022000 on Build GRCh38
rs756688792 Chr.2: 24021748-24021748 on Build GRCh38
rs112498523 Chr.2: 24021851-24021851 on Build GRCh38
rs779093340 Chr.2: 24021961-24021961 on Build GRCh38
rs779966231 Chr.2: 24021814-24021814 on Build GRCh38
rs759622870 Chr.2: 24021738-24021738 on Build GRCh38
rs756948505 Chr.2: 24021696-24021696 on Build GRCh38
rs375190597 Chr.2: 24021989-24021989 on Build GRCh38
rs571715253 Chr.2: 24021722-24021722 on Build GRCh38
rs771149993 Chr.2: 24021833-24021833 on Build GRCh38
rs142123482 Chr.2: 24021588-24021588 on Build GRCh38
rs755403213 Chr.2: 24021665-24021665 on Build GRCh38
rs745651631 Chr.2: 24021621-24021621 on Build GRCh38
rs955212752 Chr.2: 24021840-24021840 on Build GRCh38
rs887367366 Chr.2: 24021746-24021746 on Build GRCh38
rs765630372 Chr.2: 24021998-24021998 on Build GRCh38
rs975203595 Chr.2: 24021639-24021639 on Build GRCh38
rs199790975 Chr.2: 24021911-24021911 on Build GRCh38
rs1052394059 Chr.2: 24021694-24021694 on Build GRCh38
rs566363217 Chr.2: 24021745-24021745 on Build GRCh38
rs765848582 Chr.2: 24022010-24022010 on Build GRCh38
rs775525362 Chr.2: 24021633-24021633 on Build GRCh38
rs946801105 Chr.2: 24021723-24021723 on Build GRCh38
rs776965717 Chr.2: 24021992-24021992 on Build GRCh38
rs200261524 Chr.2: 24021906-24021906 on Build GRCh38
rs774167824 Chr.2: 24021610-24021610 on Build GRCh38
rs749894294 Chr.2: 24021886-24021886 on Build GRCh38
rs955415139 Chr.2: 24021654-24021654 on Build GRCh38
rs536416317 Chr.2: 24021812-24021812 on Build GRCh38
rs1053060977 Chr.2: 24021848-24021848 on Build GRCh38
rs777335703 Chr.2: 24021686-24021686 on Build GRCh38
rs762246600 Chr.2: 24021876-24021876 on Build GRCh38
rs767708664 Chr.2: 24021668-24021668 on Build GRCh38
rs774639664 Chr.2: 24021737-24021737 on Build GRCh38
rs751782427 Chr.2: 24021664-24021664 on Build GRCh38
rs754290263 Chr.2: 24021767-24021767 on Build GRCh38
rs990065313 Chr.2: 24021873-24021873 on Build GRCh38
rs759871444 Chr.2: 24021835-24021835 on Build GRCh38
rs1011803587 Chr.2: 24021617-24021617 on Build GRCh38
rs762371377 Chr.2: 24021993-24021993 on Build GRCh38
rs771711175 Chr.2: 24021625-24021625 on Build GRCh38
rs773363433 Chr.2: 24021692-24021692 on Build GRCh38
rs773412328 Chr.2: 24021647-24021647 on Build GRCh38
rs777211171 Chr.2: 24021598-24021598 on Build GRCh38
rs750101800 Chr.2: 24021807-24021807 on Build GRCh38
rs373395037 Chr.2: 24021912-24021912 on Build GRCh38
rs765369942 Chr.2: 24021641-24021641 on Build GRCh38
rs770063279 Chr.2: 24021628-24021629 on Build GRCh38
rs1003846180 Chr.2: 24021798-24021798 on Build GRCh38
rs375422477 Chr.2: 24021818-24021818 on Build GRCh38

View Full Product Details