Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69864803-69865351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMCL1
Entrez Gene ID
64395
Gene Name
germ cell-less, spermatogenesis associated 1
Gene Aliases
BTBD13, GCL, GCL1, SPATA29
UniGene
Hs.293971
Gene Chromosome Location
Chr.2: 69829606-69881396 on Build GRCh38
Transcripts
NM_178439.4 XM_011533033.1 XM_011533034.2 XM_017004704.1 XM_017004705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
126724 c.1174G>T p.G392* Chr.2: 69864931-69864931 on Build GRCh38
3972320 c.1143-1G>T p.? Chr.2: 69864899-69864899 on Build GRCh38
4523457 c.1216G>A p.E406K Chr.2: 69864973-69864973 on Build GRCh38
6671961 c.1190G>A p.C397Y Chr.2: 69864947-69864947 on Build GRCh38
5948908 c.1172A>G p.E391G Chr.2: 69864929-69864929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377022782 Chr.2: 69865013-69865013 on Build GRCh38
rs982988764 Chr.2: 69865083-69865083 on Build GRCh38
rs200583478 Chr.2: 69864851-69864851 on Build GRCh38
rs78263806 Chr.2: 69865241-69865241 on Build GRCh38
rs374737011 Chr.2: 69865239-69865239 on Build GRCh38
rs959780601 Chr.2: 69864999-69864999 on Build GRCh38
rs903035581 Chr.2: 69865254-69865254 on Build GRCh38
rs79102790 Chr.2: 69865214-69865214 on Build GRCh38
rs187800510 Chr.2: 69865142-69865142 on Build GRCh38
rs1041561383 Chr.2: 69865252-69865252 on Build GRCh38
rs753417518 Chr.2: 69864858-69864858 on Build GRCh38
rs765851779 Chr.2: 69864996-69864996 on Build GRCh38
rs868689141 Chr.2: 69864914-69864914 on Build GRCh38
rs992643175 Chr.2: 69865220-69865220 on Build GRCh38
rs926594484 Chr.2: 69865209-69865209 on Build GRCh38
rs747155646 Chr.2: 69864900-69864900 on Build GRCh38
rs200699097 Chr.2: 69864881-69864881 on Build GRCh38
rs183308985 Chr.2: 69865084-69865084 on Build GRCh38
rs753056347 Chr.2: 69864874-69864874 on Build GRCh38
rs373908725 Chr.2: 69865003-69865003 on Build GRCh38
rs974336240 Chr.2: 69865126-69865126 on Build GRCh38
rs962984911 Chr.2: 69865110-69865110 on Build GRCh38
rs753128153 Chr.2: 69865010-69865020 on Build GRCh38
rs35874751 Chr.2: 69864860-69864860 on Build GRCh38
rs759761499 Chr.2: 69864917-69864919 on Build GRCh38
rs781618097 Chr.2: 69864912-69864912 on Build GRCh38
rs201816300 Chr.2: 69865010-69865010 on Build GRCh38
rs960520022 Chr.2: 69865323-69865323 on Build GRCh38
rs1051566190 Chr.2: 69865094-69865094 on Build GRCh38
rs376128248 Chr.2: 69865020-69865020 on Build GRCh38
rs747916035 Chr.2: 69864939-69864939 on Build GRCh38
rs911752766 Chr.2: 69865135-69865135 on Build GRCh38
rs921816871 Chr.2: 69865069-69865069 on Build GRCh38
rs944127280 Chr.2: 69865145-69865145 on Build GRCh38
rs575540764 Chr.2: 69865050-69865050 on Build GRCh38
rs891829469 Chr.2: 69864892-69864892 on Build GRCh38
rs866559369 Chr.2: 69864973-69864973 on Build GRCh38
rs891777520 Chr.2: 69865238-69865238 on Build GRCh38
rs774034456 Chr.2: 69864932-69864932 on Build GRCh38
rs760234728 Chr.2: 69864971-69864971 on Build GRCh38
rs746051258 Chr.2: 69864915-69864915 on Build GRCh38
rs115076547 Chr.2: 69865064-69865064 on Build GRCh38
rs760131924 Chr.2: 69864999-69865006 on Build GRCh38
rs545860246 Chr.2: 69864958-69864958 on Build GRCh38
rs1035860491 Chr.2: 69865306-69865306 on Build GRCh38
rs1002197866 Chr.2: 69865278-69865278 on Build GRCh38
rs906092659 Chr.2: 69865261-69865261 on Build GRCh38
rs1036278901 Chr.2: 69864975-69864975 on Build GRCh38
rs907529938 Chr.2: 69865034-69865034 on Build GRCh38
rs565554116 Chr.2: 69865060-69865060 on Build GRCh38
rs142388845 Chr.2: 69865015-69865015 on Build GRCh38
rs199902498 Chr.2: 69864909-69864909 on Build GRCh38
rs138299558 Chr.2: 69864963-69864963 on Build GRCh38
rs935895313 Chr.2: 69865307-69865307 on Build GRCh38
rs111934832 Chr.2: 69864875-69864875 on Build GRCh38
rs770013386 Chr.2: 69864929-69864929 on Build GRCh38
rs779177001 Chr.2: 69864867-69864867 on Build GRCh38

View Full Product Details