Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74224762-74225267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC4A5
Entrez Gene ID
57835
Gene Name
solute carrier family 4 member 5
Gene Aliases
NBC4, NBCe2
UniGene
Hs.594099
Gene Chromosome Location
Chr.2: 74216242-74343407 on Build GRCh38
Transcripts
NM_021196.3 NM_133478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613977 c.3219delA p.A1075fs*17 Chr.2: 74224867-74224867 on Build GRCh38
4542931 c.3280G>C p.D1094H Chr.2: 74224854-74224854 on Build GRCh38
4095720 c.3234A>G p.K1078K Chr.2: 74224900-74224900 on Build GRCh38
5862854 c.3181G>A p.E1061K Chr.2: 74224905-74224905 on Build GRCh38
4542862 c.3268G>A p.G1090R Chr.2: 74224866-74224866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753413379 Chr.2: 74224873-74224873 on Build GRCh38
rs142577702 Chr.2: 74225231-74225231 on Build GRCh38
rs746339850 Chr.2: 74224920-74224922 on Build GRCh38
rs770117994 Chr.2: 74225058-74225058 on Build GRCh38
rs766077884 Chr.2: 74224957-74224957 on Build GRCh38
rs754544217 Chr.2: 74224876-74224876 on Build GRCh38
rs1048176805 Chr.2: 74225092-74225092 on Build GRCh38
rs778394945 Chr.2: 74224878-74224878 on Build GRCh38
rs776683823 Chr.2: 74224841-74224841 on Build GRCh38
rs772515122 Chr.2: 74224880-74224880 on Build GRCh38
rs747052137 Chr.2: 74224840-74224840 on Build GRCh38
rs561084407 Chr.2: 74225105-74225105 on Build GRCh38
rs753596923 Chr.2: 74224973-74224973 on Build GRCh38
rs982174264 Chr.2: 74225128-74225128 on Build GRCh38
rs751614394 Chr.2: 74224849-74224849 on Build GRCh38
rs185821227 Chr.2: 74225073-74225073 on Build GRCh38
rs757706193 Chr.2: 74225033-74225033 on Build GRCh38
rs755755685 Chr.2: 74224863-74224863 on Build GRCh38
rs747347622 Chr.2: 74225057-74225057 on Build GRCh38
rs1030511196 Chr.2: 74224842-74224842 on Build GRCh38
rs958164661 Chr.2: 74225071-74225072 on Build GRCh38
rs770270233 Chr.2: 74225195-74225195 on Build GRCh38
rs12476235 Chr.2: 74225200-74225200 on Build GRCh38
rs115649713 Chr.2: 74225244-74225244 on Build GRCh38
rs990761270 Chr.2: 74224897-74224897 on Build GRCh38
rs552107295 Chr.2: 74224798-74224798 on Build GRCh38
rs756736527 Chr.2: 74224839-74224841 on Build GRCh38
rs746151282 Chr.2: 74225053-74225053 on Build GRCh38
rs767523679 Chr.2: 74224937-74224937 on Build GRCh38
rs937925302 Chr.2: 74225170-74225170 on Build GRCh38
rs760825987 Chr.2: 74224831-74224831 on Build GRCh38
rs760570306 Chr.2: 74224941-74224941 on Build GRCh38
rs148029230 Chr.2: 74225113-74225113 on Build GRCh38
rs546607814 Chr.2: 74225196-74225196 on Build GRCh38
rs544313243 Chr.2: 74225246-74225246 on Build GRCh38
rs761775516 Chr.2: 74224850-74224850 on Build GRCh38
rs981963102 Chr.2: 74224966-74224966 on Build GRCh38
rs942368322 Chr.2: 74225127-74225127 on Build GRCh38
rs781179299 Chr.2: 74225020-74225020 on Build GRCh38
rs780757230 Chr.2: 74225041-74225042 on Build GRCh38
rs900803799 Chr.2: 74225166-74225166 on Build GRCh38
rs967240951 Chr.2: 74224843-74224843 on Build GRCh38
rs143616530 Chr.2: 74224858-74224858 on Build GRCh38
rs753895866 Chr.2: 74225035-74225035 on Build GRCh38
rs888159136 Chr.2: 74224788-74224788 on Build GRCh38
rs997145829 Chr.2: 74224990-74224990 on Build GRCh38
rs1032449775 Chr.2: 74225241-74225241 on Build GRCh38
rs776967623 Chr.2: 74224903-74224903 on Build GRCh38
rs779798842 Chr.2: 74224867-74224867 on Build GRCh38
rs778293501 Chr.2: 74224897-74224898 on Build GRCh38
rs900738267 Chr.2: 74224821-74224821 on Build GRCh38
rs828863 Chr.2: 74225132-74225132 on Build GRCh38
rs781343801 Chr.2: 74225045-74225045 on Build GRCh38
rs371252754 Chr.2: 74224808-74224808 on Build GRCh38
rs773503778 Chr.2: 74224830-74224830 on Build GRCh38
rs747456308 Chr.2: 74225031-74225031 on Build GRCh38
rs772993510 Chr.2: 74224939-74224939 on Build GRCh38
rs778391845 Chr.2: 74225010-74225010 on Build GRCh38
rs764745698 Chr.2: 74224978-74224978 on Build GRCh38
rs887172937 Chr.2: 74225230-74225230 on Build GRCh38
rs768126471 Chr.2: 74224817-74224817 on Build GRCh38
rs966006327 Chr.2: 74224981-74224981 on Build GRCh38
rs990714198 Chr.2: 74225184-74225184 on Build GRCh38
rs147834537 Chr.2: 74224857-74224857 on Build GRCh38
rs916559861 Chr.2: 74224915-74224915 on Build GRCh38
rs761865301 Chr.2: 74224922-74224922 on Build GRCh38
rs149322613 Chr.2: 74224820-74224820 on Build GRCh38
rs748644673 Chr.2: 74224822-74224822 on Build GRCh38
rs747438592 Chr.2: 74224886-74224886 on Build GRCh38
rs887204172 Chr.2: 74224892-74224892 on Build GRCh38
rs772730495 Chr.2: 74224978-74224980 on Build GRCh38
rs1047614026 Chr.2: 74225228-74225228 on Build GRCh38
rs767444223 Chr.2: 74224942-74224942 on Build GRCh38
rs144930405 Chr.2: 74224977-74224977 on Build GRCh38
rs749409054 Chr.2: 74225061-74225061 on Build GRCh38
rs145427733 Chr.2: 74224949-74224949 on Build GRCh38
rs752159341 Chr.2: 74224988-74224988 on Build GRCh38
rs563412053 Chr.2: 74224813-74224814 on Build GRCh38
rs150255670 Chr.2: 74225078-74225078 on Build GRCh38
rs754562657 Chr.2: 74224974-74224974 on Build GRCh38
rs549286065 Chr.2: 74224828-74224828 on Build GRCh38
rs997770159 Chr.2: 74225182-74225182 on Build GRCh38
rs140560393 Chr.2: 74224846-74224846 on Build GRCh38
rs757870254 Chr.2: 74224996-74224996 on Build GRCh38
rs775751231 Chr.2: 74225060-74225060 on Build GRCh38
rs771141483 Chr.2: 74224835-74224835 on Build GRCh38
rs778027532 Chr.2: 74224882-74224882 on Build GRCh38
rs571302530 Chr.2: 74224911-74224911 on Build GRCh38
rs775653246 Chr.2: 74224912-74224912 on Build GRCh38
rs180943938 Chr.2: 74224813-74224813 on Build GRCh38
rs759832977 Chr.2: 74224906-74224906 on Build GRCh38
rs779453628 Chr.2: 74224809-74224809 on Build GRCh38

View Full Product Details