Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102201268-102201767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RL2
Entrez Gene ID
8808
Gene Name
interleukin 1 receptor like 2
Gene Aliases
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2
UniGene
Hs.659863
Gene Chromosome Location
Chr.2: 102186973-102242906 on Build GRCh38
Transcripts
NM_003854.2 XM_005264035.1 XM_005264036.1 XM_006712820.2 XM_006712822.2 XM_011512091.1 XM_011512092.1 XM_011512093.1 XM_011512094.1 XM_011512096.1 XM_017005174.1 XM_017005175.1 XM_017005176.1 XM_017005177.1 XM_017005178.1 XM_017005179.1 XM_017005180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1004634 c.641T>A p.V214E Chr.2: 102201707-102201707 on Build GRCh38
1004631 c.264G>A p.Q88Q Chr.2: 102201681-102201681 on Build GRCh38
5394017 c.222G>A p.G74G Chr.2: 102201639-102201639 on Build GRCh38
5394016 c.573G>A p.G191G Chr.2: 102201639-102201639 on Build GRCh38
5394019 c.256G>A p.G86R Chr.2: 102201673-102201673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751108610 Chr.2: 102201602-102201611 on Build GRCh38
rs909535950 Chr.2: 102201609-102201609 on Build GRCh38
rs749581969 Chr.2: 102201574-102201574 on Build GRCh38
rs969575347 Chr.2: 102201350-102201350 on Build GRCh38
rs888128627 Chr.2: 102201349-102201350 on Build GRCh38
rs549627572 Chr.2: 102201425-102201425 on Build GRCh38
rs1011028511 Chr.2: 102201346-102201346 on Build GRCh38
rs772104647 Chr.2: 102201733-102201733 on Build GRCh38
rs576789803 Chr.2: 102201669-102201669 on Build GRCh38
rs115648216 Chr.2: 102201364-102201364 on Build GRCh38
rs138237710 Chr.2: 102201337-102201338 on Build GRCh38
rs770162134 Chr.2: 102201497-102201497 on Build GRCh38
rs193263994 Chr.2: 102201347-102201347 on Build GRCh38
rs771636754 Chr.2: 102201473-102201473 on Build GRCh38
rs751982557 Chr.2: 102201525-102201525 on Build GRCh38
rs986563671 Chr.2: 102201374-102201374 on Build GRCh38
rs780250200 Chr.2: 102201568-102201568 on Build GRCh38
rs76375740 Chr.2: 102201647-102201647 on Build GRCh38
rs760475039 Chr.2: 102201588-102201588 on Build GRCh38
rs897574347 Chr.2: 102201577-102201577 on Build GRCh38
rs750996139 Chr.2: 102201635-102201635 on Build GRCh38
rs985355887 Chr.2: 102201454-102201457 on Build GRCh38
rs747709241 Chr.2: 102201696-102201697 on Build GRCh38
rs144973039 Chr.2: 102201623-102201623 on Build GRCh38
rs559013846 Chr.2: 102201351-102201351 on Build GRCh38
rs1054641754 Chr.2: 102201316-102201316 on Build GRCh38
rs998939832 Chr.2: 102201314-102201314 on Build GRCh38
rs780802208 Chr.2: 102201689-102201689 on Build GRCh38
rs911021975 Chr.2: 102201313-102201314 on Build GRCh38
rs777843950 Chr.2: 102201662-102201662 on Build GRCh38
rs746996241 Chr.2: 102201664-102201664 on Build GRCh38
rs10164790 Chr.2: 102201402-102201402 on Build GRCh38
rs200534980 Chr.2: 102201564-102201564 on Build GRCh38
rs778012541 Chr.2: 102201409-102201409 on Build GRCh38
rs189640693 Chr.2: 102201589-102201589 on Build GRCh38
rs893382186 Chr.2: 102201324-102201324 on Build GRCh38
rs766823741 Chr.2: 102201739-102201739 on Build GRCh38
rs776128896 Chr.2: 102201610-102201610 on Build GRCh38
rs924663291 Chr.2: 102201444-102201444 on Build GRCh38
rs368217225 Chr.2: 102201723-102201723 on Build GRCh38
rs748205889 Chr.2: 102201581-102201581 on Build GRCh38
rs948985916 Chr.2: 102201523-102201523 on Build GRCh38
rs745675706 Chr.2: 102201675-102201675 on Build GRCh38
rs918375082 Chr.2: 102201338-102201338 on Build GRCh38
rs369721985 Chr.2: 102201740-102201740 on Build GRCh38
rs758960247 Chr.2: 102201508-102201508 on Build GRCh38
rs2302624 Chr.2: 102201373-102201373 on Build GRCh38
rs763652307 Chr.2: 102201703-102201703 on Build GRCh38
rs184118101 Chr.2: 102201580-102201580 on Build GRCh38
rs961876930 Chr.2: 102201363-102201363 on Build GRCh38
rs376758029 Chr.2: 102201621-102201621 on Build GRCh38
rs767946557 Chr.2: 102201633-102201633 on Build GRCh38
rs1027610893 Chr.2: 102201365-102201365 on Build GRCh38
rs1006407059 Chr.2: 102201349-102201349 on Build GRCh38
rs367688415 Chr.2: 102201548-102201548 on Build GRCh38
rs138453799 Chr.2: 102201673-102201673 on Build GRCh38
rs772278646 Chr.2: 102201656-102201656 on Build GRCh38
rs145899905 Chr.2: 102201648-102201648 on Build GRCh38
rs1034746594 Chr.2: 102201359-102201359 on Build GRCh38
rs944680112 Chr.2: 102201406-102201406 on Build GRCh38
rs748198462 Chr.2: 102201652-102201652 on Build GRCh38
rs560493967 Chr.2: 102201465-102201465 on Build GRCh38
rs752203916 Chr.2: 102201624-102201624 on Build GRCh38
rs754710750 Chr.2: 102201637-102201637 on Build GRCh38
rs575876084 Chr.2: 102201360-102201360 on Build GRCh38
rs372158050 Chr.2: 102201645-102201645 on Build GRCh38
rs373907400 Chr.2: 102201741-102201741 on Build GRCh38
rs781681881 Chr.2: 102201559-102201559 on Build GRCh38
rs1003582671 Chr.2: 102201555-102201555 on Build GRCh38
rs2302623 Chr.2: 102201494-102201494 on Build GRCh38
rs531626294 Chr.2: 102201582-102201582 on Build GRCh38
rs983856365 Chr.2: 102201600-102201600 on Build GRCh38
rs756476630 Chr.2: 102201565-102201565 on Build GRCh38
rs761209838 Chr.2: 102201729-102201729 on Build GRCh38
rs934865001 Chr.2: 102201453-102201453 on Build GRCh38
rs958042723 Chr.2: 102201446-102201446 on Build GRCh38
rs142080534 Chr.2: 102201646-102201646 on Build GRCh38
rs764855909 Chr.2: 102201524-102201524 on Build GRCh38
rs376417306 Chr.2: 102201696-102201696 on Build GRCh38
rs769431595 Chr.2: 102201684-102201684 on Build GRCh38
rs553419509 Chr.2: 102201685-102201685 on Build GRCh38

View Full Product Details